Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009135.hg.1 |
ENST00000320393.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009135.hg.1 |
ENST00000356489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009135.hg.1 |
ENST00000369760.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009135.hg.1 |
ENST00000541767.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009135.hg.1 |
ENST00000545529.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009135.hg.1 |
NM_000056.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009135.hg.1 |
NM_183050.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009135.hg.1 |
TC06000762.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
ENST00000320393.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
ENST00000320393.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
ENST00000320393.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
ENST00000320393.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
ENST00000320393.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
ENST00000320393.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
ENST00000320393.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
ENST00000320393.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
ENST00000320393.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
ENST00000320393.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
ENST00000320393.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
ENST00000320393.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
ENST00000320393.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
ENST00000356489.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
ENST00000356489.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
ENST00000356489.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
ENST00000356489.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
ENST00000356489.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
ENST00000356489.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
ENST00000356489.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
ENST00000356489.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
ENST00000356489.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
ENST00000356489.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
ENST00000356489.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
ENST00000356489.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
ENST00000369760.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
ENST00000369760.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
ENST00000369760.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
ENST00000369760.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
ENST00000369760.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
ENST00000369760.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
ENST00000468520.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
ENST00000468520.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
ENST00000468520.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
ENST00000486968.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
ENST00000486968.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
ENST00000486968.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
ENST00000486968.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
ENST00000491328.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
ENST00000491328.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
ENST00000491328.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
NM_000056.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
NM_000056.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
NM_000056.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
NM_000056.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
NM_000056.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
NM_000056.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
NM_000056.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
NM_000056.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
NM_000056.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
NM_000056.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
NM_000056.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
NM_000056.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
NM_183050.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
NM_183050.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
NM_183050.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
NM_183050.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
NM_183050.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
NM_183050.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
NM_183050.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
NM_183050.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
NM_183050.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
NM_183050.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
NM_183050.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
NM_183050.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008632.hg.1 |
NM_183050.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
ENST00000320393.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
ENST00000320393.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
ENST00000320393.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
ENST00000320393.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
ENST00000320393.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
ENST00000320393.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
ENST00000320393.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
ENST00000320393.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
ENST00000320393.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
ENST00000320393.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
ENST00000320393.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
ENST00000320393.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
ENST00000320393.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
ENST00000356489.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
ENST00000356489.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
ENST00000356489.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
ENST00000356489.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
ENST00000356489.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
ENST00000356489.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
ENST00000356489.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
ENST00000356489.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
ENST00000356489.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
ENST00000356489.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
ENST00000356489.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
ENST00000356489.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
ENST00000369760.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
ENST00000369760.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
ENST00000369760.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
ENST00000369760.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
ENST00000369760.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
ENST00000369760.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
ENST00000468520.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
ENST00000468520.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
ENST00000468520.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
ENST00000486968.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
ENST00000486968.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
ENST00000486968.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
ENST00000486968.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
ENST00000491328.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
ENST00000491328.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
ENST00000491328.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
NM_000056.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
NM_000056.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
NM_000056.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
NM_000056.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
NM_000056.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
NM_000056.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
NM_000056.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
NM_000056.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
NM_000056.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
NM_000056.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
NM_000056.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
NM_000056.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
NM_183050.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
NM_183050.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
NM_183050.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
NM_183050.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
NM_183050.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
NM_183050.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
NM_183050.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
NM_183050.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
NM_183050.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
NM_183050.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
NM_183050.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
NM_183050.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008632.hg.1 |
NM_183050.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600159948.hg.1 |
ENST00000320393.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159948.hg.1 |
ENST00000356489.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159948.hg.1 |
ENST00000369760.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159948.hg.1 |
NM_000056.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600159948.hg.1 |
NM_183050.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600159948.hg.1 |
ENST00000320393.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159948.hg.1 |
ENST00000356489.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159948.hg.1 |
ENST00000369760.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159948.hg.1 |
NM_000056.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600159948.hg.1 |
NM_183050.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600159948.hg.1 |
TC0600008632.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600159949.hg.1 |
ENST00000369760.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159949.hg.1 |
ENST00000369760.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159949.hg.1 |
TC0600008632.hg.1 |
Clariom D TC |