This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00609049_m1
See other BCKDHB GE Assays ›
Gene Symbol:
BCKDHB
Gene Name:
branched chain keto acid dehydrogenase E1 subunit beta
Gene Aliases:
BCKDE1B, BCKDH E1-beta, E1B
Chromosome Location:
Chr.6: 80106610 - 80469088 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00609049_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BCKDHB
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000056.4 NP_000047.1 2 - 3 358 111
NM_183050.3 NP_898871.1 2 - 3 358 111
NR_134945.1 2 - 3 358 111
XM_005248756.4 XP_005248813.1 2 - 3 307 111
XM_011536023.2 XP_011534325.1 2 - 3 307 111
XM_011536024.2 XP_011534326.1 2 - 3 307 111
XM_011536025.2 XP_011534327.1 2 - 3 307 111
GenBank mRNA AK289977.1 - 2 - 3 287 111
AK316545.1 - 3 - 4 271 111
BC034481.1 - 2 - 3 302 4824846 111
BC040139.2 - 2 - 3 279 5493159 111
BT020063.1 - 2 - 3 274 111
DC372349.1 - 2 - 3 346 111
DC422376.1 - 2 - 3 361 111
M55575.1 - 2 - 3 278 111
U50708.1 - 2 - 3 321 111
X52446.1 - 2 - 3 217 111

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0600008632.hg.1
This assay targets the same location as HTA transcript cluster(s) TC06000762.hg.1

Additional Information:

This assay is in the 5'UTR region of the AK316545.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene BCKDHB

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

branched-chain amino acid catabolic process
glyoxylate metabolic process
oxidation-reduction process
alpha-ketoacid dehydrogenase activity
3-methyl-2-oxobutanoate dehydrogenase (2-methylpropanoyl-transferring) activity
protein binding
carboxy-lyase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 ENST00000320393.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 ENST00000320393.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 ENST00000320393.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 ENST00000320393.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 ENST00000320393.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 ENST00000320393.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 ENST00000320393.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 ENST00000320393.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 ENST00000320393.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 ENST00000320393.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 ENST00000320393.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 ENST00000320393.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 ENST00000320393.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 ENST00000356489.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 ENST00000356489.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 ENST00000356489.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 ENST00000356489.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 ENST00000356489.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 ENST00000356489.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 ENST00000356489.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 ENST00000356489.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 ENST00000356489.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 ENST00000356489.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 ENST00000356489.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 ENST00000356489.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 ENST00000369760.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 ENST00000369760.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 ENST00000369760.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 ENST00000369760.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 ENST00000369760.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 ENST00000369760.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 ENST00000468520.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 ENST00000468520.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 ENST00000468520.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 ENST00000486968.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 ENST00000486968.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 ENST00000486968.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 ENST00000486968.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 ENST00000491328.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 ENST00000491328.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 ENST00000491328.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 NM_000056.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 NM_000056.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 NM_000056.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 NM_000056.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 NM_000056.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 NM_000056.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 NM_000056.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 NM_000056.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 NM_000056.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 NM_000056.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 NM_000056.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 NM_000056.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 NM_183050.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 NM_183050.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 NM_183050.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 NM_183050.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 NM_183050.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 NM_183050.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 NM_183050.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 NM_183050.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 NM_183050.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 NM_183050.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 NM_183050.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 NM_183050.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008632.hg.1 NM_183050.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06009132.hg.1 ENST00000320393.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06009132.hg.1 ENST00000356489.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06009132.hg.1 ENST00000369760.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06009132.hg.1 ENST00000541767.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06009132.hg.1 ENST00000545529.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06009132.hg.1 NM_000056.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06009132.hg.1 NM_183050.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06009132.hg.1 TC06000762.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC0600008632.hg.1 ENST00000320393.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008632.hg.1 ENST00000320393.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008632.hg.1 ENST00000320393.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008632.hg.1 ENST00000320393.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008632.hg.1 ENST00000320393.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008632.hg.1 ENST00000320393.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008632.hg.1 ENST00000320393.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008632.hg.1 ENST00000320393.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008632.hg.1 ENST00000320393.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008632.hg.1 ENST00000320393.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008632.hg.1 ENST00000320393.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008632.hg.1 ENST00000320393.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008632.hg.1 ENST00000320393.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008632.hg.1 ENST00000356489.9 Ensembl
Affymetrix Human Clariom D Assay TC0600008632.hg.1 ENST00000356489.9 Ensembl
Affymetrix Human Clariom D Assay TC0600008632.hg.1 ENST00000356489.9 Ensembl
Affymetrix Human Clariom D Assay TC0600008632.hg.1 ENST00000356489.9 Ensembl
Affymetrix Human Clariom D Assay TC0600008632.hg.1 ENST00000356489.9 Ensembl
Affymetrix Human Clariom D Assay TC0600008632.hg.1 ENST00000356489.9 Ensembl
Affymetrix Human Clariom D Assay TC0600008632.hg.1 ENST00000356489.9 Ensembl
Affymetrix Human Clariom D Assay TC0600008632.hg.1 ENST00000356489.9 Ensembl
Affymetrix Human Clariom D Assay TC0600008632.hg.1 ENST00000356489.9 Ensembl
Affymetrix Human Clariom D Assay TC0600008632.hg.1 ENST00000356489.9 Ensembl
Affymetrix Human Clariom D Assay TC0600008632.hg.1 ENST00000356489.9 Ensembl
Affymetrix Human Clariom D Assay TC0600008632.hg.1 ENST00000356489.9 Ensembl
Affymetrix Human Clariom D Assay TC0600008632.hg.1 ENST00000369760.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008632.hg.1 ENST00000369760.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008632.hg.1 ENST00000369760.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008632.hg.1 ENST00000369760.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008632.hg.1 ENST00000369760.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008632.hg.1 ENST00000369760.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008632.hg.1 ENST00000468520.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008632.hg.1 ENST00000468520.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008632.hg.1 ENST00000468520.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008632.hg.1 ENST00000486968.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008632.hg.1 ENST00000486968.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008632.hg.1 ENST00000486968.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008632.hg.1 ENST00000486968.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008632.hg.1 ENST00000491328.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008632.hg.1 ENST00000491328.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008632.hg.1 ENST00000491328.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008632.hg.1 NM_000056.4 Refseq
Affymetrix Human Clariom D Assay TC0600008632.hg.1 NM_000056.4 Refseq
Affymetrix Human Clariom D Assay TC0600008632.hg.1 NM_000056.4 Refseq
Affymetrix Human Clariom D Assay TC0600008632.hg.1 NM_000056.4 Refseq
Affymetrix Human Clariom D Assay TC0600008632.hg.1 NM_000056.4 Refseq
Affymetrix Human Clariom D Assay TC0600008632.hg.1 NM_000056.4 Refseq
Affymetrix Human Clariom D Assay TC0600008632.hg.1 NM_000056.4 Refseq
Affymetrix Human Clariom D Assay TC0600008632.hg.1 NM_000056.4 Refseq
Affymetrix Human Clariom D Assay TC0600008632.hg.1 NM_000056.4 Refseq
Affymetrix Human Clariom D Assay TC0600008632.hg.1 NM_000056.4 Refseq
Affymetrix Human Clariom D Assay TC0600008632.hg.1 NM_000056.4 Refseq
Affymetrix Human Clariom D Assay TC0600008632.hg.1 NM_000056.4 Refseq
Affymetrix Human Clariom D Assay TC0600008632.hg.1 NM_183050.3 Refseq
Affymetrix Human Clariom D Assay TC0600008632.hg.1 NM_183050.3 Refseq
Affymetrix Human Clariom D Assay TC0600008632.hg.1 NM_183050.3 Refseq
Affymetrix Human Clariom D Assay TC0600008632.hg.1 NM_183050.3 Refseq
Affymetrix Human Clariom D Assay TC0600008632.hg.1 NM_183050.3 Refseq
Affymetrix Human Clariom D Assay TC0600008632.hg.1 NM_183050.3 Refseq
Affymetrix Human Clariom D Assay TC0600008632.hg.1 NM_183050.3 Refseq
Affymetrix Human Clariom D Assay TC0600008632.hg.1 NM_183050.3 Refseq
Affymetrix Human Clariom D Assay TC0600008632.hg.1 NM_183050.3 Refseq
Affymetrix Human Clariom D Assay TC0600008632.hg.1 NM_183050.3 Refseq
Affymetrix Human Clariom D Assay TC0600008632.hg.1 NM_183050.3 Refseq
Affymetrix Human Clariom D Assay TC0600008632.hg.1 NM_183050.3 Refseq
Affymetrix Human Clariom D Assay TC0600008632.hg.1 NM_183050.3 Refseq
Affymetrix Human Clariom D Assay PSR0600159945.hg.1 ENST00000320393.8 Ensembl
Affymetrix Human Clariom D Assay PSR0600159945.hg.1 ENST00000356489.9 Ensembl
Affymetrix Human Clariom D Assay PSR0600159945.hg.1 ENST00000369760.8 Ensembl
Affymetrix Human Clariom D Assay PSR0600159945.hg.1 ENST00000486968.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600159945.hg.1 NM_000056.4 Refseq
Affymetrix Human Clariom D Assay PSR0600159945.hg.1 NM_183050.3 Refseq
Affymetrix Human Clariom D Assay PSR0600159945.hg.1 ENST00000320393.8 Ensembl
Affymetrix Human Clariom D Assay PSR0600159945.hg.1 ENST00000356489.9 Ensembl
Affymetrix Human Clariom D Assay PSR0600159945.hg.1 ENST00000369760.8 Ensembl
Affymetrix Human Clariom D Assay PSR0600159945.hg.1 ENST00000486968.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600159945.hg.1 NM_000056.4 Refseq
Affymetrix Human Clariom D Assay PSR0600159945.hg.1 NM_183050.3 Refseq
Affymetrix Human Clariom D Assay PSR0600159945.hg.1 TC0600008632.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0