This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00560002_m1
See other XRN2 GE Assays ›
Gene Symbol:
XRN2
Gene Name:
5'-3' exoribonuclease 2
Gene Aliases:
-
Chromosome Location:
Chr.20: 21303284 - 21389825 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00560002_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: XRN2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001317960.1 NP_001304889.1 21 - 22 2298 86
NM_012255.4 NP_036387.2 21 - 22 2138 86
XM_017027723.1 XP_016883212.1 21 - 22 2091 86
GenBank mRNA AF064257.1 - 21 - 22 2091 86
AF152169.1 - 21 - 22 2041 86
AK000084.1 - 10 - 11 953 86
AK001573.1 - 21 - 22 2108 86
AK302846.1 - 21 - 22 2002 86
AK303312.1 - 20 - 21 1949 86
AK314602.1 - 21 - 22 2077 86
AL136841.1 - 21 - 22 2064 86
AY382900.1 - 20 - 21 1953 86
BC006417.1 - 9 - 10 913 86

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2000006899.hg.1
This assay targets the same location as HTA transcript cluster(s) TC20000140.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene XRN2

Panther Classification:

Molecular Function -

exoribonuclease

Gene Ontology Categories:

Function(s) Process(es)

DNA catabolic process, exonucleolytic
DNA-templated transcription, termination
regulation of transcription, DNA-templated
rRNA processing
RNA processing
mRNA processing
RNA catabolic process
spermatogenesis
cell growth
RNA metabolic process
RNA phosphodiester bond hydrolysis, exonucleolytic
3'-5'-exoribonuclease activity
transcription termination site sequence-specific DNA binding
nuclease activity
5'-3' exoribonuclease activity
protein binding
5'-3' exonuclease activity
poly(A) RNA binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR20001884.hg.1 ENST00000377191.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20001884.hg.1 ENST00000430571.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20001884.hg.1 ENST00000539513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20001884.hg.1 NM_012255.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR20001884.hg.1 TC20000140.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR20001883.hg.1 ENST00000377191.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20001883.hg.1 ENST00000430571.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20001883.hg.1 ENST00000539513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20001883.hg.1 NM_012255.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR20001883.hg.1 TC20000140.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Clariom D Assay TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Clariom D Assay TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Clariom D Assay TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Clariom D Assay TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Clariom D Assay TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Clariom D Assay TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Clariom D Assay TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Clariom D Assay TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Clariom D Assay TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Clariom D Assay TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Clariom D Assay TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Clariom D Assay TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Clariom D Assay TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Clariom D Assay TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Clariom D Assay TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Clariom D Assay TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Clariom D Assay TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Clariom D Assay TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Clariom D Assay TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Clariom D Assay TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Clariom D Assay TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Clariom D Assay TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Clariom D Assay TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Clariom D Assay TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Clariom D Assay TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Clariom D Assay TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Clariom D Assay TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Clariom D Assay TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Clariom D Assay TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Clariom D Assay TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Clariom D Assay TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Clariom D Assay TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Clariom D Assay TC2000006899.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Clariom D Assay TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Clariom D Assay TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Clariom D Assay TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Clariom D Assay TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Clariom D Assay TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Clariom D Assay TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Clariom D Assay TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Clariom D Assay TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Clariom D Assay TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Clariom D Assay TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Clariom D Assay TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Clariom D Assay TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Clariom D Assay TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Clariom D Assay TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Clariom D Assay TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Clariom D Assay TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Clariom D Assay TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Clariom D Assay TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Clariom D Assay TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Clariom D Assay TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Clariom D Assay TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Clariom D Assay TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Clariom D Assay TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Clariom D Assay TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Clariom D Assay TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Clariom D Assay TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Clariom D Assay TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Clariom D Assay TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Clariom D Assay TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Clariom D Assay TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Clariom D Assay TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Clariom D Assay TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Clariom D Assay TC2000006899.hg.1 NM_012255.4 Refseq
Affymetrix Human Clariom D Assay PSR2000147260.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Clariom D Assay PSR2000147260.hg.1 NM_012255.4 Refseq
Affymetrix Human Clariom D Assay PSR2000147260.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Clariom D Assay PSR2000147260.hg.1 NM_012255.4 Refseq
Affymetrix Human Clariom D Assay PSR2000147260.hg.1 TC2000006899.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2000147261.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Clariom D Assay PSR2000147261.hg.1 NM_012255.4 Refseq
Affymetrix Human Clariom D Assay PSR2000147261.hg.1 ENST00000377191.4 Ensembl
Affymetrix Human Clariom D Assay PSR2000147261.hg.1 NM_012255.4 Refseq
Affymetrix Human Clariom D Assay PSR2000147261.hg.1 TC2000006899.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0