Assay Details
Gene Symbol: PSMB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004159.4 | NP_004150.1 | 2 - 3 | 811 | 72 | |
NM_148919.3 | NP_683720.2 | 2 - 3 | 344 | 72 | ||
GenBank mRNA | BC001114.2 | - | 2 - 3 | 527 | 2967119 | 72 |
BG547344.1 | - | 2 - 3 | 339 | 72 | ||
BG760674.1 | - | 2 - 3 | 343 | 72 | ||
BM547457.1 | - | 2 - 3 | 581 | 72 | ||
BM781620.1 | - | 2 - 3 | 344 | 72 | ||
CR541661.1 | - | 2 - 3 | 289 | 72 | ||
U17496.1 | - | 2 - 3 | 329 | 72 | ||
U17497.1 | - | 2 - 3 | 329 | 72 | ||
X62598.1 | - | 2 - 3 | 509 | 72 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06001573.hg.1, TC6_apd_hap1000104.hg.1, TC6_cox_hap2000198.hg.1, TC6_dbb_hap3000184.hg.1, TC6_mann_hap4000166.hg.1, TC6_mcf_hap5000173.hg.1, TC6_qbl_hap6000186.hg.1, TC6_ssto_hap7000168.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
protein polyubiquitination
stimulatory C-type lectin receptor signaling pathway
antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-dependent
regulation of cellular amino acid metabolic process
viral process
anaphase-promoting complex-dependent catabolic process
tumor necrosis factor-mediated signaling pathway
NIK/NF-kappaB signaling
Fc-epsilon receptor signaling pathway
proteasome-mediated ubiquitin-dependent protein catabolic process
regulation of mRNA stability
fat cell differentiation
T cell receptor signaling pathway
negative regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
positive regulation of ubiquitin-protein ligase activity involved in regulation of mitotic cell cycle transition
regulation of endopeptidase activity
Wnt signaling pathway, planar cell polarity pathway
type I interferon signaling pathway
negative regulation of canonical Wnt signaling pathway
positive regulation of canonical Wnt signaling pathway
threonine-type endopeptidase activity
protein binding
MAPK cascade
protein polyubiquitination
stimulatory C-type lectin receptor signaling pathway
antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-dependent
regulation of cellular amino acid metabolic process
viral process
anaphase-promoting complex-dependent catabolic process
tumor necrosis factor-mediated signaling pathway
NIK/NF-kappaB signaling
Fc-epsilon receptor signaling pathway
proteasome-mediated ubiquitin-dependent protein catabolic process
regulation of mRNA stability
fat cell differentiation
T cell receptor signaling pathway
negative regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
positive regulation of ubiquitin-protein ligase activity involved in regulation of mitotic cell cycle transition
regulation of endopeptidase activity
Wnt signaling pathway, planar cell polarity pathway
type I interferon signaling pathway
negative regulation of canonical Wnt signaling pathway
positive regulation of canonical Wnt signaling pathway
threonine-type endopeptidase activity
protein binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR06018783.hg.1 | ENST00000374881.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06018783.hg.1 | ENST00000374882.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06018783.hg.1 | NM_004159.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06018783.hg.1 | NM_148919.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06018783.hg.1 | TC06001573.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2003930.hg.1 | NM_148919.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2003930.hg.1 | TC6_cox_hap2000198.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3003679.hg.1 | NM_004159.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3003679.hg.1 | NM_148919.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3003679.hg.1 | TC6_dbb_hap3000184.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4003033.hg.1 | NM_148919.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4003033.hg.1 | TC6_mann_hap4000166.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3003685.hg.1 | NM_148919.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3003685.hg.1 | TC6_dbb_hap3000184.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4003030.hg.1 | NM_004159.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4003030.hg.1 | NM_148919.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4003030.hg.1 | TC6_mann_hap4000166.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6003741.hg.1 | NM_004159.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6003741.hg.1 | NM_148919.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6003741.hg.1 | TC6_qbl_hap6000186.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06018786.hg.1 | ENST00000374882.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06018786.hg.1 | ENST00000395339.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06018786.hg.1 | NM_148919.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06018786.hg.1 | TC06001573.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2003927.hg.1 | NM_004159.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2003927.hg.1 | NM_148919.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2003927.hg.1 | TC6_cox_hap2000198.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5003566.hg.1 | NM_004159.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5003566.hg.1 | NM_148919.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5003566.hg.1 | TC6_mcf_hap5000173.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5003569.hg.1 | NM_148919.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5003569.hg.1 | TC6_mcf_hap5000173.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1001737.hg.1 | NM_148919.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1001737.hg.1 | TC6_apd_hap1000104.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6003744.hg.1 | NM_148919.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6003744.hg.1 | TC6_qbl_hap6000186.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7003503.hg.1 | NM_148919.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7003503.hg.1 | TC6_ssto_hap7000168.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3003684.hg.1 | NM_148919.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3003684.hg.1 | TC6_dbb_hap3000184.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5003565.hg.1 | NM_004159.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5003565.hg.1 | NM_148919.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5003565.hg.1 | TC6_mcf_hap5000173.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | ENST00000374881.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | ENST00000374881.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | ENST00000374881.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | ENST00000374881.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | ENST00000374881.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | ENST00000374881.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | ENST00000374881.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | ENST00000374881.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | ENST00000374881.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | ENST00000374881.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | ENST00000374881.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | ENST00000374882.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | ENST00000374882.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | ENST00000374882.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | ENST00000374882.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | ENST00000374882.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | ENST00000374882.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | ENST00000374882.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | ENST00000374882.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | ENST00000374882.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | ENST00000395339.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | ENST00000395339.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | ENST00000395339.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | ENST00000395339.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | ENST00000395339.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | ENST00000395339.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | ENST00000395339.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | ENST00000395339.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | ENST00000484003.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | ENST00000484003.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | ENST00000484003.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | ENST00000484003.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | ENST00000484003.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | ENST00000484003.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | ENST00000484003.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | ENST00000484003.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | ENST00000484003.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | ENST00000490613.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | ENST00000490613.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | ENST00000490613.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | ENST00000490613.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | ENST00000490613.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | NM_148919.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | NM_148919.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | NM_148919.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | NM_148919.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | NM_148919.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | NM_148919.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | NM_148919.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | NM_148919.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011507.hg.1 | NM_148919.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1001734.hg.1 | NM_004159.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1001734.hg.1 | NM_148919.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1001734.hg.1 | TC6_apd_hap1000104.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7003500.hg.1 | NM_004159.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7003500.hg.1 | NM_148919.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7003500.hg.1 | TC6_ssto_hap7000168.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR0600178572.hg.1 | ENST00000374881.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600178572.hg.1 | ENST00000374882.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600178572.hg.1 | ENST00000484003.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600178572.hg.1 | NM_148919.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600178572.hg.1 | ENST00000374881.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600178572.hg.1 | ENST00000374882.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600178572.hg.1 | ENST00000484003.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600178572.hg.1 | NM_148919.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600178572.hg.1 | TC0600011507.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | ENST00000374881.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | ENST00000374881.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | ENST00000374881.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | ENST00000374881.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | ENST00000374881.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | ENST00000374881.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | ENST00000374881.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | ENST00000374881.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | ENST00000374881.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | ENST00000374881.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | ENST00000374881.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | ENST00000374882.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | ENST00000374882.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | ENST00000374882.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | ENST00000374882.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | ENST00000374882.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | ENST00000374882.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | ENST00000374882.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | ENST00000374882.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | ENST00000374882.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | ENST00000395339.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | ENST00000395339.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | ENST00000395339.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | ENST00000395339.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | ENST00000395339.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | ENST00000395339.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | ENST00000395339.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | ENST00000395339.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | ENST00000484003.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | ENST00000484003.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | ENST00000484003.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | ENST00000484003.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | ENST00000484003.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | ENST00000484003.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | ENST00000484003.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | ENST00000484003.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | ENST00000484003.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | ENST00000490613.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | ENST00000490613.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | ENST00000490613.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | ENST00000490613.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | ENST00000490613.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | NM_148919.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | NM_148919.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | NM_148919.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | NM_148919.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | NM_148919.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | NM_148919.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | NM_148919.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | NM_148919.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011507.hg.1 | NM_148919.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600178575.hg.1 | ENST00000374882.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600178575.hg.1 | ENST00000395339.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600178575.hg.1 | NM_148919.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600178575.hg.1 | ENST00000374882.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600178575.hg.1 | ENST00000395339.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600178575.hg.1 | NM_148919.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600178575.hg.1 | TC0600011507.hg.1 | Clariom D TC |
