This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00544391_m1
See other ARHGAP15 GE Assays ›
Gene Symbol:
ARHGAP15
Gene Name:
Rho GTPase activating protein 15
Gene Aliases:
BM046
Chromosome Location:
Chr.2: 143070973 - 143768352 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00544391_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ARHGAP15
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_018460.3 NP_060930.3 2 - 3 312 86
XM_011511479.2 XP_011509781.1 3 - 4 344 86
XM_011511481.2 XP_011509783.1 2 - 3 247 86
XM_011511482.2 XP_011509784.1 2 - 3 247 86
XM_011511484.1 XP_011509786.1 2 - 3 247 86
XM_017004499.1 XP_016859988.1 2 - 3 273 86
XM_017004501.1 XP_016859990.1 2 - 3 247 86
GenBank mRNA AF212222.1 - 2 - 3 250 86
AF217507.1 - 2 - 3 249 86
AF217522.1 - 2 - 3 249 86
AK226014.1 - 2 - 3 247 86
AK303490.1 - 2 - 3 230 86
AK308219.1 - 2 - 3 270 86
AY219338.1 - 2 - 3 250 86
BC016701.2 - 2 - 3 218 4108644 86
BC038976.1 - 2 - 3 217 5553184 86
BP299206.1 - 2 - 3 312 86

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ARHGAP15

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
regulation of cell shape
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
GTPase activator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000409869.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000409869.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000409869.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000409869.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000409869.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000409869.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000409869.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000419455.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000419455.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000419455.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000460776.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000460776.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000460776.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000460776.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000460776.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000460776.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000469117.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000469117.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000469117.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000469117.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000474474.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000474474.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000474474.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000474474.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000548242.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000548242.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000548242.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000548242.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000548242.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000548800.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000548800.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000548800.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000548800.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000548929.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000548929.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000548929.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000548929.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000549060.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000549060.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000549060.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000549436.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000549436.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000549436.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000549436.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000549436.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000552289.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000552289.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000552289.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000552641.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000552641.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000552641.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000552641.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000552641.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000552641.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000552641.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000552641.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000552641.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000552641.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000409869.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000409869.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000409869.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000409869.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000409869.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000409869.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000409869.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000419455.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000419455.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000419455.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000460776.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000460776.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000460776.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000460776.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000460776.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000460776.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000469117.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000469117.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000469117.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000469117.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000474474.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000474474.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000474474.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000474474.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000548242.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000548242.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000548242.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000548242.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000548242.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000548800.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000548800.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000548800.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000548800.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000548929.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000548929.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000548929.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000548929.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000549060.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000549060.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000549060.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000549436.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000549436.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000549436.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000549436.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000549436.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000552289.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000552289.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000552289.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000552641.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000552641.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000552641.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000552641.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000552641.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000552641.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000552641.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000552641.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000552641.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000552641.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Clariom D Assay TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Clariom D Assay TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Clariom D Assay TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Clariom D Assay TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Clariom D Assay TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Clariom D Assay TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Clariom D Assay TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Clariom D Assay TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Clariom D Assay TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Clariom D Assay TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Clariom D Assay TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Clariom D Assay TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Clariom D Assay TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Clariom D Assay TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Clariom D Assay TC0200009539.hg.1 NM_018460.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0