This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00537793_g1
See other GPR156 GE Assays ›
Gene Symbol:
GPR156
Gene Name:
G protein-coupled receptor 156
Gene Aliases:
GABABL, PGR28
Chromosome Location:
Chr.3: 120165481 - 120285122 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00537793_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR156
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001168271.1 NP_001161743.1 3 - 4 561 96
NM_153002.2 NP_694547.2 3 - 4 561 96
XM_005247165.2 XP_005247222.1 4 - 5 838 96
XM_011512484.2 XP_011510786.1 5 - 6 911 96
XM_011512487.1 XP_011510789.1 3 - 4 463 96
XM_017005795.1 XP_016861284.1 4 - 5 874 96
XM_017005796.1 XP_016861285.1 4 - 5 883 96
XM_017005797.1 XP_016861286.1 4 - 5 838 96
GenBank mRNA AF488739.1 - 3 - 4 364 96
BC113701.1 - 3 - 4 384 8322753 96
BC143606.1 - 3 - 4 384 9052119 96

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300012140.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03001689.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GPR156

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
G-protein coupled GABA receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR03030471.hg.1 ENST00000315843.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03030471.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03030471.hg.1 ENST00000464295.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03030471.hg.1 NM_001168271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03030471.hg.1 NM_153002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03030471.hg.1 TC03001689.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000481907.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000481907.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000481907.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000481907.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000481907.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000481907.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000495912.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000495912.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000495912.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000495912.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000495912.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000495912.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000495912.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Clariom D Assay PSR0300190339.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Clariom D Assay PSR0300190339.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300190339.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300190339.hg.1 NM_001168271.1 Refseq
Affymetrix Human Clariom D Assay PSR0300190339.hg.1 NM_153002.2 Refseq
Affymetrix Human Clariom D Assay PSR0300190339.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Clariom D Assay PSR0300190339.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300190339.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300190339.hg.1 NM_001168271.1 Refseq
Affymetrix Human Clariom D Assay PSR0300190339.hg.1 NM_153002.2 Refseq
Affymetrix Human Clariom D Assay PSR0300190339.hg.1 TC0300012140.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000481907.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000481907.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000481907.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000481907.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000481907.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000481907.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000495912.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000495912.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000495912.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000495912.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000495912.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000495912.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000495912.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_153002.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0