This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00536308_s1
See other ZCCHC2 GE Assays ›
Gene Symbol:
ZCCHC2
Gene Name:
zinc finger CCHC-type containing 2
Gene Aliases:
C18orf49
Chromosome Location:
Chr.18: 62523410 - 62587709 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00536308_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZCCHC2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011526044.1 XP_011524346.1 1 338 72
GenBank mRNA BC006340.1 - 1 1543 72

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_011526044.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Gene Ontology Categories:

Process(es)

nucleic acid binding
zinc ion binding
phosphatidylinositol binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0