This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00426589_m1
See other TYMS GE Assays ›
Gene Symbol:
TYMS
Gene Name:
thymidylate synthetase
Gene Aliases:
HST422, TMS, TS
Chromosome Location:
Chr.18: 657604 - 673499 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00426589_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TYMS
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001071.2 NP_001062.1 4 - 5 693 68
GenBank mRNA AB062290.1 - 5 - 6 635 68
AB077207.1 - 2 - 3 305 68
AK223372.1 - 4 - 5 681 68
AK292620.1 - 4 - 5 588 68
AK314140.1 - 4 - 5 652 68
BC002567.2 - 4 - 5 622 3138877 68
BC013919.1 - 5 - 6 649 4048625 68
BC018858.2 - 4 - 5 606 68
BC083512.1 - 5 - 6 660 4299083 68
BT006811.1 - 4 - 5 554 68
X02308.1 - 4 - 5 659 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1800006448.hg.1
This assay targets the same location as HTA transcript cluster(s) TC18000006.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TYMS

Panther Classification:

Molecular Function -

methyltransferase

Gene Ontology Categories:

Function(s) Process(es)

G1/S transition of mitotic cell cycle
regulation of transcription involved in G1/S transition of mitotic cell cycle
nucleobase-containing compound metabolic process
dTMP biosynthetic process
dTTP biosynthetic process
aging
circadian rhythm
cell proliferation
deoxyribonucleoside monophosphate biosynthetic process
response to toxic substance
cell growth
immortalization of host cell by virus
uracil metabolic process
organ regeneration
methylation
response to progesterone
response to vitamin A
response to cytokine
tetrahydrofolate interconversion
response to drug
response to ethanol
dUMP metabolic process
pyrimidine nucleoside biosynthetic process
response to organophosphorus
developmental growth
cartilage development
response to glucocorticoid
response to folic acid
intestinal epithelial cell maturation
DNA biosynthetic process
nucleotide binding
mRNA binding
thymidylate synthase activity
folic acid binding
drug binding
protein homodimerization activity
cofactor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323224.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323224.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323224.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323224.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323224.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323224.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323224.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323224.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323224.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323250.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323250.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323250.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323250.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323250.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323274.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323274.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323274.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323274.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323274.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323274.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323274.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323274.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323274.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323274.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323274.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323274.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323274.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000579128.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000579128.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000579128.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000579128.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000579128.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000579128.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000579128.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000581920.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000581920.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000581920.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000581920.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000581920.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000581920.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000584122.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000584122.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 NM_001071.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 NM_001071.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 NM_001071.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 NM_001071.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 NM_001071.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 NM_001071.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 NM_001071.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 NM_001071.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 NM_001071.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 NM_001071.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 NM_001071.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 NM_001071.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR18000060.hg.1 ENST00000323224.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR18000060.hg.1 ENST00000323250.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR18000060.hg.1 ENST00000323274.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR18000060.hg.1 NM_001071.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR18000060.hg.1 TC18000006.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1800144217.hg.1 ENST00000323224.7 Ensembl
Affymetrix Human Clariom D Assay PSR1800144217.hg.1 ENST00000323250.9 Ensembl
Affymetrix Human Clariom D Assay PSR1800144217.hg.1 ENST00000323274.14 Ensembl
Affymetrix Human Clariom D Assay PSR1800144217.hg.1 ENST00000581920.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800144217.hg.1 NM_001071.2 Refseq
Affymetrix Human Clariom D Assay PSR1800144217.hg.1 ENST00000323224.7 Ensembl
Affymetrix Human Clariom D Assay PSR1800144217.hg.1 ENST00000323250.9 Ensembl
Affymetrix Human Clariom D Assay PSR1800144217.hg.1 ENST00000323274.14 Ensembl
Affymetrix Human Clariom D Assay PSR1800144217.hg.1 ENST00000581920.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800144217.hg.1 NM_001071.2 Refseq
Affymetrix Human Clariom D Assay PSR1800144217.hg.1 TC1800006448.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323224.7 Ensembl
Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323224.7 Ensembl
Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323224.7 Ensembl
Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323224.7 Ensembl
Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323224.7 Ensembl
Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323224.7 Ensembl
Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323224.7 Ensembl
Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323224.7 Ensembl
Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323224.7 Ensembl
Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323250.9 Ensembl
Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323250.9 Ensembl
Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323250.9 Ensembl
Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323250.9 Ensembl
Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323250.9 Ensembl
Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323274.14 Ensembl
Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323274.14 Ensembl
Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323274.14 Ensembl
Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323274.14 Ensembl
Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323274.14 Ensembl
Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323274.14 Ensembl
Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323274.14 Ensembl
Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323274.14 Ensembl
Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323274.14 Ensembl
Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323274.14 Ensembl
Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323274.14 Ensembl
Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323274.14 Ensembl
Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323274.14 Ensembl
Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000579128.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000579128.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000579128.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000579128.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000579128.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000579128.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000579128.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000581920.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000581920.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000581920.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000581920.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000581920.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000581920.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000584122.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000584122.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006448.hg.1 NM_001071.2 Refseq
Affymetrix Human Clariom D Assay TC1800006448.hg.1 NM_001071.2 Refseq
Affymetrix Human Clariom D Assay TC1800006448.hg.1 NM_001071.2 Refseq
Affymetrix Human Clariom D Assay TC1800006448.hg.1 NM_001071.2 Refseq
Affymetrix Human Clariom D Assay TC1800006448.hg.1 NM_001071.2 Refseq
Affymetrix Human Clariom D Assay TC1800006448.hg.1 NM_001071.2 Refseq
Affymetrix Human Clariom D Assay TC1800006448.hg.1 NM_001071.2 Refseq
Affymetrix Human Clariom D Assay TC1800006448.hg.1 NM_001071.2 Refseq
Affymetrix Human Clariom D Assay TC1800006448.hg.1 NM_001071.2 Refseq
Affymetrix Human Clariom D Assay TC1800006448.hg.1 NM_001071.2 Refseq
Affymetrix Human Clariom D Assay TC1800006448.hg.1 NM_001071.2 Refseq
Affymetrix Human Clariom D Assay TC1800006448.hg.1 NM_001071.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0