This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00418775_m1
See other LINC00520 GE Assays ›
Gene Symbol:
LINC00520
Gene Name:
long intergenic non-protein coding RNA 520
Gene Aliases:
C14orf34
Chromosome Location:
Chr.14: 55781135 - 55796674 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00418775_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LINC00520
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BG104219.1 - 2 - 3 142 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Homo sapiens Inventoried Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000554186.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000554186.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000554186.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000554186.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000554186.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000554186.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000554186.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000554196.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000554196.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000554221.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000554221.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000554221.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000554221.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000554221.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000554221.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000554221.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000554221.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000556417.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000556417.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000556417.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000556417.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000556417.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000556417.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000556417.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000556417.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000556417.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000556417.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000556417.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000556417.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000556417.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000556417.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000560267.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000560267.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000560267.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000560267.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000560267.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000560267.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000560267.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000560267.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000560267.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000560267.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000560336.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000560336.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000560336.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000560336.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000560336.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000560336.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000560336.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000560336.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000560336.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000560336.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000560336.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 ENST00000560336.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 NR_026796.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 NR_026796.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 NR_026796.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 NR_026796.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 NR_026796.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 NR_026796.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 NR_026796.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 NR_026796.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 NR_026796.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 NR_026796.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 NR_026796.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 NR_026797.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 NR_026797.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 NR_026797.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 NR_026797.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 NR_026797.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 NR_026797.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 NR_026797.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 NR_026797.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 NR_026797.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009267.hg.1 NR_026797.1 Refseq
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000554186.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000554186.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000554186.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000554186.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000554186.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000554186.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000554186.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000554196.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000554196.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000554221.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000554221.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000554221.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000554221.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000554221.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000554221.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000554221.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000554221.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000556417.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000556417.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000556417.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000556417.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000556417.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000556417.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000556417.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000556417.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000556417.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000556417.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000556417.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000556417.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000556417.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000556417.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000560267.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000560267.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000560267.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000560267.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000560267.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000560267.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000560267.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000560267.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000560267.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000560267.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000560336.2 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000560336.2 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000560336.2 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000560336.2 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000560336.2 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000560336.2 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000560336.2 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000560336.2 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000560336.2 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000560336.2 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000560336.2 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 ENST00000560336.2 Ensembl
Affymetrix Human Clariom D Assay TC1400009267.hg.1 NR_026796.1 Refseq
Affymetrix Human Clariom D Assay TC1400009267.hg.1 NR_026796.1 Refseq
Affymetrix Human Clariom D Assay TC1400009267.hg.1 NR_026796.1 Refseq
Affymetrix Human Clariom D Assay TC1400009267.hg.1 NR_026796.1 Refseq
Affymetrix Human Clariom D Assay TC1400009267.hg.1 NR_026796.1 Refseq
Affymetrix Human Clariom D Assay TC1400009267.hg.1 NR_026796.1 Refseq
Affymetrix Human Clariom D Assay TC1400009267.hg.1 NR_026796.1 Refseq
Affymetrix Human Clariom D Assay TC1400009267.hg.1 NR_026796.1 Refseq
Affymetrix Human Clariom D Assay TC1400009267.hg.1 NR_026796.1 Refseq
Affymetrix Human Clariom D Assay TC1400009267.hg.1 NR_026796.1 Refseq
Affymetrix Human Clariom D Assay TC1400009267.hg.1 NR_026796.1 Refseq
Affymetrix Human Clariom D Assay TC1400009267.hg.1 NR_026797.1 Refseq
Affymetrix Human Clariom D Assay TC1400009267.hg.1 NR_026797.1 Refseq
Affymetrix Human Clariom D Assay TC1400009267.hg.1 NR_026797.1 Refseq
Affymetrix Human Clariom D Assay TC1400009267.hg.1 NR_026797.1 Refseq
Affymetrix Human Clariom D Assay TC1400009267.hg.1 NR_026797.1 Refseq
Affymetrix Human Clariom D Assay TC1400009267.hg.1 NR_026797.1 Refseq
Affymetrix Human Clariom D Assay TC1400009267.hg.1 NR_026797.1 Refseq
Affymetrix Human Clariom D Assay TC1400009267.hg.1 NR_026797.1 Refseq
Affymetrix Human Clariom D Assay TC1400009267.hg.1 NR_026797.1 Refseq
Affymetrix Human Clariom D Assay TC1400009267.hg.1 NR_026797.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0