This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00418313_m1
See other KANK3 GE Assays ›
Gene Symbol:
KANK3
Gene Name:
KN motif and ankyrin repeat domains 3
Gene Aliases:
ANKRD47
Chromosome Location:
Chr.19: 8322584 - 8343262 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00418313_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: KANK3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_198471.2 NP_940873.2 7 - 8 2001 75
XM_006722718.3 XP_006722781.1 7 - 8 2120 75
XM_011527884.2 XP_011526186.1 7 - 8 2377 75
XM_017026563.1 XP_016882052.1 6 - 7 2137 75
GenBank mRNA AB284126.1 - 7 - 8 2001 75
AK128815.1 - 7 - 8 2001 75
BC066124.1 - 7 - 8 2001 30345663 75
BC066775.1 - 7 - 8 2001 30333203 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1900009528.hg.1
This assay targets the same location as HTA transcript cluster(s) TC19001129.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene KANK3

Panther Classification:

Molecular Function -

non-motor actin binding protein

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR19015531.hg.1 ENST00000330915.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19015531.hg.1 NM_198471.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19015531.hg.1 TC19001129.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 ENST00000330915.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 ENST00000330915.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 ENST00000330915.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 ENST00000330915.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 ENST00000330915.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 ENST00000330915.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 ENST00000330915.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 ENST00000330915.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 ENST00000330915.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 ENST00000330915.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 ENST00000330915.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 ENST00000330915.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 ENST00000330915.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 ENST00000330915.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 ENST00000593331.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 ENST00000593331.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 ENST00000593331.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 ENST00000593649.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 ENST00000593649.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 ENST00000593649.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 ENST00000593649.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 ENST00000593649.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 ENST00000593649.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 ENST00000593649.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 ENST00000593649.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 ENST00000593649.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 ENST00000593649.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 ENST00000593649.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 ENST00000593649.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 ENST00000593649.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 ENST00000595639.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 ENST00000595639.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 ENST00000595639.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 ENST00000595639.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 ENST00000595639.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 ENST00000595639.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 ENST00000610351.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 ENST00000610351.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 ENST00000610351.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 ENST00000610351.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 ENST00000610351.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 ENST00000610351.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 ENST00000610351.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 NM_198471.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 NM_198471.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 NM_198471.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 NM_198471.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 NM_198471.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 NM_198471.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 NM_198471.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 NM_198471.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 NM_198471.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 NM_198471.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 NM_198471.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 NM_198471.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 NM_198471.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009528.hg.1 NM_198471.2 Refseq
Affymetrix Human Clariom D Assay PSR1900176641.hg.1 ENST00000330915.7 Ensembl
Affymetrix Human Clariom D Assay PSR1900176641.hg.1 ENST00000593649.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900176641.hg.1 NM_198471.2 Refseq
Affymetrix Human Clariom D Assay PSR1900176641.hg.1 ENST00000330915.7 Ensembl
Affymetrix Human Clariom D Assay PSR1900176641.hg.1 ENST00000593649.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900176641.hg.1 NM_198471.2 Refseq
Affymetrix Human Clariom D Assay PSR1900176641.hg.1 TC1900009528.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1900009528.hg.1 ENST00000330915.7 Ensembl
Affymetrix Human Clariom D Assay TC1900009528.hg.1 ENST00000330915.7 Ensembl
Affymetrix Human Clariom D Assay TC1900009528.hg.1 ENST00000330915.7 Ensembl
Affymetrix Human Clariom D Assay TC1900009528.hg.1 ENST00000330915.7 Ensembl
Affymetrix Human Clariom D Assay TC1900009528.hg.1 ENST00000330915.7 Ensembl
Affymetrix Human Clariom D Assay TC1900009528.hg.1 ENST00000330915.7 Ensembl
Affymetrix Human Clariom D Assay TC1900009528.hg.1 ENST00000330915.7 Ensembl
Affymetrix Human Clariom D Assay TC1900009528.hg.1 ENST00000330915.7 Ensembl
Affymetrix Human Clariom D Assay TC1900009528.hg.1 ENST00000330915.7 Ensembl
Affymetrix Human Clariom D Assay TC1900009528.hg.1 ENST00000330915.7 Ensembl
Affymetrix Human Clariom D Assay TC1900009528.hg.1 ENST00000330915.7 Ensembl
Affymetrix Human Clariom D Assay TC1900009528.hg.1 ENST00000330915.7 Ensembl
Affymetrix Human Clariom D Assay TC1900009528.hg.1 ENST00000330915.7 Ensembl
Affymetrix Human Clariom D Assay TC1900009528.hg.1 ENST00000330915.7 Ensembl
Affymetrix Human Clariom D Assay TC1900009528.hg.1 ENST00000593331.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009528.hg.1 ENST00000593331.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009528.hg.1 ENST00000593331.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009528.hg.1 ENST00000593649.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009528.hg.1 ENST00000593649.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009528.hg.1 ENST00000593649.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009528.hg.1 ENST00000593649.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009528.hg.1 ENST00000593649.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009528.hg.1 ENST00000593649.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009528.hg.1 ENST00000593649.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009528.hg.1 ENST00000593649.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009528.hg.1 ENST00000593649.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009528.hg.1 ENST00000593649.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009528.hg.1 ENST00000593649.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009528.hg.1 ENST00000593649.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009528.hg.1 ENST00000593649.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009528.hg.1 ENST00000595639.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009528.hg.1 ENST00000595639.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009528.hg.1 ENST00000595639.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009528.hg.1 ENST00000595639.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009528.hg.1 ENST00000595639.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009528.hg.1 ENST00000595639.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009528.hg.1 ENST00000610351.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009528.hg.1 ENST00000610351.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009528.hg.1 ENST00000610351.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009528.hg.1 ENST00000610351.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009528.hg.1 ENST00000610351.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009528.hg.1 ENST00000610351.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009528.hg.1 ENST00000610351.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009528.hg.1 NM_198471.2 Refseq
Affymetrix Human Clariom D Assay TC1900009528.hg.1 NM_198471.2 Refseq
Affymetrix Human Clariom D Assay TC1900009528.hg.1 NM_198471.2 Refseq
Affymetrix Human Clariom D Assay TC1900009528.hg.1 NM_198471.2 Refseq
Affymetrix Human Clariom D Assay TC1900009528.hg.1 NM_198471.2 Refseq
Affymetrix Human Clariom D Assay TC1900009528.hg.1 NM_198471.2 Refseq
Affymetrix Human Clariom D Assay TC1900009528.hg.1 NM_198471.2 Refseq
Affymetrix Human Clariom D Assay TC1900009528.hg.1 NM_198471.2 Refseq
Affymetrix Human Clariom D Assay TC1900009528.hg.1 NM_198471.2 Refseq
Affymetrix Human Clariom D Assay TC1900009528.hg.1 NM_198471.2 Refseq
Affymetrix Human Clariom D Assay TC1900009528.hg.1 NM_198471.2 Refseq
Affymetrix Human Clariom D Assay TC1900009528.hg.1 NM_198471.2 Refseq
Affymetrix Human Clariom D Assay TC1900009528.hg.1 NM_198471.2 Refseq
Affymetrix Human Clariom D Assay TC1900009528.hg.1 NM_198471.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0