Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003735.hg.1 |
ENST00000216144.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003735.hg.1 |
NM_182527.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003735.hg.1 |
TC22000200.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007032.hg.1 |
ENST00000216144.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007032.hg.1 |
ENST00000216144.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007032.hg.1 |
ENST00000216144.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007032.hg.1 |
ENST00000216144.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007032.hg.1 |
ENST00000216144.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007032.hg.1 |
ENST00000216144.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007032.hg.1 |
ENST00000216144.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007032.hg.1 |
ENST00000216144.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007032.hg.1 |
NM_182527.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007032.hg.1 |
NM_182527.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007032.hg.1 |
NM_182527.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007032.hg.1 |
NM_182527.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007032.hg.1 |
NM_182527.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007032.hg.1 |
NM_182527.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007032.hg.1 |
NM_182527.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200149157.hg.1 |
ENST00000216144.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200149157.hg.1 |
NM_182527.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200149157.hg.1 |
ENST00000216144.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200149157.hg.1 |
NM_182527.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200149157.hg.1 |
TC2200007032.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC2200007032.hg.1 |
ENST00000216144.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007032.hg.1 |
ENST00000216144.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007032.hg.1 |
ENST00000216144.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007032.hg.1 |
ENST00000216144.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007032.hg.1 |
ENST00000216144.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007032.hg.1 |
ENST00000216144.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007032.hg.1 |
ENST00000216144.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007032.hg.1 |
ENST00000216144.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007032.hg.1 |
NM_182527.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007032.hg.1 |
NM_182527.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007032.hg.1 |
NM_182527.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007032.hg.1 |
NM_182527.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007032.hg.1 |
NM_182527.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007032.hg.1 |
NM_182527.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007032.hg.1 |
NM_182527.2 |
Refseq |