This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00417121_m1
See other IFNLR1 GE Assays ›
Gene Symbol:
IFNLR1
Gene Name:
interferon lambda receptor 1
Gene Aliases:
CRF2/12, IFNLR, IL-28R1, IL28RA, LICR2
Chromosome Location:
Chr.1: 24154156 - 24187275 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00417121_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IFNLR1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_170743.3 NP_734464.1 4 - 5 532 92
NM_173064.2 NP_775087.1 4 - 5 532 92
NM_173065.2 NP_775088.1 4 - 5 532 92
XM_006710394.3 XP_006710457.2 3 - 4 596 92
XM_017000479.1 XP_016855968.1 3 - 4 561 92
GenBank mRNA AF439325.1 - 4 - 5 505 92
AJ534330.1 - 4 - 5 518 92
AJ534331.1 - 4 - 5 518 92
AK160364.1 - 4 - 5 508 92
AY129151.1 - 4 - 5 505 92
AY129152.1 - 4 - 5 505 92
BC101407.1 - 3 - 4 394 92
BC101408.1 - 3 - 4 394 92
BC101975.1 - 4 - 5 518 92
BC139722.1 - 4 - 5 518 92
BC140872.1 - 4 - 5 581 9021740 92
JN591376.1 - 4 - 5 505 92

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100013315.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01002359.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene IFNLR1

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

mucosal immune response
negative regulation of cell proliferation
cytokine-mediated signaling pathway
response to type III interferon
regulation of defense response to virus by host
defense response to virus
cytokine receptor activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000327575.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000327575.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000327575.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000327575.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000327575.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000327575.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000327575.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000327575.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000374418.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000374418.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000374418.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000374418.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000374418.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000374419.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000374419.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000374419.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000374419.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000374419.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000374419.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000374419.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000374421.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000374421.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000374421.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000374421.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000374421.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000374421.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 ENST00000374421.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_170743.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_170743.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_170743.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_170743.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_170743.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_170743.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_170743.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_170743.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_170743.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_170743.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_170743.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_173064.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_173064.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_173064.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_173064.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_173064.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_173064.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_173064.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_173064.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_173064.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_173064.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_173065.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_173065.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_173065.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_173065.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_173065.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_173065.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_173065.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_173065.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_173065.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013315.hg.1 NM_173065.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01036461.hg.1 ENST00000327535.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01036461.hg.1 ENST00000327575.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01036461.hg.1 ENST00000374419.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01036461.hg.1 ENST00000374421.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01036461.hg.1 NM_170743.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01036461.hg.1 NM_173064.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01036461.hg.1 NM_173065.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01036461.hg.1 TC01002359.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000327575.6 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000327575.6 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000327575.6 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000327575.6 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000327575.6 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000327575.6 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000327575.6 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000327575.6 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000374418.3 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000374418.3 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000374418.3 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000374418.3 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000374418.3 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000374419.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000374419.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000374419.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000374419.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000374419.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000374419.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000374419.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000374421.7 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000374421.7 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000374421.7 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000374421.7 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000374421.7 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000374421.7 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 ENST00000374421.7 Ensembl
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_170743.3 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_170743.3 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_170743.3 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_170743.3 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_170743.3 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_170743.3 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_170743.3 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_170743.3 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_170743.3 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_170743.3 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_170743.3 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_173064.2 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_173064.2 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_173064.2 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_173064.2 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_173064.2 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_173064.2 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_173064.2 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_173064.2 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_173064.2 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_173064.2 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_173065.2 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_173065.2 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_173065.2 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_173065.2 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_173065.2 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_173065.2 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_173065.2 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_173065.2 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_173065.2 Refseq
Affymetrix Human Clariom D Assay TC0100013315.hg.1 NM_173065.2 Refseq
Affymetrix Human Clariom D Assay PSR0100198885.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100198885.hg.1 ENST00000327575.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100198885.hg.1 ENST00000374419.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100198885.hg.1 ENST00000374421.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100198885.hg.1 NM_170743.3 Refseq
Affymetrix Human Clariom D Assay PSR0100198885.hg.1 NM_173064.2 Refseq
Affymetrix Human Clariom D Assay PSR0100198885.hg.1 NM_173065.2 Refseq
Affymetrix Human Clariom D Assay PSR0100198885.hg.1 ENST00000327535.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100198885.hg.1 ENST00000327575.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100198885.hg.1 ENST00000374419.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100198885.hg.1 ENST00000374421.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100198885.hg.1 NM_170743.3 Refseq
Affymetrix Human Clariom D Assay PSR0100198885.hg.1 NM_173064.2 Refseq
Affymetrix Human Clariom D Assay PSR0100198885.hg.1 NM_173065.2 Refseq
Affymetrix Human Clariom D Assay PSR0100198885.hg.1 TC0100013315.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-fd98957d6-rbs6c:80/100.66.128.142:80.
git-commit: 278a916a254c4ca07f9ccbb2b9efa8cd7578b5be
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0