Assay Details
Gene Symbol: MBNL1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_027037.1 | 1 - 2 | 194 | 61 | ||
GenBank mRNA | AK304857.1 | - | 1 - 2 | 114 | 61 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03001911.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK304857.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300012831.hg.1 | ENST00000445466.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012831.hg.1 | ENST00000445466.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012831.hg.1 | ENST00000445466.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012831.hg.1 | ENST00000608395.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012831.hg.1 | ENST00000608395.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012831.hg.1 | ENST00000608395.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012831.hg.1 | ENST00000608395.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012831.hg.1 | ENST00000608395.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012831.hg.1 | ENST00000608395.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012831.hg.1 | NR_027037.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012831.hg.1 | NR_027037.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012831.hg.1 | NR_027037.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012831.hg.1 | NR_027037.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012831.hg.1 | NR_027037.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012831.hg.1 | NR_027037.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012831.hg.1 | NR_027037.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012831.hg.1 | NR_027038.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012831.hg.1 | NR_027038.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012831.hg.1 | NR_027038.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012831.hg.1 | NR_027038.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012831.hg.1 | NR_027038.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012831.hg.1 | NR_027038.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012831.hg.1 | NR_027038.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03034409.hg.1 | ENST00000445466.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03034409.hg.1 | NR_027037.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03034409.hg.1 | NR_027038.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03034409.hg.1 | TC03001911.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03034407.hg.1 | ENST00000445466.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03034407.hg.1 | NR_027037.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03034407.hg.1 | NR_027038.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03034407.hg.1 | TC03001911.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03034410.hg.1 | NR_027037.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03034410.hg.1 | NR_027038.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03034410.hg.1 | TC03001911.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR0300196029.hg.1 | ENST00000445466.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300196029.hg.1 | ENST00000608395.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300196029.hg.1 | NR_027037.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300196029.hg.1 | NR_027038.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300196029.hg.1 | ENST00000445466.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300196029.hg.1 | ENST00000608395.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300196029.hg.1 | NR_027037.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300196029.hg.1 | NR_027038.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300196029.hg.1 | TC0300012831.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300196031.hg.1 | ENST00000445466.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300196031.hg.1 | ENST00000608395.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300196031.hg.1 | NR_027037.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300196031.hg.1 | NR_027038.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300196031.hg.1 | ENST00000445466.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300196031.hg.1 | ENST00000608395.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300196031.hg.1 | NR_027037.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300196031.hg.1 | NR_027038.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300196031.hg.1 | TC0300012831.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0300012831.hg.1 | ENST00000445466.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012831.hg.1 | ENST00000445466.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012831.hg.1 | ENST00000445466.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012831.hg.1 | ENST00000608395.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012831.hg.1 | ENST00000608395.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012831.hg.1 | ENST00000608395.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012831.hg.1 | ENST00000608395.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012831.hg.1 | ENST00000608395.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012831.hg.1 | ENST00000608395.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012831.hg.1 | NR_027037.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012831.hg.1 | NR_027037.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012831.hg.1 | NR_027037.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012831.hg.1 | NR_027037.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012831.hg.1 | NR_027037.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012831.hg.1 | NR_027037.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012831.hg.1 | NR_027037.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012831.hg.1 | NR_027038.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012831.hg.1 | NR_027038.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012831.hg.1 | NR_027038.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012831.hg.1 | NR_027038.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012831.hg.1 | NR_027038.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012831.hg.1 | NR_027038.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012831.hg.1 | NR_027038.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300196033.hg.1 | NR_027037.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300196033.hg.1 | NR_027038.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300196033.hg.1 | NR_027037.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300196033.hg.1 | TC0300012831.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300196033.hg.1 | NR_027038.1 | Refseq |
