This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00393450_m1
See other GRID1 GE Assays ›
Gene Symbol:
GRID1
Gene Name:
glutamate ionotropic receptor delta type subunit 1
Gene Aliases:
GluD1
Chromosome Location:
Chr.10: 85599555 - 86366493 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00393450_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GRID1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_017551.2 NP_060021.1 10 - 11 1639 74
XM_011539720.2 XP_011538022.1 10 - 11 1636 74
GenBank mRNA AB033046.1 - 7 - 8 886 74
AK127168.1 - 10 - 11 1624 74
AK293104.1 - 10 - 11 1597 74
AK302192.1 - 3 - 4 411 74
BC039263.1 - 10 - 11 1639 5273869 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000011287.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10001475.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GRID1

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

ion transmembrane transport
social behavior
ionotropic glutamate receptor signaling pathway
ionotropic glutamate receptor activity
extracellular-glutamate-gated ion channel activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000327946.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000327946.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000327946.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000327946.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000327946.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000327946.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000327946.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000327946.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000327946.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000327946.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000327946.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000327946.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000327946.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000327946.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000327946.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000327946.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000327946.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000327946.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000327946.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000464741.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000464741.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000464741.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000464741.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000464741.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000464741.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000464741.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000464741.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000464741.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000464741.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000464741.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000464741.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000464741.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000464741.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000464741.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000464741.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000464741.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000536331.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000536331.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000536331.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000536331.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000536331.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000536331.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000536331.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000536331.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000536331.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000536331.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000536331.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000536331.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000536331.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000536331.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000552278.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000552278.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000552278.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000614988.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000614988.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 ENST00000614988.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 NM_017551.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 NM_017551.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 NM_017551.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 NM_017551.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 NM_017551.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 NM_017551.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 NM_017551.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 NM_017551.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 NM_017551.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 NM_017551.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 NM_017551.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 NM_017551.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 NM_017551.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 NM_017551.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 NM_017551.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 NM_017551.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 NM_017551.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 NM_017551.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011287.hg.1 NM_017551.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10018673.hg.1 ENST00000327946.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10018673.hg.1 ENST00000536331.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10018673.hg.1 NM_017551.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10018673.hg.1 TC10001475.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000327946.11 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000327946.11 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000327946.11 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000327946.11 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000327946.11 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000327946.11 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000327946.11 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000327946.11 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000327946.11 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000327946.11 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000327946.11 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000327946.11 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000327946.11 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000327946.11 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000327946.11 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000327946.11 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000327946.11 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000327946.11 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000327946.11 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000464741.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000464741.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000464741.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000464741.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000464741.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000464741.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000464741.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000464741.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000464741.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000464741.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000464741.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000464741.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000464741.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000464741.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000464741.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000464741.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000464741.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000536331.5 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000536331.5 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000536331.5 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000536331.5 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000536331.5 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000536331.5 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000536331.5 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000536331.5 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000536331.5 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000536331.5 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000536331.5 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000536331.5 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000536331.5 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000536331.5 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000552278.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000552278.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000552278.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000614988.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000614988.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 ENST00000614988.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011287.hg.1 NM_017551.2 Refseq
Affymetrix Human Clariom D Assay TC1000011287.hg.1 NM_017551.2 Refseq
Affymetrix Human Clariom D Assay TC1000011287.hg.1 NM_017551.2 Refseq
Affymetrix Human Clariom D Assay TC1000011287.hg.1 NM_017551.2 Refseq
Affymetrix Human Clariom D Assay TC1000011287.hg.1 NM_017551.2 Refseq
Affymetrix Human Clariom D Assay TC1000011287.hg.1 NM_017551.2 Refseq
Affymetrix Human Clariom D Assay TC1000011287.hg.1 NM_017551.2 Refseq
Affymetrix Human Clariom D Assay TC1000011287.hg.1 NM_017551.2 Refseq
Affymetrix Human Clariom D Assay TC1000011287.hg.1 NM_017551.2 Refseq
Affymetrix Human Clariom D Assay TC1000011287.hg.1 NM_017551.2 Refseq
Affymetrix Human Clariom D Assay TC1000011287.hg.1 NM_017551.2 Refseq
Affymetrix Human Clariom D Assay TC1000011287.hg.1 NM_017551.2 Refseq
Affymetrix Human Clariom D Assay TC1000011287.hg.1 NM_017551.2 Refseq
Affymetrix Human Clariom D Assay TC1000011287.hg.1 NM_017551.2 Refseq
Affymetrix Human Clariom D Assay TC1000011287.hg.1 NM_017551.2 Refseq
Affymetrix Human Clariom D Assay TC1000011287.hg.1 NM_017551.2 Refseq
Affymetrix Human Clariom D Assay TC1000011287.hg.1 NM_017551.2 Refseq
Affymetrix Human Clariom D Assay TC1000011287.hg.1 NM_017551.2 Refseq
Affymetrix Human Clariom D Assay TC1000011287.hg.1 NM_017551.2 Refseq
Affymetrix Human Clariom D Assay PSR1000176778.hg.1 ENST00000327946.11 Ensembl
Affymetrix Human Clariom D Assay PSR1000176778.hg.1 ENST00000464741.2 Ensembl
Affymetrix Human Clariom D Assay PSR1000176778.hg.1 ENST00000536331.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000176778.hg.1 NM_017551.2 Refseq
Affymetrix Human Clariom D Assay PSR1000176778.hg.1 ENST00000327946.11 Ensembl
Affymetrix Human Clariom D Assay PSR1000176778.hg.1 ENST00000464741.2 Ensembl
Affymetrix Human Clariom D Assay PSR1000176778.hg.1 ENST00000536331.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000176778.hg.1 NM_017551.2 Refseq
Affymetrix Human Clariom D Assay PSR1000176778.hg.1 TC1000011287.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0