This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00390836_m1
See other VWA8 GE Assays ›
Gene Symbol:
VWA8
Gene Name:
von Willebrand factor A domain containing 8
Gene Aliases:
KIAA0564
Chromosome Location:
Chr.13: 41566825 - 41961111 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00390836_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: VWA8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_015058.1 NP_055873.1 43 - 44 5440 74
XM_006719791.3 XP_006719854.1 35 - 36 4396 74
XM_017020469.1 XP_016875958.1 45 - 46 5499 74
XM_017020472.1 XP_016875961.1 27 - 28 3428 74
XM_017020473.1 XP_016875962.1 26 - 27 3321 74
GenBank mRNA AB011136.1 - 32 - 33 3980 74
AK025432.1 - 3 - 4 382 74
AK308568.1 - 2 - 3 165 74
BC131802.1 - 44 - 45 5273 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1300008735.hg.1
This assay targets the same location as HTA transcript cluster(s) TC13000607.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene VWA8

Gene Ontology Categories:

Process(es)

ATP binding
ATPase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR13006904.hg.1 ENST00000251030.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13006904.hg.1 ENST00000379310.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13006904.hg.1 NM_015058.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13006904.hg.1 TC13000607.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1300158917.hg.1 ENST00000379310.7 Ensembl
Affymetrix Human Clariom D Assay PSR1300158917.hg.1 NM_015058.1 Refseq
Affymetrix Human Clariom D Assay PSR1300158917.hg.1 ENST00000379310.7 Ensembl
Affymetrix Human Clariom D Assay PSR1300158917.hg.1 NM_015058.1 Refseq
Affymetrix Human Clariom D Assay PSR1300158917.hg.1 TC1300008735.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0