This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00390140_m1
See other SEC14L5 GE Assays ›
Gene Symbol:
SEC14L5
Gene Name:
SEC14 like lipid binding 5
Gene Aliases:
PRELID4B
Chromosome Location:
Chr.16: 4958317 - 5019155 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00390140_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SEC14L5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_014692.1 NP_055507.1 13 - 14 1754 89
GenBank mRNA AB007880.2 - 13 - 14 1754 89

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1600011346.hg.1
This assay targets the same location as HTA transcript cluster(s) TC16000138.hg.1

Set Membership:

Amplicon length between 86 and 100 Transporter Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

transport
transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1600011346.hg.1 ENST00000251170.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011346.hg.1 ENST00000251170.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011346.hg.1 ENST00000251170.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011346.hg.1 ENST00000251170.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011346.hg.1 ENST00000251170.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011346.hg.1 ENST00000251170.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011346.hg.1 ENST00000251170.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011346.hg.1 ENST00000251170.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011346.hg.1 ENST00000251170.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011346.hg.1 ENST00000251170.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011346.hg.1 ENST00000251170.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011346.hg.1 ENST00000251170.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011346.hg.1 ENST00000251170.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011346.hg.1 ENST00000251170.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011346.hg.1 ENST00000251170.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011346.hg.1 ENST00000251170.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011346.hg.1 ENST00000251170.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011346.hg.1 ENST00000251170.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011346.hg.1 ENST00000251170.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011346.hg.1 ENST00000587469.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011346.hg.1 ENST00000587469.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011346.hg.1 ENST00000587469.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011346.hg.1 ENST00000587469.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011346.hg.1 ENST00000587469.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011346.hg.1 ENST00000587469.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011346.hg.1 NM_014692.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011346.hg.1 NM_014692.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011346.hg.1 NM_014692.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011346.hg.1 NM_014692.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011346.hg.1 NM_014692.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011346.hg.1 NM_014692.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011346.hg.1 NM_014692.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011346.hg.1 NM_014692.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011346.hg.1 NM_014692.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011346.hg.1 NM_014692.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011346.hg.1 NM_014692.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011346.hg.1 NM_014692.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011346.hg.1 NM_014692.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011346.hg.1 NM_014692.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011346.hg.1 NM_014692.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011346.hg.1 NM_014692.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011346.hg.1 NM_014692.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011346.hg.1 NM_014692.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011346.hg.1 NM_014692.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16002191.hg.1 ENST00000251170.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16002191.hg.1 NM_014692.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16002191.hg.1 TC16000138.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC1600011346.hg.1 ENST00000251170.11 Ensembl
Affymetrix Human Clariom D Assay TC1600011346.hg.1 ENST00000251170.11 Ensembl
Affymetrix Human Clariom D Assay TC1600011346.hg.1 ENST00000251170.11 Ensembl
Affymetrix Human Clariom D Assay TC1600011346.hg.1 ENST00000251170.11 Ensembl
Affymetrix Human Clariom D Assay TC1600011346.hg.1 ENST00000251170.11 Ensembl
Affymetrix Human Clariom D Assay TC1600011346.hg.1 ENST00000251170.11 Ensembl
Affymetrix Human Clariom D Assay TC1600011346.hg.1 ENST00000251170.11 Ensembl
Affymetrix Human Clariom D Assay TC1600011346.hg.1 ENST00000251170.11 Ensembl
Affymetrix Human Clariom D Assay TC1600011346.hg.1 ENST00000251170.11 Ensembl
Affymetrix Human Clariom D Assay TC1600011346.hg.1 ENST00000251170.11 Ensembl
Affymetrix Human Clariom D Assay TC1600011346.hg.1 ENST00000251170.11 Ensembl
Affymetrix Human Clariom D Assay TC1600011346.hg.1 ENST00000251170.11 Ensembl
Affymetrix Human Clariom D Assay TC1600011346.hg.1 ENST00000251170.11 Ensembl
Affymetrix Human Clariom D Assay TC1600011346.hg.1 ENST00000251170.11 Ensembl
Affymetrix Human Clariom D Assay TC1600011346.hg.1 ENST00000251170.11 Ensembl
Affymetrix Human Clariom D Assay TC1600011346.hg.1 ENST00000251170.11 Ensembl
Affymetrix Human Clariom D Assay TC1600011346.hg.1 ENST00000251170.11 Ensembl
Affymetrix Human Clariom D Assay TC1600011346.hg.1 ENST00000251170.11 Ensembl
Affymetrix Human Clariom D Assay TC1600011346.hg.1 ENST00000251170.11 Ensembl
Affymetrix Human Clariom D Assay TC1600011346.hg.1 ENST00000587469.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011346.hg.1 ENST00000587469.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011346.hg.1 ENST00000587469.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011346.hg.1 ENST00000587469.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011346.hg.1 ENST00000587469.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011346.hg.1 ENST00000587469.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011346.hg.1 NM_014692.1 Refseq
Affymetrix Human Clariom D Assay TC1600011346.hg.1 NM_014692.1 Refseq
Affymetrix Human Clariom D Assay TC1600011346.hg.1 NM_014692.1 Refseq
Affymetrix Human Clariom D Assay TC1600011346.hg.1 NM_014692.1 Refseq
Affymetrix Human Clariom D Assay TC1600011346.hg.1 NM_014692.1 Refseq
Affymetrix Human Clariom D Assay TC1600011346.hg.1 NM_014692.1 Refseq
Affymetrix Human Clariom D Assay TC1600011346.hg.1 NM_014692.1 Refseq
Affymetrix Human Clariom D Assay TC1600011346.hg.1 NM_014692.1 Refseq
Affymetrix Human Clariom D Assay TC1600011346.hg.1 NM_014692.1 Refseq
Affymetrix Human Clariom D Assay TC1600011346.hg.1 NM_014692.1 Refseq
Affymetrix Human Clariom D Assay TC1600011346.hg.1 NM_014692.1 Refseq
Affymetrix Human Clariom D Assay TC1600011346.hg.1 NM_014692.1 Refseq
Affymetrix Human Clariom D Assay TC1600011346.hg.1 NM_014692.1 Refseq
Affymetrix Human Clariom D Assay TC1600011346.hg.1 NM_014692.1 Refseq
Affymetrix Human Clariom D Assay TC1600011346.hg.1 NM_014692.1 Refseq
Affymetrix Human Clariom D Assay TC1600011346.hg.1 NM_014692.1 Refseq
Affymetrix Human Clariom D Assay TC1600011346.hg.1 NM_014692.1 Refseq
Affymetrix Human Clariom D Assay TC1600011346.hg.1 NM_014692.1 Refseq
Affymetrix Human Clariom D Assay TC1600011346.hg.1 NM_014692.1 Refseq
Affymetrix Human Clariom D Assay PSR1600189252.hg.1 ENST00000251170.11 Ensembl
Affymetrix Human Clariom D Assay PSR1600189252.hg.1 NM_014692.1 Refseq
Affymetrix Human Clariom D Assay PSR1600189252.hg.1 ENST00000251170.11 Ensembl
Affymetrix Human Clariom D Assay PSR1600189252.hg.1 NM_014692.1 Refseq
Affymetrix Human Clariom D Assay PSR1600189252.hg.1 TC1600011346.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0