This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00384260_m1
See other STAMBPL1 GE Assays ›
Gene Symbol:
STAMBPL1
Gene Name:
STAM binding protein like 1
Gene Aliases:
ALMalpha, AMSH-FP, AMSH-LP, bA399O19.2
Chromosome Location:
Chr.10: 88879368 - 88923502 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00384260_m1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: STAMBPL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_020799.3 NP_065850.1 8 - 9 1544 133
XM_006717928.2 XP_006717991.1 9 - 10 1645 133
XM_006717930.3 XP_006717993.1 8 - 9 2132 133
XM_011539983.1 XP_011538285.1 9 - 10 2052 133
XM_011539985.1 XP_011538287.1 9 - 10 1375 133
XM_017016453.1 XP_016871942.1 9 - 10 2369 133
XM_017016454.1 XP_016871943.1 10 - 11 1853 133
XM_017016455.1 XP_016871944.1 11 - 12 2308 133
XM_017016456.1 XP_016871945.1 9 - 10 1965 133
XM_017016458.1 XP_016871947.1 6 - 7 1069 133
GenBank mRNA AB010120.1 - 7 - 8 1072 133
AB037794.1 - 8 - 9 1865 133
AK056086.1 - 8 - 9 1473 133
AK090678.1 - 3 - 4 1365 133
AK092946.1 - 7 - 8 1673 133
BC010846.1 - 8 - 9 1462 3898533 133
BX649096.1 - 9 - 10 9967 133

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000008385.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10002941.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene STAMBPL1

Panther Classification:

Molecular Function -

metalloprotease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
protein binding
metallopeptidase activity
metal ion binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371922.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371922.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371922.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371922.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371922.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371922.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371922.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371922.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371924.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371924.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371924.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371924.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371924.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371924.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371924.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371924.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371924.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371924.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371924.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371924.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371924.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371926.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371926.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371926.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371926.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371926.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371926.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371926.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371926.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371926.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371926.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371926.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371926.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371926.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371926.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371926.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371926.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371927.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371927.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371927.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371927.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371927.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371927.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371927.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371927.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371927.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371927.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371927.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371927.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371927.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371927.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000371927.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000461650.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000461650.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000461650.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000461650.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000468698.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000468698.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 ENST00000468698.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 NM_020799.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 NM_020799.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 NM_020799.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 NM_020799.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 NM_020799.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 NM_020799.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 NM_020799.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 NM_020799.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 NM_020799.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 NM_020799.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 NM_020799.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 NM_020799.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 NM_020799.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 NM_020799.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008385.hg.1 NM_020799.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10006950.hg.1 ENST00000371922.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10006950.hg.1 ENST00000371924.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10006950.hg.1 ENST00000371926.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10006950.hg.1 ENST00000371927.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10006950.hg.1 NM_020799.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10006950.hg.1 TC10002941.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371922.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371922.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371922.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371922.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371922.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371922.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371922.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371922.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371924.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371924.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371924.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371924.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371924.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371924.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371924.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371924.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371924.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371924.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371924.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371924.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371924.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371926.7 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371926.7 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371926.7 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371926.7 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371926.7 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371926.7 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371926.7 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371926.7 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371926.7 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371926.7 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371926.7 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371926.7 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371926.7 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371926.7 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371926.7 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371926.7 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371927.7 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371927.7 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371927.7 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371927.7 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371927.7 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371927.7 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371927.7 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371927.7 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371927.7 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371927.7 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371927.7 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371927.7 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371927.7 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371927.7 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000371927.7 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000461650.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000461650.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000461650.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000461650.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000468698.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000468698.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 ENST00000468698.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008385.hg.1 NM_020799.3 Refseq
Affymetrix Human Clariom D Assay TC1000008385.hg.1 NM_020799.3 Refseq
Affymetrix Human Clariom D Assay TC1000008385.hg.1 NM_020799.3 Refseq
Affymetrix Human Clariom D Assay TC1000008385.hg.1 NM_020799.3 Refseq
Affymetrix Human Clariom D Assay TC1000008385.hg.1 NM_020799.3 Refseq
Affymetrix Human Clariom D Assay TC1000008385.hg.1 NM_020799.3 Refseq
Affymetrix Human Clariom D Assay TC1000008385.hg.1 NM_020799.3 Refseq
Affymetrix Human Clariom D Assay TC1000008385.hg.1 NM_020799.3 Refseq
Affymetrix Human Clariom D Assay TC1000008385.hg.1 NM_020799.3 Refseq
Affymetrix Human Clariom D Assay TC1000008385.hg.1 NM_020799.3 Refseq
Affymetrix Human Clariom D Assay TC1000008385.hg.1 NM_020799.3 Refseq
Affymetrix Human Clariom D Assay TC1000008385.hg.1 NM_020799.3 Refseq
Affymetrix Human Clariom D Assay TC1000008385.hg.1 NM_020799.3 Refseq
Affymetrix Human Clariom D Assay TC1000008385.hg.1 NM_020799.3 Refseq
Affymetrix Human Clariom D Assay TC1000008385.hg.1 NM_020799.3 Refseq
Affymetrix Human Clariom D Assay PSR1000156822.hg.1 ENST00000371922.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000156822.hg.1 ENST00000371924.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000156822.hg.1 ENST00000371926.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000156822.hg.1 ENST00000371927.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000156822.hg.1 NM_020799.3 Refseq
Affymetrix Human Clariom D Assay PSR1000156822.hg.1 ENST00000371922.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000156822.hg.1 ENST00000371924.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000156822.hg.1 ENST00000371926.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000156822.hg.1 ENST00000371927.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000156822.hg.1 NM_020799.3 Refseq
Affymetrix Human Clariom D Assay PSR1000156822.hg.1 TC1000008385.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 124733 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0