This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00383612_g1
See other TIMM13 GE Assays ›
Gene Symbol:
TIMM13
Gene Name:
translocase of inner mitochondrial membrane 13
Gene Aliases:
TIM13, TIM13B, TIMM13A, TIMM13B, ppv1
Chromosome Location:
Chr.19: 2425624 - 2427916 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00383612_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TIMM13
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_012458.3 NP_036590.1 2 - 3 574 81
GenBank mRNA AF144700.1 - 2 - 3 237 81
AF152352.1 - 2 - 3 191 81
AK024764.1 - 2 - 3 209 81
BC008607.1 - 2 - 3 232 3453921 81
BI908100.1 - 2 - 3 535 81

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TIMM13

Gene Ontology Categories:

Function(s) Process(es)

protein targeting to mitochondrion
sensory perception of sound
chaperone-mediated protein transport
zinc ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0