This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00383304_m1
See other KCNG1 GE Assays ›
Gene Symbol:
KCNG1
Gene Name:
potassium voltage-gated channel modifier subfamily G member 1
Gene Aliases:
K13, KCNG, KV6.1, kH2
Chromosome Location:
Chr.20: 51003656 - 51023176 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00383304_m1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: KCNG1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011528804.1 XP_011527106.1 1 - 2 519 104
GenBank mRNA BC006367.1 - 1 - 2 423 4131541 104
DA576272.1 - 1 - 2 104

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2000009448.hg.1

Additional Information:

This assay is in the 5'UTR region of the BC006367.1,XM_011528804.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens 1700 Inventoried Probe spans exons

Panther Classification:

Molecular Function -

voltage-gated ion channel

Gene Ontology Categories:

Function(s) Process(es)

potassium ion transport
protein homooligomerization
potassium ion transmembrane transport
regulation of delayed rectifier potassium channel activity
delayed rectifier potassium channel activity
potassium channel activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC2000009448.hg.1 ENST00000371571.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009448.hg.1 ENST00000371571.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009448.hg.1 ENST00000371571.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009448.hg.1 ENST00000371571.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009448.hg.1 ENST00000371571.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009448.hg.1 ENST00000371571.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009448.hg.1 ENST00000371571.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009448.hg.1 ENST00000371571.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009448.hg.1 ENST00000424171.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009448.hg.1 ENST00000424171.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009448.hg.1 ENST00000424171.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009448.hg.1 ENST00000424171.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009448.hg.1 ENST00000424171.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009448.hg.1 ENST00000424171.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009448.hg.1 ENST00000424171.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009448.hg.1 ENST00000433903.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009448.hg.1 ENST00000433903.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009448.hg.1 ENST00000433903.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009448.hg.1 ENST00000433903.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009448.hg.1 ENST00000433903.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009448.hg.1 ENST00000433903.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009448.hg.1 ENST00000439216.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009448.hg.1 ENST00000439216.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009448.hg.1 ENST00000439216.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009448.hg.1 ENST00000439216.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009448.hg.1 ENST00000439216.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009448.hg.1 ENST00000439216.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009448.hg.1 ENST00000439216.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009448.hg.1 ENST00000447736.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009448.hg.1 ENST00000447736.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009448.hg.1 ENST00000447736.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009448.hg.1 ENST00000447736.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009448.hg.1 ENST00000447736.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009448.hg.1 ENST00000506387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009448.hg.1 ENST00000506387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009448.hg.1 ENST00000506387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009448.hg.1 ENST00000506387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009448.hg.1 NM_002237.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009448.hg.1 NM_002237.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009448.hg.1 NM_002237.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009448.hg.1 NM_002237.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009448.hg.1 NM_002237.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009448.hg.1 NM_002237.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009448.hg.1 NM_002237.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009448.hg.1 NM_002237.3 Refseq
Affymetrix Human Clariom D Assay TC2000009448.hg.1 ENST00000371571.4 Ensembl
Affymetrix Human Clariom D Assay TC2000009448.hg.1 ENST00000371571.4 Ensembl
Affymetrix Human Clariom D Assay TC2000009448.hg.1 ENST00000371571.4 Ensembl
Affymetrix Human Clariom D Assay TC2000009448.hg.1 ENST00000371571.4 Ensembl
Affymetrix Human Clariom D Assay TC2000009448.hg.1 ENST00000371571.4 Ensembl
Affymetrix Human Clariom D Assay TC2000009448.hg.1 ENST00000371571.4 Ensembl
Affymetrix Human Clariom D Assay TC2000009448.hg.1 ENST00000371571.4 Ensembl
Affymetrix Human Clariom D Assay TC2000009448.hg.1 ENST00000371571.4 Ensembl
Affymetrix Human Clariom D Assay TC2000009448.hg.1 ENST00000424171.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009448.hg.1 ENST00000424171.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009448.hg.1 ENST00000424171.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009448.hg.1 ENST00000424171.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009448.hg.1 ENST00000424171.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009448.hg.1 ENST00000424171.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009448.hg.1 ENST00000424171.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009448.hg.1 ENST00000433903.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009448.hg.1 ENST00000433903.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009448.hg.1 ENST00000433903.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009448.hg.1 ENST00000433903.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009448.hg.1 ENST00000433903.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009448.hg.1 ENST00000433903.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009448.hg.1 ENST00000439216.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009448.hg.1 ENST00000439216.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009448.hg.1 ENST00000439216.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009448.hg.1 ENST00000439216.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009448.hg.1 ENST00000439216.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009448.hg.1 ENST00000439216.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009448.hg.1 ENST00000439216.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009448.hg.1 ENST00000447736.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009448.hg.1 ENST00000447736.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009448.hg.1 ENST00000447736.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009448.hg.1 ENST00000447736.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009448.hg.1 ENST00000447736.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009448.hg.1 ENST00000506387.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009448.hg.1 ENST00000506387.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009448.hg.1 ENST00000506387.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009448.hg.1 ENST00000506387.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009448.hg.1 NM_002237.3 Refseq
Affymetrix Human Clariom D Assay TC2000009448.hg.1 NM_002237.3 Refseq
Affymetrix Human Clariom D Assay TC2000009448.hg.1 NM_002237.3 Refseq
Affymetrix Human Clariom D Assay TC2000009448.hg.1 NM_002237.3 Refseq
Affymetrix Human Clariom D Assay TC2000009448.hg.1 NM_002237.3 Refseq
Affymetrix Human Clariom D Assay TC2000009448.hg.1 NM_002237.3 Refseq
Affymetrix Human Clariom D Assay TC2000009448.hg.1 NM_002237.3 Refseq
Affymetrix Human Clariom D Assay TC2000009448.hg.1 NM_002237.3 Refseq
Affymetrix Human Clariom D Assay PSR2000166092.hg.1 ENST00000371571.4 Ensembl
Affymetrix Human Clariom D Assay PSR2000166092.hg.1 ENST00000424171.5 Ensembl
Affymetrix Human Clariom D Assay PSR2000166092.hg.1 ENST00000433903.5 Ensembl
Affymetrix Human Clariom D Assay PSR2000166092.hg.1 ENST00000439216.5 Ensembl
Affymetrix Human Clariom D Assay PSR2000166092.hg.1 NM_002237.3 Refseq
Affymetrix Human Clariom D Assay PSR2000166092.hg.1 ENST00000371571.4 Ensembl
Affymetrix Human Clariom D Assay PSR2000166092.hg.1 ENST00000424171.5 Ensembl
Affymetrix Human Clariom D Assay PSR2000166092.hg.1 ENST00000433903.5 Ensembl
Affymetrix Human Clariom D Assay PSR2000166092.hg.1 ENST00000439216.5 Ensembl
Affymetrix Human Clariom D Assay PSR2000166092.hg.1 NM_002237.3 Refseq
Affymetrix Human Clariom D Assay PSR2000166092.hg.1 TC2000009448.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2000166088.hg.1 ENST00000424171.5 Ensembl
Affymetrix Human Clariom D Assay PSR2000166088.hg.1 ENST00000433903.5 Ensembl
Affymetrix Human Clariom D Assay PSR2000166088.hg.1 ENST00000439216.5 Ensembl
Affymetrix Human Clariom D Assay PSR2000166088.hg.1 ENST00000447736.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000166088.hg.1 ENST00000424171.5 Ensembl
Affymetrix Human Clariom D Assay PSR2000166088.hg.1 ENST00000433903.5 Ensembl
Affymetrix Human Clariom D Assay PSR2000166088.hg.1 ENST00000439216.5 Ensembl
Affymetrix Human Clariom D Assay PSR2000166088.hg.1 ENST00000447736.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000166088.hg.1 TC2000009448.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2000166091.hg.1 ENST00000433903.5 Ensembl
Affymetrix Human Clariom D Assay PSR2000166091.hg.1 ENST00000433903.5 Ensembl
Affymetrix Human Clariom D Assay PSR2000166091.hg.1 TC2000009448.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 179345 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0