This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00380997_m1
See other ZYG11B GE Assays ›
Gene Symbol:
ZYG11B
Gene Name:
zyg-11 family member B, cell cycle regulator
Gene Aliases:
ZYG11
Chromosome Location:
Chr.1: 52726454 - 52827342 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00380997_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZYG11B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_024646.2 NP_078922.1 4 - 5 1287 111
XM_006710898.3 XP_006710961.1 4 - 5 1174 111
XM_017002336.1 XP_016857825.1 4 - 5 1292 111
GenBank mRNA AB051517.1 - 4 - 5 1237 111
AK056645.1 - 2 - 3 231 111
AK095643.1 - 3 - 4 927 111
AK296460.1 - 4 - 5 1292 111
BC029832.1 - 2 - 3 301 111
BX647643.1 - 2 - 3 460 111
DQ646393.1 - 4 - 5 1306 111

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100008332.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01000645.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ZYG11B

Gene Ontology Categories:

Function(s) Process(es)

protein ubiquitination
regulation of ubiquitin-protein transferase activity
ubiquitin-protein transferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR01010524.hg.1 ENST00000294353.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01010524.hg.1 ENST00000443756.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01010524.hg.1 ENST00000545132.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01010524.hg.1 NM_024646.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01010524.hg.1 TC01000645.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 ENST00000294353.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 ENST00000294353.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 ENST00000294353.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 ENST00000294353.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 ENST00000294353.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 ENST00000294353.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 ENST00000294353.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 ENST00000294353.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 ENST00000294353.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 ENST00000294353.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 ENST00000294353.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 ENST00000294353.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 ENST00000294353.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 ENST00000294353.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 ENST00000294353.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 ENST00000294353.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 ENST00000545132.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 ENST00000545132.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 ENST00000545132.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 ENST00000545132.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 ENST00000545132.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 ENST00000545132.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 ENST00000545132.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 ENST00000545132.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 ENST00000545132.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 ENST00000545132.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 ENST00000545132.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 ENST00000545132.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 ENST00000545132.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 ENST00000545132.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 ENST00000545132.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 ENST00000545132.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 NM_024646.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 NM_024646.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 NM_024646.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 NM_024646.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 NM_024646.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 NM_024646.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 NM_024646.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 NM_024646.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 NM_024646.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 NM_024646.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 NM_024646.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 NM_024646.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 NM_024646.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 NM_024646.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 NM_024646.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 NM_024646.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008332.hg.1 NM_024646.2 Refseq
Affymetrix Human Clariom D Assay PSR0100159330.hg.1 ENST00000294353.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100159330.hg.1 ENST00000545132.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100159330.hg.1 NM_024646.2 Refseq
Affymetrix Human Clariom D Assay PSR0100159330.hg.1 ENST00000294353.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100159330.hg.1 ENST00000545132.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100159330.hg.1 NM_024646.2 Refseq
Affymetrix Human Clariom D Assay PSR0100159330.hg.1 TC0100008332.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0100008332.hg.1 ENST00000294353.6 Ensembl
Affymetrix Human Clariom D Assay TC0100008332.hg.1 ENST00000294353.6 Ensembl
Affymetrix Human Clariom D Assay TC0100008332.hg.1 ENST00000294353.6 Ensembl
Affymetrix Human Clariom D Assay TC0100008332.hg.1 ENST00000294353.6 Ensembl
Affymetrix Human Clariom D Assay TC0100008332.hg.1 ENST00000294353.6 Ensembl
Affymetrix Human Clariom D Assay TC0100008332.hg.1 ENST00000294353.6 Ensembl
Affymetrix Human Clariom D Assay TC0100008332.hg.1 ENST00000294353.6 Ensembl
Affymetrix Human Clariom D Assay TC0100008332.hg.1 ENST00000294353.6 Ensembl
Affymetrix Human Clariom D Assay TC0100008332.hg.1 ENST00000294353.6 Ensembl
Affymetrix Human Clariom D Assay TC0100008332.hg.1 ENST00000294353.6 Ensembl
Affymetrix Human Clariom D Assay TC0100008332.hg.1 ENST00000294353.6 Ensembl
Affymetrix Human Clariom D Assay TC0100008332.hg.1 ENST00000294353.6 Ensembl
Affymetrix Human Clariom D Assay TC0100008332.hg.1 ENST00000294353.6 Ensembl
Affymetrix Human Clariom D Assay TC0100008332.hg.1 ENST00000294353.6 Ensembl
Affymetrix Human Clariom D Assay TC0100008332.hg.1 ENST00000294353.6 Ensembl
Affymetrix Human Clariom D Assay TC0100008332.hg.1 ENST00000294353.6 Ensembl
Affymetrix Human Clariom D Assay TC0100008332.hg.1 ENST00000545132.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008332.hg.1 ENST00000545132.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008332.hg.1 ENST00000545132.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008332.hg.1 ENST00000545132.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008332.hg.1 ENST00000545132.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008332.hg.1 ENST00000545132.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008332.hg.1 ENST00000545132.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008332.hg.1 ENST00000545132.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008332.hg.1 ENST00000545132.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008332.hg.1 ENST00000545132.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008332.hg.1 ENST00000545132.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008332.hg.1 ENST00000545132.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008332.hg.1 ENST00000545132.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008332.hg.1 ENST00000545132.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008332.hg.1 ENST00000545132.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008332.hg.1 ENST00000545132.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008332.hg.1 NM_024646.2 Refseq
Affymetrix Human Clariom D Assay TC0100008332.hg.1 NM_024646.2 Refseq
Affymetrix Human Clariom D Assay TC0100008332.hg.1 NM_024646.2 Refseq
Affymetrix Human Clariom D Assay TC0100008332.hg.1 NM_024646.2 Refseq
Affymetrix Human Clariom D Assay TC0100008332.hg.1 NM_024646.2 Refseq
Affymetrix Human Clariom D Assay TC0100008332.hg.1 NM_024646.2 Refseq
Affymetrix Human Clariom D Assay TC0100008332.hg.1 NM_024646.2 Refseq
Affymetrix Human Clariom D Assay TC0100008332.hg.1 NM_024646.2 Refseq
Affymetrix Human Clariom D Assay TC0100008332.hg.1 NM_024646.2 Refseq
Affymetrix Human Clariom D Assay TC0100008332.hg.1 NM_024646.2 Refseq
Affymetrix Human Clariom D Assay TC0100008332.hg.1 NM_024646.2 Refseq
Affymetrix Human Clariom D Assay TC0100008332.hg.1 NM_024646.2 Refseq
Affymetrix Human Clariom D Assay TC0100008332.hg.1 NM_024646.2 Refseq
Affymetrix Human Clariom D Assay TC0100008332.hg.1 NM_024646.2 Refseq
Affymetrix Human Clariom D Assay TC0100008332.hg.1 NM_024646.2 Refseq
Affymetrix Human Clariom D Assay TC0100008332.hg.1 NM_024646.2 Refseq
Affymetrix Human Clariom D Assay TC0100008332.hg.1 NM_024646.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0