Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011690.hg.1 |
ENST00000536262.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011690.hg.1 |
ENST00000536262.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011690.hg.1 |
ENST00000536262.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011690.hg.1 |
ENST00000536262.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011690.hg.1 |
ENST00000536262.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011690.hg.1 |
ENST00000536262.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011690.hg.1 |
ENST00000536262.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011690.hg.1 |
ENST00000536262.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011690.hg.1 |
ENST00000536262.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011690.hg.1 |
ENST00000536262.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011690.hg.1 |
ENST00000536262.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011690.hg.1 |
ENST00000536262.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011690.hg.1 |
ENST00000536262.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011690.hg.1 |
ENST00000536262.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011690.hg.1 |
ENST00000536262.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011690.hg.1 |
ENST00000536262.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011690.hg.1 |
ENST00000536262.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011690.hg.1 |
ENST00000536262.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011690.hg.1 |
ENST00000536262.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011690.hg.1 |
ENST00000536262.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011690.hg.1 |
ENST00000536262.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12024519.hg.1 |
ENST00000536262.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12024519.hg.1 |
NM_145913.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12024519.hg.1 |
TC12001873.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200190564.hg.1 |
ENST00000536262.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200190564.hg.1 |
ENST00000536262.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200190564.hg.1 |
TC1200011690.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200011690.hg.1 |
ENST00000536262.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011690.hg.1 |
ENST00000536262.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011690.hg.1 |
ENST00000536262.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011690.hg.1 |
ENST00000536262.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011690.hg.1 |
ENST00000536262.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011690.hg.1 |
ENST00000536262.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011690.hg.1 |
ENST00000536262.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011690.hg.1 |
ENST00000536262.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011690.hg.1 |
ENST00000536262.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011690.hg.1 |
ENST00000536262.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011690.hg.1 |
ENST00000536262.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011690.hg.1 |
ENST00000536262.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011690.hg.1 |
ENST00000536262.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011690.hg.1 |
ENST00000536262.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011690.hg.1 |
ENST00000536262.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011690.hg.1 |
ENST00000536262.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011690.hg.1 |
ENST00000536262.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011690.hg.1 |
ENST00000536262.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011690.hg.1 |
ENST00000536262.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011690.hg.1 |
ENST00000536262.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011690.hg.1 |
ENST00000536262.2 |
Ensembl |