This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00376173_g1
See other DOCK11 GE Assays ›
Gene Symbol:
DOCK11
Gene Name:
dedicator of cytokinesis 11
Gene Aliases:
ACG, ZIZ2, bB128O4.1
Chromosome Location:
Chr.X: 118495664 - 118686162 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00376173_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DOCK11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_144658.3 NP_653259.3 33 - 34 3724 104
XM_005262368.3 XP_005262425.1 34 - 35 4008 104
XM_005262369.4 XP_005262426.1 34 - 35 3702 104
XM_011531276.2 XP_011529578.1 34 - 35 3643 104
XM_011531278.2 XP_011529580.1 21 - 22 2129 104
GenBank mRNA AK056684.1 - 3 - 4 238 104
AK124735.1 - 28 - 29 3147 104
AK125641.1 - 12 - 13 1236 104
AY692226.1 - 33 - 34 3724 104

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0X00008228.hg.1
This assay targets the same location as HTA transcript cluster(s) TC0X000574.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene DOCK11

Panther Classification:

Molecular Function -

guanyl-nucleotide exchange factor

Gene Ontology Categories:

Function(s) Process(es)

small GTPase mediated signal transduction
blood coagulation
positive regulation of GTPase activity
Rho guanyl-nucleotide exchange factor activity
protein binding
Rho GTPase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR0X007601.hg.1 ENST00000276202.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X007601.hg.1 ENST00000276204.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X007601.hg.1 NM_144658.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X007601.hg.1 TC0X000574.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0X00156169.hg.1 ENST00000276202.8 Ensembl
Affymetrix Human Clariom D Assay PSR0X00156169.hg.1 ENST00000276204.10 Ensembl
Affymetrix Human Clariom D Assay PSR0X00156169.hg.1 ENST00000633080.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00156169.hg.1 NM_144658.3 Refseq
Affymetrix Human Clariom D Assay PSR0X00156169.hg.1 ENST00000276202.8 Ensembl
Affymetrix Human Clariom D Assay PSR0X00156169.hg.1 ENST00000276204.10 Ensembl
Affymetrix Human Clariom D Assay PSR0X00156169.hg.1 ENST00000633080.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00156169.hg.1 NM_144658.3 Refseq
Affymetrix Human Clariom D Assay PSR0X00156169.hg.1 TC0X00008228.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header