Assay Details
Gene Symbol: TAF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138572.2 | NP_612639.2 | 5 - 6 | 521 | 67 | |
XM_011514296.1 | XP_011512598.1 | 5 - 6 | 512 | 67 | ||
XM_017010241.1 | XP_016865730.1 | 5 - 6 | 512 | 67 | ||
XM_017010242.1 | XP_016865731.1 | 5 - 6 | 675 | 67 | ||
XM_017010243.1 | XP_016865732.1 | 5 - 6 | 675 | 67 | ||
GenBank mRNA | AF465841.1 | - | 5 - 6 | 495 | 67 | |
AK057383.1 | - | 5 - 6 | 512 | 67 | ||
AK303097.1 | - | 5 - 6 | 679 | 67 | ||
AL833401.1 | - | 5 - 6 | 511 | 67 | ||
BC119678.1 | - | 5 - 6 | 512 | 40116287 | 67 | |
BC119679.1 | - | 5 - 6 | 512 | 40116290 | 67 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06000579.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
inner cell mass cell proliferation
cell differentiation
snRNA transcription from RNA polymerase II promoter
regulation of fat cell differentiation
positive regulation of transcription, DNA-templated
maintenance of protein location in nucleus
protein binding
protein heterodimerization activity
inner cell mass cell proliferation
cell differentiation
snRNA transcription from RNA polymerase II promoter
regulation of fat cell differentiation
positive regulation of transcription, DNA-templated
maintenance of protein location in nucleus
protein binding
protein heterodimerization activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR06006517.hg.1 | ENST00000372977.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06006517.hg.1 | ENST00000372982.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06006517.hg.1 | ENST00000456846.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06006517.hg.1 | ENST00000465926.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06006517.hg.1 | ENST00000494547.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06006517.hg.1 | NM_138572.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06006517.hg.1 | TC06000579.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000372977.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000372977.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000372977.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000372977.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000372977.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000372977.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000372977.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000372977.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000372977.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000372977.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000372977.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000372977.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000372977.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000372978.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000372978.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000372978.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000372978.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000372978.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000372978.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000372982.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000372982.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000372982.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000372982.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000372982.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000372982.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000372982.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000372982.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000372982.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000372982.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000372982.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000372982.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000456846.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000456846.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000456846.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000456846.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000456846.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000456846.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000456846.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000456846.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000456846.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000456846.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000465926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000465926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000465926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000465926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000465926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000465926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000465926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000465926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000465926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000465926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000472818.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000472818.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000472818.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000472818.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000472818.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000482432.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000482432.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000482432.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000482432.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000482926.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000482926.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000482926.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000482926.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000482926.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000482926.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000486070.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000486070.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000486070.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000494547.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000494547.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000494547.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000494547.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000494547.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000494547.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000494547.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000494547.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000494547.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000494547.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000494547.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | ENST00000494547.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | NM_138572.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | NM_138572.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | NM_138572.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | NM_138572.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | NM_138572.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | NM_138572.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | NM_138572.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | NM_138572.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | NM_138572.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | NM_138572.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | NM_138572.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008035.hg.1 | NM_138572.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000372977.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000372977.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000372977.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000372977.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000372977.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000372977.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000372977.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000372977.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000372977.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000372977.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000372977.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000372977.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000372977.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000372978.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000372978.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000372978.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000372978.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000372978.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000372978.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000372982.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000372982.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000372982.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000372982.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000372982.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000372982.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000372982.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000372982.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000372982.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000372982.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000372982.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000372982.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000456846.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000456846.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000456846.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000456846.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000456846.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000456846.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000456846.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000456846.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000456846.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000456846.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000465926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000465926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000465926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000465926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000465926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000465926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000465926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000465926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000465926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000465926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000472818.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000472818.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000472818.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000472818.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000472818.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000482432.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000482432.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000482432.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000482432.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000482926.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000482926.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000482926.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000482926.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000482926.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000482926.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000486070.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000486070.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000486070.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000494547.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000494547.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000494547.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000494547.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000494547.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000494547.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000494547.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000494547.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000494547.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000494547.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000494547.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | ENST00000494547.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | NM_138572.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | NM_138572.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | NM_138572.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | NM_138572.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | NM_138572.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | NM_138572.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | NM_138572.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | NM_138572.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | NM_138572.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | NM_138572.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | NM_138572.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008035.hg.1 | NM_138572.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600155702.hg.1 | ENST00000372977.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600155702.hg.1 | ENST00000372982.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600155702.hg.1 | ENST00000456846.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600155702.hg.1 | ENST00000465926.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600155702.hg.1 | ENST00000494547.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600155702.hg.1 | NM_138572.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600155702.hg.1 | ENST00000372977.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600155702.hg.1 | ENST00000372982.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600155702.hg.1 | ENST00000456846.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600155702.hg.1 | ENST00000465926.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600155702.hg.1 | ENST00000494547.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600155702.hg.1 | NM_138572.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600155702.hg.1 | TC0600008035.hg.1 | Clariom D TC |
