Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04022788.hg.1 |
ENST00000357115.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04022788.hg.1 |
ENST00000503716.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04022788.hg.1 |
ENST00000507224.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04022788.hg.1 |
ENST00000510413.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04022788.hg.1 |
ENST00000535741.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04022788.hg.1 |
NM_001199282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04022788.hg.1 |
NM_006726.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04022788.hg.1 |
TC04001634.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0400183076.hg.1 |
ENST00000357115.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400183076.hg.1 |
ENST00000503716.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400183076.hg.1 |
ENST00000507224.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400183076.hg.1 |
ENST00000509835.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400183076.hg.1 |
ENST00000510413.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400183076.hg.1 |
NM_001199282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400183076.hg.1 |
NM_006726.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400183076.hg.1 |
ENST00000357115.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400183076.hg.1 |
ENST00000503716.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400183076.hg.1 |
ENST00000507224.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400183076.hg.1 |
ENST00000509835.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400183076.hg.1 |
ENST00000510413.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400183076.hg.1 |
NM_001199282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400183076.hg.1 |
NM_006726.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400183076.hg.1 |
TC0400012102.hg.1 |
Clariom D TC |