Assay Details
Gene Symbol: MED12L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053002.5 | NP_443728.3 | 7 - 8 | 1236 | 66 | |
XM_006713487.3 | XP_006713550.1 | 8 - 9 | 1508 | 66 | ||
XM_011512390.2 | XP_011510692.1 | 8 - 9 | 1508 | 66 | ||
XM_011512394.2 | XP_011510696.1 | 8 - 9 | 1508 | 66 | ||
XM_011512399.2 | XP_011510701.1 | 8 - 9 | 1508 | 66 | ||
XM_017005676.1 | XP_016861165.1 | 8 - 9 | 1508 | 66 | ||
XM_017005677.1 | XP_016861166.1 | 8 - 9 | 1508 | 66 | ||
XM_017005678.1 | XP_016861167.1 | 8 - 9 | 1508 | 66 | ||
XM_017005680.1 | XP_016861169.1 | 8 - 9 | 1508 | 66 | ||
GenBank mRNA | AF388364.1 | - | 7 - 8 | 1640 | 66 | |
AF388365.1 | - | 7 - 8 | 1640 | 66 | ||
AF399708.1 | - | 7 - 8 | 1145 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
