This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00359931_g1
See other XPOT GE Assays ›
Gene Symbol:
XPOT
Gene Name:
exportin for tRNA
Gene Aliases:
XPO3
Chromosome Location:
Chr.12: 64404350 - 64451125 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00359931_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: XPOT
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007235.4 NP_009166.2 10 - 11 1619 117
XM_017018748.1 XP_016874237.1 10 - 11 1479 117
GenBank mRNA AF039022.1 - 9 - 10 1113 117
AK292884.1 - 10 - 11 1432 117
BC020569.1 - 9 - 10 1198 4420052 117
Y16414.1 - 9 - 10 1171 117

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1200008002.hg.1
This assay targets the same location as HTA transcript cluster(s) TC12000569.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene XPOT

Panther Classification:

Molecular Function -

RNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

tRNA export from nucleus
intracellular protein transport
tRNA re-export from nucleus
tRNA binding
Ran GTPase binding
nucleobase-containing compound transmembrane transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0