Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002916.hg.1 |
ENST00000330953.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002916.hg.1 |
NM_005508.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002916.hg.1 |
TC03000164.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002917.hg.1 |
ENST00000330953.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002917.hg.1 |
NM_005508.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002917.hg.1 |
TC03000164.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006985.hg.1 |
ENST00000330953.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006985.hg.1 |
ENST00000330953.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006985.hg.1 |
ENST00000330953.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006985.hg.1 |
ENST00000330953.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006985.hg.1 |
ENST00000330953.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006985.hg.1 |
ENST00000330953.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006985.hg.1 |
ENST00000330953.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006985.hg.1 |
ENST00000330953.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006985.hg.1 |
NM_005508.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006985.hg.1 |
NM_005508.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006985.hg.1 |
NM_005508.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006985.hg.1 |
NM_005508.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006985.hg.1 |
NM_005508.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002913.hg.1 |
ENST00000330953.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002913.hg.1 |
NM_005508.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002913.hg.1 |
TC03000164.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0300148586.hg.1 |
ENST00000330953.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300148586.hg.1 |
NM_005508.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300148586.hg.1 |
ENST00000330953.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300148586.hg.1 |
NM_005508.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300148586.hg.1 |
TC0300006985.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300148589.hg.1 |
ENST00000330953.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300148589.hg.1 |
NM_005508.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300148589.hg.1 |
ENST00000330953.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300148589.hg.1 |
NM_005508.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300148589.hg.1 |
TC0300006985.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300148590.hg.1 |
ENST00000330953.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300148590.hg.1 |
NM_005508.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300148590.hg.1 |
ENST00000330953.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300148590.hg.1 |
NM_005508.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300148590.hg.1 |
TC0300006985.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300006985.hg.1 |
ENST00000330953.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006985.hg.1 |
ENST00000330953.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006985.hg.1 |
ENST00000330953.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006985.hg.1 |
ENST00000330953.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006985.hg.1 |
ENST00000330953.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006985.hg.1 |
ENST00000330953.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006985.hg.1 |
ENST00000330953.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006985.hg.1 |
ENST00000330953.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006985.hg.1 |
NM_005508.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006985.hg.1 |
NM_005508.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006985.hg.1 |
NM_005508.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006985.hg.1 |
NM_005508.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006985.hg.1 |
NM_005508.4 |
Refseq |