Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07018204.hg.1 |
ENST00000401412.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07018204.hg.1 |
ENST00000419304.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07018204.hg.1 |
ENST00000419572.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07018204.hg.1 |
NM_006408.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07018204.hg.1 |
TC07001163.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000401412.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000401412.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000401412.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000401412.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000401412.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000401412.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000401412.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000401412.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000401412.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000412973.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000412973.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000412973.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000412973.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000412973.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000412973.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000412973.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000419304.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000419304.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000419304.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000419304.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000419304.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000419304.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000419304.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000419304.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000419304.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000419304.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000419304.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000419304.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000450569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000450569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000450569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000450569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000450569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000450569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000450569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000450569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000468419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000468419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000468419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000468419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000468419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000486219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000486219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000489523.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000489523.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000489523.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000489523.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000489523.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
ENST00000489523.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
NM_006408.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
NM_006408.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
NM_006408.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
NM_006408.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
NM_006408.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
NM_006408.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
NM_006408.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
NM_006408.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
NM_006408.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
NM_006408.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
NM_006408.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010355.hg.1 |
NM_006408.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700173615.hg.1 |
ENST00000401412.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700173615.hg.1 |
ENST00000412973.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700173615.hg.1 |
ENST00000419304.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700173615.hg.1 |
ENST00000468419.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700173615.hg.1 |
ENST00000489523.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700173615.hg.1 |
NM_006408.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700173615.hg.1 |
ENST00000401412.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700173615.hg.1 |
ENST00000412973.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700173615.hg.1 |
ENST00000419304.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700173615.hg.1 |
ENST00000468419.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700173615.hg.1 |
ENST00000489523.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700173615.hg.1 |
NM_006408.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700173615.hg.1 |
TC0700010355.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000401412.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000401412.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000401412.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000401412.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000401412.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000401412.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000401412.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000401412.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000401412.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000412973.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000412973.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000412973.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000412973.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000412973.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000412973.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000412973.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000419304.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000419304.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000419304.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000419304.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000419304.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000419304.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000419304.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000419304.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000419304.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000419304.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000419304.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000419304.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000450569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000450569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000450569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000450569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000450569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000450569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000450569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000450569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000468419.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000468419.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000468419.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000468419.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000468419.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000486219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000486219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000489523.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000489523.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000489523.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000489523.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000489523.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
ENST00000489523.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
NM_006408.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
NM_006408.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
NM_006408.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
NM_006408.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
NM_006408.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
NM_006408.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
NM_006408.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
NM_006408.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
NM_006408.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
NM_006408.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
NM_006408.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010355.hg.1 |
NM_006408.3 |
Refseq |