This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00332222_s1
See other LOC100506473 GE Assays ›
Gene Symbol:
LOC100506473 C2orf49
Gene Name:
uncharacterized LOC100506473
chromosome 2 open reading frame 49
Gene Aliases:
-
asw
Chromosome Location:
Chr.2: 105331542 - 105337278 on Build GRCh38
Chromosome Location:
Chr.2: 105337257 - 105359940 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
-
Assay ID Hs00332222_s1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100506473
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK058000.1 - 1 151 60
Gene Symbol: C2orf49
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017004892.1 XP_016860381.1 1 60

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_017004892.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Inventoried Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

tRNA splicing, via endonucleolytic cleavage and ligation
biological_process
embryonic morphogenesis
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200013749.hg.1 ENST00000610036.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013749.hg.1 ENST00000610036.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013749.hg.1 ENST00000610036.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013749.hg.1 ENST00000610036.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013749.hg.1 ENST00000610036.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013749.hg.1 ENST00000610036.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013749.hg.1 ENST00000610036.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013749.hg.1 ENST00000610036.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013749.hg.1 ENST00000610036.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008734.hg.1 ENST00000258457.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008734.hg.1 ENST00000258457.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008734.hg.1 ENST00000258457.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008734.hg.1 ENST00000258457.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008734.hg.1 ENST00000258457.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008734.hg.1 ENST00000258457.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008734.hg.1 ENST00000258457.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008734.hg.1 ENST00000258457.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008734.hg.1 ENST00000258457.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008734.hg.1 ENST00000258457.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008734.hg.1 ENST00000258457.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008734.hg.1 ENST00000258457.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008734.hg.1 ENST00000410049.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008734.hg.1 ENST00000410049.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008734.hg.1 ENST00000410049.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008734.hg.1 ENST00000410049.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008734.hg.1 ENST00000410049.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008734.hg.1 ENST00000410049.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008734.hg.1 ENST00000410049.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008734.hg.1 ENST00000410049.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008734.hg.1 NM_001286537.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008734.hg.1 NM_001286537.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008734.hg.1 NM_001286537.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008734.hg.1 NM_001286537.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008734.hg.1 NM_001286537.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008734.hg.1 NM_001286537.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008734.hg.1 NM_001286537.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008734.hg.1 NM_001286537.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008734.hg.1 NM_001286537.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008734.hg.1 NM_001286537.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008734.hg.1 NM_024093.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008734.hg.1 NM_024093.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008734.hg.1 NM_024093.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008734.hg.1 NM_024093.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008734.hg.1 NM_024093.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008734.hg.1 NM_024093.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008734.hg.1 NM_024093.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008734.hg.1 NM_024093.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008734.hg.1 NM_024093.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008734.hg.1 NM_024093.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008734.hg.1 NM_024093.2 Refseq
Affymetrix Human Clariom D Assay TC0200008734.hg.1 ENST00000258457.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008734.hg.1 ENST00000258457.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008734.hg.1 ENST00000258457.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008734.hg.1 ENST00000258457.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008734.hg.1 ENST00000258457.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008734.hg.1 ENST00000258457.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008734.hg.1 ENST00000258457.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008734.hg.1 ENST00000258457.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008734.hg.1 ENST00000258457.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008734.hg.1 ENST00000258457.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008734.hg.1 ENST00000258457.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008734.hg.1 ENST00000258457.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008734.hg.1 ENST00000410049.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008734.hg.1 ENST00000410049.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008734.hg.1 ENST00000410049.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008734.hg.1 ENST00000410049.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008734.hg.1 ENST00000410049.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008734.hg.1 ENST00000410049.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008734.hg.1 ENST00000410049.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008734.hg.1 ENST00000410049.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008734.hg.1 NM_001286537.1 Refseq
Affymetrix Human Clariom D Assay TC0200008734.hg.1 NM_001286537.1 Refseq
Affymetrix Human Clariom D Assay TC0200008734.hg.1 NM_001286537.1 Refseq
Affymetrix Human Clariom D Assay TC0200008734.hg.1 NM_001286537.1 Refseq
Affymetrix Human Clariom D Assay TC0200008734.hg.1 NM_001286537.1 Refseq
Affymetrix Human Clariom D Assay TC0200008734.hg.1 NM_001286537.1 Refseq
Affymetrix Human Clariom D Assay TC0200008734.hg.1 NM_001286537.1 Refseq
Affymetrix Human Clariom D Assay TC0200008734.hg.1 NM_001286537.1 Refseq
Affymetrix Human Clariom D Assay TC0200008734.hg.1 NM_001286537.1 Refseq
Affymetrix Human Clariom D Assay TC0200008734.hg.1 NM_001286537.1 Refseq
Affymetrix Human Clariom D Assay TC0200008734.hg.1 NM_024093.2 Refseq
Affymetrix Human Clariom D Assay TC0200008734.hg.1 NM_024093.2 Refseq
Affymetrix Human Clariom D Assay TC0200008734.hg.1 NM_024093.2 Refseq
Affymetrix Human Clariom D Assay TC0200008734.hg.1 NM_024093.2 Refseq
Affymetrix Human Clariom D Assay TC0200008734.hg.1 NM_024093.2 Refseq
Affymetrix Human Clariom D Assay TC0200008734.hg.1 NM_024093.2 Refseq
Affymetrix Human Clariom D Assay TC0200008734.hg.1 NM_024093.2 Refseq
Affymetrix Human Clariom D Assay TC0200008734.hg.1 NM_024093.2 Refseq
Affymetrix Human Clariom D Assay TC0200008734.hg.1 NM_024093.2 Refseq
Affymetrix Human Clariom D Assay TC0200008734.hg.1 NM_024093.2 Refseq
Affymetrix Human Clariom D Assay TC0200008734.hg.1 NM_024093.2 Refseq
Affymetrix Human Clariom D Assay TC0200013749.hg.1 ENST00000610036.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013749.hg.1 ENST00000610036.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013749.hg.1 ENST00000610036.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013749.hg.1 ENST00000610036.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013749.hg.1 ENST00000610036.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013749.hg.1 ENST00000610036.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013749.hg.1 ENST00000610036.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013749.hg.1 ENST00000610036.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013749.hg.1 ENST00000610036.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0