This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00324907_m1
See other ZNF512B GE Assays ›
Gene Symbol:
ZNF512B
Gene Name:
zinc finger protein 512B
Gene Aliases:
GM632
Chromosome Location:
Chr.20: 63956703 - 63969870 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00324907_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZNF512B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_020713.2 NP_065764.1 5 - 6 1096 75
XM_005260226.3 XP_005260283.1 6 - 7 1168 75
XM_011528929.2 XP_011527231.1 5 - 6 1058 75
XM_011528930.2 XP_011527232.1 5 - 6 1285 75
XM_011528933.2 XP_011527235.1 5 - 6 1281 75
GenBank mRNA AB033022.1 - 3 - 4 915 75
BC125128.1 - 5 - 6 1087 40119047 75
BC125129.1 - 5 - 6 1086 40119053 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2000010034.hg.1
This assay targets the same location as HTA transcript cluster(s) TC20001043.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ZNF512B

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
protein binding
metal ion binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR20014324.hg.1 ENST00000217130.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20014324.hg.1 ENST00000369888.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20014324.hg.1 ENST00000450537.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20014324.hg.1 NM_020713.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR20014324.hg.1 TC20001043.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR2000172021.hg.1 ENST00000217130.4 Ensembl
Affymetrix Human Clariom D Assay PSR2000172021.hg.1 ENST00000369888.5 Ensembl
Affymetrix Human Clariom D Assay PSR2000172021.hg.1 NM_020713.2 Refseq
Affymetrix Human Clariom D Assay PSR2000172021.hg.1 ENST00000217130.4 Ensembl
Affymetrix Human Clariom D Assay PSR2000172021.hg.1 ENST00000369888.5 Ensembl
Affymetrix Human Clariom D Assay PSR2000172021.hg.1 NM_020713.2 Refseq
Affymetrix Human Clariom D Assay PSR2000172021.hg.1 TC2000010034.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 123640 - -
ABI Human Genome Survey Microarray Version 2.0 200706 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0