Assay Details
Gene Symbol: TMIGD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169126.1 | NP_001162597.1 | 1 - 2 | 91 | 92 | |
NM_144615.2 | NP_653216.2 | 1 - 2 | 91 | 92 | ||
XM_017026284.1 | XP_016881773.1 | 1 - 2 | 99 | 92 | ||
GenBank mRNA | BC015655.1 | - | 1 - 2 | 91 | 4811204 | 92 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC19001060.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC015655.1,NM_001169126.1,NM_144615.2,XM_017026284.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR19014481.hg.1 | ENST00000301272.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19014481.hg.1 | NM_001169126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19014481.hg.1 | NM_144615.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19014481.hg.1 | TC19001060.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR19014480.hg.1 | ENST00000301272.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19014480.hg.1 | NM_001169126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19014480.hg.1 | NM_144615.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19014480.hg.1 | TC19001060.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1900009330.hg.1 | ENST00000301272.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009330.hg.1 | ENST00000301272.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009330.hg.1 | ENST00000301272.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009330.hg.1 | ENST00000301272.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009330.hg.1 | ENST00000301272.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009330.hg.1 | ENST00000301272.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009330.hg.1 | ENST00000301272.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009330.hg.1 | ENST00000595645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009330.hg.1 | ENST00000595645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009330.hg.1 | ENST00000595645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009330.hg.1 | ENST00000595645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009330.hg.1 | ENST00000595645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009330.hg.1 | ENST00000595645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009330.hg.1 | ENST00000600114.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009330.hg.1 | ENST00000600114.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009330.hg.1 | ENST00000600114.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009330.hg.1 | ENST00000600114.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009330.hg.1 | ENST00000600349.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009330.hg.1 | ENST00000600349.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009330.hg.1 | NM_001169126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009330.hg.1 | NM_001169126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009330.hg.1 | NM_001169126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009330.hg.1 | NM_001169126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009330.hg.1 | NM_001169126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009330.hg.1 | NM_001169126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009330.hg.1 | NM_001169126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009330.hg.1 | NM_001308232.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009330.hg.1 | NM_001308232.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009330.hg.1 | NM_001308232.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009330.hg.1 | NM_001308232.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009330.hg.1 | NM_001308232.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009330.hg.1 | NM_001308232.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009330.hg.1 | NM_144615.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009330.hg.1 | NM_144615.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009330.hg.1 | NM_144615.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009330.hg.1 | NM_144615.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009330.hg.1 | NM_144615.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009330.hg.1 | NM_144615.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009330.hg.1 | NM_144615.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900174543.hg.1 | ENST00000301272.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900174543.hg.1 | ENST00000595645.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900174543.hg.1 | NM_001169126.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900174543.hg.1 | NM_144615.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900174543.hg.1 | ENST00000301272.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900174543.hg.1 | ENST00000595645.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900174543.hg.1 | NM_001169126.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900174543.hg.1 | NM_144615.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900174543.hg.1 | TC1900009330.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900174544.hg.1 | ENST00000301272.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900174544.hg.1 | ENST00000595645.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900174544.hg.1 | ENST00000600114.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900174544.hg.1 | ENST00000600349.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900174544.hg.1 | NM_001169126.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900174544.hg.1 | NM_001308232.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900174544.hg.1 | NM_144615.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900174544.hg.1 | ENST00000301272.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900174544.hg.1 | ENST00000595645.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900174544.hg.1 | ENST00000600114.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900174544.hg.1 | ENST00000600349.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900174544.hg.1 | NM_001169126.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900174544.hg.1 | NM_001308232.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900174544.hg.1 | NM_144615.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900174544.hg.1 | TC1900009330.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900174546.hg.1 | ENST00000301272.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900174546.hg.1 | NM_001169126.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900174546.hg.1 | NM_001308232.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900174546.hg.1 | NM_144615.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900174546.hg.1 | ENST00000301272.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900174546.hg.1 | NM_001169126.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900174546.hg.1 | NM_001308232.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900174546.hg.1 | NM_144615.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900174546.hg.1 | TC1900009330.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1900009330.hg.1 | ENST00000301272.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009330.hg.1 | ENST00000301272.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009330.hg.1 | ENST00000301272.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009330.hg.1 | ENST00000301272.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009330.hg.1 | ENST00000301272.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009330.hg.1 | ENST00000301272.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009330.hg.1 | ENST00000301272.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009330.hg.1 | ENST00000595645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009330.hg.1 | ENST00000595645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009330.hg.1 | ENST00000595645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009330.hg.1 | ENST00000595645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009330.hg.1 | ENST00000595645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009330.hg.1 | ENST00000595645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009330.hg.1 | ENST00000600114.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009330.hg.1 | ENST00000600114.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009330.hg.1 | ENST00000600114.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009330.hg.1 | ENST00000600114.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009330.hg.1 | ENST00000600349.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009330.hg.1 | ENST00000600349.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009330.hg.1 | NM_001169126.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009330.hg.1 | NM_001169126.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009330.hg.1 | NM_001169126.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009330.hg.1 | NM_001169126.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009330.hg.1 | NM_001169126.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009330.hg.1 | NM_001169126.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009330.hg.1 | NM_001169126.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009330.hg.1 | NM_001308232.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009330.hg.1 | NM_001308232.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009330.hg.1 | NM_001308232.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009330.hg.1 | NM_001308232.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009330.hg.1 | NM_001308232.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009330.hg.1 | NM_001308232.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009330.hg.1 | NM_144615.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009330.hg.1 | NM_144615.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009330.hg.1 | NM_144615.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009330.hg.1 | NM_144615.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009330.hg.1 | NM_144615.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009330.hg.1 | NM_144615.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009330.hg.1 | NM_144615.2 | Refseq |
