This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00271044_s1
See other GPR15 GE Assays ›
Gene Symbol:
GPR15
Gene Name:
G protein-coupled receptor 15
Gene Aliases:
BOB
Chromosome Location:
Chr.3: 98531899 - 98534679 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00271044_s1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR15
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005290.3 NP_005281.1 1 946 78
GenBank mRNA AK313375.1 - 1 946 78
BC069437.1 - 1 811 7262234 78
BC101779.1 - 1 837 8069285 78
BC101781.1 - 1 834 8069287 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300008114.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03000494.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GPR15

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
T cell migration
G-protein coupled receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR03009329.hg.1 ENST00000284311.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03009329.hg.1 NM_005290.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03009329.hg.1 TC03000494.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0300008114.hg.1 ENST00000284311.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008114.hg.1 ENST00000284311.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008114.hg.1 ENST00000284311.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008114.hg.1 NM_005290.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008114.hg.1 NM_005290.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008114.hg.1 NM_005290.3 Refseq
Affymetrix Human Clariom D Assay PSR0300157764.hg.1 ENST00000284311.4 Ensembl
Affymetrix Human Clariom D Assay PSR0300157764.hg.1 NM_005290.3 Refseq
Affymetrix Human Clariom D Assay PSR0300157764.hg.1 ENST00000284311.4 Ensembl
Affymetrix Human Clariom D Assay PSR0300157764.hg.1 NM_005290.3 Refseq
Affymetrix Human Clariom D Assay PSR0300157764.hg.1 TC0300008114.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0300157761.hg.1 ENST00000284311.4 Ensembl
Affymetrix Human Clariom D Assay PSR0300157761.hg.1 NM_005290.3 Refseq
Affymetrix Human Clariom D Assay PSR0300157761.hg.1 ENST00000284311.4 Ensembl
Affymetrix Human Clariom D Assay PSR0300157761.hg.1 NM_005290.3 Refseq
Affymetrix Human Clariom D Assay PSR0300157761.hg.1 TC0300008114.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0300008114.hg.1 ENST00000284311.4 Ensembl
Affymetrix Human Clariom D Assay TC0300008114.hg.1 ENST00000284311.4 Ensembl
Affymetrix Human Clariom D Assay TC0300008114.hg.1 ENST00000284311.4 Ensembl
Affymetrix Human Clariom D Assay TC0300008114.hg.1 NM_005290.3 Refseq
Affymetrix Human Clariom D Assay TC0300008114.hg.1 NM_005290.3 Refseq
Affymetrix Human Clariom D Assay TC0300008114.hg.1 NM_005290.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0