This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00270172_m1
See other KCNQ2 GE Assays ›
Gene Symbol:
KCNQ2
Gene Name:
potassium voltage-gated channel subfamily Q member 2
Gene Aliases:
BFNC, EBN, EBN1, ENB1, HNSPC, KCNA11, KV7.2
Chromosome Location:
Chr.20: 63400208 - 63472677 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00270172_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: KCNQ2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA Y15065.1 - 15 - 16 2777 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Homo sapiens Inventoried MicroArray Quality Control (MAQC) Probe spans exons

Panther Classification:

Molecular Function -

voltage-gated ion channel

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0