This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00263152_m1
See other PCDH11X GE Assays ›
Gene Symbol:
PCDH11X
Gene Name:
protocadherin 11 X-linked
Gene Aliases:
PCDH-X, PCDH11, PCDHX, PPP1R119
Chromosome Location:
Chr.X: 91779310 - 92623230 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00263152_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PCDH11X
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001168360.1 NP_001161832.1 4 - 5 3193 64
NM_032968.4 NP_116750.1 4 - 5 3193 64
XM_011530910.1 XP_011529212.1 7 - 8 3668 64
XM_011530911.2 XP_011529213.1 9 - 10 3956 64
XM_011530912.2 XP_011529214.1 8 - 9 3902 64
XM_011530914.2 XP_011529216.1 5 - 6 3568 64
XM_017029416.1 XP_016884905.1 4 - 5 3177 64
XM_017029417.1 XP_016884906.1 8 - 9 3902 64
GenBank mRNA AB037747.1 - 3 - 4 381 64
AF206516.1 - 4 - 5 3149 64
AF332218.1 - 4 - 5 3994 64
AY861432.1 - 4 - 5 3149 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0X00007834.hg.1

Set Membership:

Amplicon length less than or equal to 70 3' Most Homo sapiens Inventoried Probe spans exons

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

homophilic cell adhesion via plasma membrane adhesion molecules
negative regulation of phosphatase activity
calcium ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000298274.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000298274.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000298274.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000298274.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000298274.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000298274.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000298274.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000298274.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000361655.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000361655.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000361655.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000361655.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000361655.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000361655.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000361724.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000361724.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000361724.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000361724.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000361724.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000373088.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000373088.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000373088.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000373088.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000373088.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000373094.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000373094.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000373094.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000373094.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000373094.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000373094.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000373094.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000373094.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000373094.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000373094.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000373097.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000373097.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000373097.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000373097.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000373097.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000373097.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000373097.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000373097.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000373097.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000395337.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000395337.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000395337.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000395337.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000395337.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000395337.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000406881.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000406881.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000406881.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000406881.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000406881.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000406881.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000406881.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000504220.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000504220.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000504220.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000504220.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000504220.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000504220.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 ENST00000504220.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_001168360.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_001168360.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_001168360.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_001168360.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_001168360.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_001168360.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_001168360.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_001168360.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_001168360.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_001168361.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_001168361.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_001168361.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_001168361.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_001168361.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_001168361.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_001168361.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_001168361.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_001168361.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_001168362.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_001168362.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_001168362.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_001168362.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_001168362.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_001168362.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_001168362.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_001168363.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_001168363.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_001168363.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_001168363.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_001168363.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_001168363.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_001168363.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_001168363.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_032968.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_032968.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_032968.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_032968.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_032968.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_032968.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_032968.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_032968.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_032968.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_032969.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_032969.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_032969.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_032969.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_032969.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_032969.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_032969.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007834.hg.1 NM_032969.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0