This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00257456_s1
See other OBSCN GE Assays ›
Gene Symbol:
OBSCN
Gene Name:
obscurin, cytoskeletal calmodulin and titin-interacting RhoGEF
Gene Aliases:
ARHGEF30, UNC89
Chromosome Location:
Chr.1: 228208013 - 228381431 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00257456_s1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: OBSCN
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK024186.1 - 1 1248 63
BC098118.1 - 1 810 63
BC098262.1 - 1 810 63
BC098292.1 - 1 810 63
BC099731.1 - 1 810 63
BC114382.1 - 1 810 63
BC114461.1 - 1 810 63

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200009809.hg.1 ENST00000436506.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009809.hg.1 ENST00000436506.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009809.hg.1 ENST00000436506.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009809.hg.1 ENST00000436506.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009809.hg.1 ENST00000436506.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009809.hg.1 ENST00000439050.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009809.hg.1 ENST00000439050.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009809.hg.1 ENST00000439050.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009809.hg.1 ENST00000439050.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009809.hg.1 ENST00000439050.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009809.hg.1 ENST00000439050.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009809.hg.1 ENST00000439050.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009809.hg.1 ENST00000439050.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009809.hg.1 ENST00000439050.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009809.hg.1 NR_110163.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009809.hg.1 NR_110163.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009809.hg.1 NR_110163.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009809.hg.1 NR_110163.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009809.hg.1 NR_110163.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009809.hg.1 NR_110163.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009809.hg.1 NR_110163.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009809.hg.1 NR_110163.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009809.hg.1 NR_110163.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009809.hg.1 NR_110163.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009809.hg.1 NR_110164.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009809.hg.1 NR_110164.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009809.hg.1 NR_110164.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009809.hg.1 NR_110164.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009809.hg.1 NR_110164.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009809.hg.1 NR_110164.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009809.hg.1 NR_110164.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009809.hg.1 NR_110164.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009809.hg.1 NR_110164.1 Refseq
Affymetrix Human Clariom D Assay TC0200009809.hg.1 ENST00000436506.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009809.hg.1 ENST00000436506.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009809.hg.1 ENST00000436506.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009809.hg.1 ENST00000436506.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009809.hg.1 ENST00000436506.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009809.hg.1 ENST00000439050.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009809.hg.1 ENST00000439050.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009809.hg.1 ENST00000439050.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009809.hg.1 ENST00000439050.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009809.hg.1 ENST00000439050.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009809.hg.1 ENST00000439050.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009809.hg.1 ENST00000439050.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009809.hg.1 ENST00000439050.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009809.hg.1 ENST00000439050.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009809.hg.1 NR_110163.1 Refseq
Affymetrix Human Clariom D Assay TC0200009809.hg.1 NR_110163.1 Refseq
Affymetrix Human Clariom D Assay TC0200009809.hg.1 NR_110163.1 Refseq
Affymetrix Human Clariom D Assay TC0200009809.hg.1 NR_110163.1 Refseq
Affymetrix Human Clariom D Assay TC0200009809.hg.1 NR_110163.1 Refseq
Affymetrix Human Clariom D Assay TC0200009809.hg.1 NR_110163.1 Refseq
Affymetrix Human Clariom D Assay TC0200009809.hg.1 NR_110163.1 Refseq
Affymetrix Human Clariom D Assay TC0200009809.hg.1 NR_110163.1 Refseq
Affymetrix Human Clariom D Assay TC0200009809.hg.1 NR_110163.1 Refseq
Affymetrix Human Clariom D Assay TC0200009809.hg.1 NR_110163.1 Refseq
Affymetrix Human Clariom D Assay TC0200009809.hg.1 NR_110164.1 Refseq
Affymetrix Human Clariom D Assay TC0200009809.hg.1 NR_110164.1 Refseq
Affymetrix Human Clariom D Assay TC0200009809.hg.1 NR_110164.1 Refseq
Affymetrix Human Clariom D Assay TC0200009809.hg.1 NR_110164.1 Refseq
Affymetrix Human Clariom D Assay TC0200009809.hg.1 NR_110164.1 Refseq
Affymetrix Human Clariom D Assay TC0200009809.hg.1 NR_110164.1 Refseq
Affymetrix Human Clariom D Assay TC0200009809.hg.1 NR_110164.1 Refseq
Affymetrix Human Clariom D Assay TC0200009809.hg.1 NR_110164.1 Refseq
Affymetrix Human Clariom D Assay TC0200009809.hg.1 NR_110164.1 Refseq
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 214216 - -
ABI Human Genome Survey Microarray Version 2.0 230395 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0