This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00244504_m1
See other ANXA7 GE Assays ›
Gene Symbol:
ANXA7
Gene Name:
annexin A7
Gene Aliases:
ANX7, SNX, SYNEXIN
Chromosome Location:
Chr.10: 73375101 - 73414085 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00244504_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ANXA7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001320880.1 NP_001307809.1 5 - 6 521 80
NM_004034.3 NP_004025.1 6 - 7 576 80
XM_017016162.1 XP_016871651.1 6 - 7 612 80
GenBank mRNA AK295319.1 - 5 - 6 387 80
AK312447.1 - 6 - 7 549 80
DA131184.1 - 5 - 6 494 80

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000011020.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10001408.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ANXA7

Panther Classification:

Molecular Function -

calcium-binding protein

Gene Ontology Categories:

Function(s) Process(es)

cellular calcium ion homeostasis
autophagy
hemostasis
cell proliferation
regulation of cell shape
response to salt stress
cellular water homeostasis
negative regulation of gene expression
response to organic cyclic compound
epithelial cell differentiation
social behavior
response to calcium ion
membrane fusion
integrin binding
calcium ion binding
protein binding
calcium-dependent phospholipid binding
poly(A) RNA binding
calcium-dependent protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000394847.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000394847.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000394847.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000394847.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000394847.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000394847.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000463788.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000463788.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000463788.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000463788.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000463788.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000492380.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000492380.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000492380.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000492380.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000492380.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000492380.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000492380.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000492380.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000492380.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10017755.hg.1 ENST00000372919.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10017755.hg.1 ENST00000372921.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10017755.hg.1 ENST00000492380.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10017755.hg.1 ENST00000535178.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10017755.hg.1 NM_001156.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10017755.hg.1 NM_004034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10017755.hg.1 TC10001408.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000394847.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000394847.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000394847.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000394847.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000394847.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000394847.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000463788.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000463788.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000463788.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000463788.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000463788.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000492380.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000492380.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000492380.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000492380.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000492380.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000492380.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000492380.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000492380.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000492380.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Clariom D Assay PSR1000175037.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Clariom D Assay PSR1000175037.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Clariom D Assay PSR1000175037.hg.1 ENST00000492380.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000175037.hg.1 NM_001156.4 Refseq
Affymetrix Human Clariom D Assay PSR1000175037.hg.1 NM_004034.3 Refseq
Affymetrix Human Clariom D Assay PSR1000175037.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Clariom D Assay PSR1000175037.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Clariom D Assay PSR1000175037.hg.1 ENST00000492380.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000175037.hg.1 NM_001156.4 Refseq
Affymetrix Human Clariom D Assay PSR1000175037.hg.1 NM_004034.3 Refseq
Affymetrix Human Clariom D Assay PSR1000175037.hg.1 TC1000011020.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0