This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00222404_m1
See other MRPS35 GE Assays ›
Gene Symbol:
MRPS35
Gene Name:
mitochondrial ribosomal protein S35
Gene Aliases:
HDCMD11P, MDS023, MRP-S28, MRPS28
Chromosome Location:
Chr.12: 27710773 - 27756304 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00222404_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MRPS35
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001190864.1 NP_001177793.1 1 - 2 185 89
NM_021821.3 NP_068593.2 1 - 2 185 89
XM_017019780.1 XP_016875269.1 1 - 2 185 89
GenBank mRNA AF182422.1 - 1 - 2 162 89
AK075378.1 - 1 - 2 125 89
AK075515.1 - 1 - 2 123 89
BC015862.1 - 1 - 2 130 4689849 89
BC028346.2 - 1 - 2 102 89
BC063515.1 - 1 - 2 108 89
BC109372.1 - 1 - 2 122 6715646 89
DA747094.1 - 1 - 2 185 89

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1200007161.hg.1
This assay targets the same location as HTA transcript cluster(s) TC12000270.hg.1

Additional Information:

This assay is in the 5'UTR region of the AF182422.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MRPS35

Panther Classification:

Molecular Function -

ribosomal protein translational protein

Gene Ontology Categories:

Function(s) Process(es)

DNA damage response, detection of DNA damage
mitochondrial translational elongation
mitochondrial translational termination
structural constituent of ribosome
poly(A) RNA binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR12003298.hg.1 ENST00000081029.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12003298.hg.1 ENST00000321446.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12003298.hg.1 ENST00000538315.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12003298.hg.1 NM_001190864.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12003298.hg.1 NM_021821.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12003298.hg.1 TC12000270.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 ENST00000081029.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 ENST00000081029.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 ENST00000081029.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 ENST00000081029.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 ENST00000081029.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 ENST00000081029.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 ENST00000081029.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 ENST00000081029.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 ENST00000081029.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 ENST00000081029.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 ENST00000081029.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 ENST00000081029.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 ENST00000081029.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 ENST00000536569.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 ENST00000536569.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 ENST00000536569.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 ENST00000536569.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 ENST00000536569.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 ENST00000536569.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 ENST00000538315.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 ENST00000538315.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 ENST00000538315.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 ENST00000538315.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 ENST00000538315.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 ENST00000538315.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 ENST00000538315.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 ENST00000538315.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 ENST00000538315.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 ENST00000538315.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 ENST00000542199.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 ENST00000542199.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 ENST00000542199.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 ENST00000542199.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 ENST00000542199.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 ENST00000542199.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 ENST00000542199.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 ENST00000542199.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 ENST00000542199.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 ENST00000542791.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 ENST00000542791.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 ENST00000542791.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 ENST00000542791.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 NM_001190864.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 NM_001190864.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 NM_001190864.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 NM_001190864.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 NM_001190864.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 NM_001190864.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 NM_001190864.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 NM_001190864.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 NM_001190864.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 NM_001190864.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 NM_001190864.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 NM_021821.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 NM_021821.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 NM_021821.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 NM_021821.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 NM_021821.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 NM_021821.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 NM_021821.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 NM_021821.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 NM_021821.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 NM_021821.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 NM_021821.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 NM_021821.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007161.hg.1 NM_021821.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12003297.hg.1 ENST00000081029.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12003297.hg.1 ENST00000321446.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12003297.hg.1 ENST00000538315.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12003297.hg.1 NM_001190864.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12003297.hg.1 NM_021821.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12003297.hg.1 TC12000270.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR12003293.hg.1 ENST00000081029.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12003293.hg.1 NM_001190864.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12003293.hg.1 NM_021821.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12003293.hg.1 TC12000270.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR12003296.hg.1 ENST00000081029.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12003296.hg.1 ENST00000321446.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12003296.hg.1 ENST00000538315.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12003296.hg.1 NM_001190864.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12003296.hg.1 NM_021821.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12003296.hg.1 TC12000270.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1200150296.hg.1 ENST00000081029.7 Ensembl
Affymetrix Human Clariom D Assay PSR1200150296.hg.1 ENST00000538315.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200150296.hg.1 ENST00000542199.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200150296.hg.1 NM_001190864.1 Refseq
Affymetrix Human Clariom D Assay PSR1200150296.hg.1 NM_021821.3 Refseq
Affymetrix Human Clariom D Assay PSR1200150296.hg.1 ENST00000081029.7 Ensembl
Affymetrix Human Clariom D Assay PSR1200150296.hg.1 ENST00000538315.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200150296.hg.1 ENST00000542199.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200150296.hg.1 NM_001190864.1 Refseq
Affymetrix Human Clariom D Assay PSR1200150296.hg.1 NM_021821.3 Refseq
Affymetrix Human Clariom D Assay PSR1200150296.hg.1 TC1200007161.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1200150287.hg.1 ENST00000081029.7 Ensembl
Affymetrix Human Clariom D Assay PSR1200150287.hg.1 NM_001190864.1 Refseq
Affymetrix Human Clariom D Assay PSR1200150287.hg.1 NM_021821.3 Refseq
Affymetrix Human Clariom D Assay PSR1200150287.hg.1 ENST00000081029.7 Ensembl
Affymetrix Human Clariom D Assay PSR1200150287.hg.1 NM_001190864.1 Refseq
Affymetrix Human Clariom D Assay PSR1200150287.hg.1 NM_021821.3 Refseq
Affymetrix Human Clariom D Assay PSR1200150287.hg.1 TC1200007161.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1200150293.hg.1 ENST00000081029.7 Ensembl
Affymetrix Human Clariom D Assay PSR1200150293.hg.1 ENST00000536569.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200150293.hg.1 ENST00000538315.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200150293.hg.1 ENST00000542791.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200150293.hg.1 NM_001190864.1 Refseq
Affymetrix Human Clariom D Assay PSR1200150293.hg.1 NM_021821.3 Refseq
Affymetrix Human Clariom D Assay PSR1200150293.hg.1 ENST00000081029.7 Ensembl
Affymetrix Human Clariom D Assay PSR1200150293.hg.1 ENST00000536569.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200150293.hg.1 ENST00000538315.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200150293.hg.1 ENST00000542791.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200150293.hg.1 NM_001190864.1 Refseq
Affymetrix Human Clariom D Assay PSR1200150293.hg.1 NM_021821.3 Refseq
Affymetrix Human Clariom D Assay PSR1200150293.hg.1 TC1200007161.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1200007161.hg.1 ENST00000081029.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007161.hg.1 ENST00000081029.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007161.hg.1 ENST00000081029.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007161.hg.1 ENST00000081029.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007161.hg.1 ENST00000081029.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007161.hg.1 ENST00000081029.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007161.hg.1 ENST00000081029.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007161.hg.1 ENST00000081029.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007161.hg.1 ENST00000081029.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007161.hg.1 ENST00000081029.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007161.hg.1 ENST00000081029.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007161.hg.1 ENST00000081029.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007161.hg.1 ENST00000081029.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007161.hg.1 ENST00000536569.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007161.hg.1 ENST00000536569.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007161.hg.1 ENST00000536569.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007161.hg.1 ENST00000536569.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007161.hg.1 ENST00000536569.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007161.hg.1 ENST00000536569.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007161.hg.1 ENST00000538315.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007161.hg.1 ENST00000538315.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007161.hg.1 ENST00000538315.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007161.hg.1 ENST00000538315.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007161.hg.1 ENST00000538315.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007161.hg.1 ENST00000538315.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007161.hg.1 ENST00000538315.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007161.hg.1 ENST00000538315.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007161.hg.1 ENST00000538315.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007161.hg.1 ENST00000538315.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007161.hg.1 ENST00000542199.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007161.hg.1 ENST00000542199.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007161.hg.1 ENST00000542199.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007161.hg.1 ENST00000542199.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007161.hg.1 ENST00000542199.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007161.hg.1 ENST00000542199.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007161.hg.1 ENST00000542199.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007161.hg.1 ENST00000542199.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007161.hg.1 ENST00000542199.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007161.hg.1 ENST00000542791.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007161.hg.1 ENST00000542791.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007161.hg.1 ENST00000542791.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007161.hg.1 ENST00000542791.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007161.hg.1 NM_001190864.1 Refseq
Affymetrix Human Clariom D Assay TC1200007161.hg.1 NM_001190864.1 Refseq
Affymetrix Human Clariom D Assay TC1200007161.hg.1 NM_001190864.1 Refseq
Affymetrix Human Clariom D Assay TC1200007161.hg.1 NM_001190864.1 Refseq
Affymetrix Human Clariom D Assay TC1200007161.hg.1 NM_001190864.1 Refseq
Affymetrix Human Clariom D Assay TC1200007161.hg.1 NM_001190864.1 Refseq
Affymetrix Human Clariom D Assay TC1200007161.hg.1 NM_001190864.1 Refseq
Affymetrix Human Clariom D Assay TC1200007161.hg.1 NM_001190864.1 Refseq
Affymetrix Human Clariom D Assay TC1200007161.hg.1 NM_001190864.1 Refseq
Affymetrix Human Clariom D Assay TC1200007161.hg.1 NM_001190864.1 Refseq
Affymetrix Human Clariom D Assay TC1200007161.hg.1 NM_001190864.1 Refseq
Affymetrix Human Clariom D Assay TC1200007161.hg.1 NM_021821.3 Refseq
Affymetrix Human Clariom D Assay TC1200007161.hg.1 NM_021821.3 Refseq
Affymetrix Human Clariom D Assay TC1200007161.hg.1 NM_021821.3 Refseq
Affymetrix Human Clariom D Assay TC1200007161.hg.1 NM_021821.3 Refseq
Affymetrix Human Clariom D Assay TC1200007161.hg.1 NM_021821.3 Refseq
Affymetrix Human Clariom D Assay TC1200007161.hg.1 NM_021821.3 Refseq
Affymetrix Human Clariom D Assay TC1200007161.hg.1 NM_021821.3 Refseq
Affymetrix Human Clariom D Assay TC1200007161.hg.1 NM_021821.3 Refseq
Affymetrix Human Clariom D Assay TC1200007161.hg.1 NM_021821.3 Refseq
Affymetrix Human Clariom D Assay TC1200007161.hg.1 NM_021821.3 Refseq
Affymetrix Human Clariom D Assay TC1200007161.hg.1 NM_021821.3 Refseq
Affymetrix Human Clariom D Assay TC1200007161.hg.1 NM_021821.3 Refseq
Affymetrix Human Clariom D Assay TC1200007161.hg.1 NM_021821.3 Refseq
Affymetrix Human Clariom D Assay PSR1200150298.hg.1 ENST00000081029.7 Ensembl
Affymetrix Human Clariom D Assay PSR1200150298.hg.1 ENST00000536569.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200150298.hg.1 ENST00000538315.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200150298.hg.1 ENST00000542199.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200150298.hg.1 NM_001190864.1 Refseq
Affymetrix Human Clariom D Assay PSR1200150298.hg.1 NM_021821.3 Refseq
Affymetrix Human Clariom D Assay PSR1200150298.hg.1 ENST00000081029.7 Ensembl
Affymetrix Human Clariom D Assay PSR1200150298.hg.1 ENST00000536569.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200150298.hg.1 ENST00000538315.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200150298.hg.1 ENST00000542199.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200150298.hg.1 NM_001190864.1 Refseq
Affymetrix Human Clariom D Assay PSR1200150298.hg.1 NM_021821.3 Refseq
Affymetrix Human Clariom D Assay PSR1200150298.hg.1 TC1200007161.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1200150294.hg.1 ENST00000081029.7 Ensembl
Affymetrix Human Clariom D Assay PSR1200150294.hg.1 ENST00000536569.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200150294.hg.1 ENST00000538315.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200150294.hg.1 ENST00000542199.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200150294.hg.1 ENST00000542791.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200150294.hg.1 NM_001190864.1 Refseq
Affymetrix Human Clariom D Assay PSR1200150294.hg.1 NM_021821.3 Refseq
Affymetrix Human Clariom D Assay PSR1200150294.hg.1 ENST00000081029.7 Ensembl
Affymetrix Human Clariom D Assay PSR1200150294.hg.1 ENST00000536569.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200150294.hg.1 ENST00000538315.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200150294.hg.1 ENST00000542199.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200150294.hg.1 ENST00000542791.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200150294.hg.1 NM_001190864.1 Refseq
Affymetrix Human Clariom D Assay PSR1200150294.hg.1 NM_021821.3 Refseq
Affymetrix Human Clariom D Assay PSR1200150294.hg.1 TC1200007161.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 122196 - -
ABI Human Genome Survey Microarray Version 2.0 234938 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0