This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00200681_m1
See other GPR182 GE Assays ›
Gene Symbol:
GPR182
Gene Name:
G protein-coupled receptor 182
Gene Aliases:
7TMR, ADMR, AM-R, AMR, G10D, gamrh, hrhAMR
Chromosome Location:
Chr.12: 56993895 - 56998441 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00200681_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR182
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007264.3 NP_009195.1 1 - 2 77 107
XM_011537824.2 XP_011536126.1 1 - 2 753 107
XM_011537825.2 XP_011536127.1 1 - 2 753 107
XM_011537827.2 XP_011536129.1 1 - 2 753 107
GenBank mRNA BC034761.1 - 1 - 2 202 5185932 107
Y13583.1 - 1 - 2 77 107

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1200007853.hg
This assay targets the same location as HTA transcript cluster(s) TC12000519.hg

Additional Information:

This assay is in the 5'UTR region of the BC034761.1,NM_007264.3,XM_011537824.2,XM_011537825.2,XM_011537827.2,Y13583.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GPR182

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

response to hypoxia
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
intracellular signal transduction
adrenomedullin receptor activity
transmembrane signaling receptor activity
G-protein coupled receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR12006787.hg.1 ENST00000300098.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12006787.hg.1 NM_007264.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12006787.hg.1 TC12000519.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR12006785.hg.1 ENST00000300098.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12006785.hg.1 NM_007264.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12006785.hg.1 TC12000519.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1200007853.hg.1 ENST00000300098.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007853.hg.1 ENST00000300098.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007853.hg.1 ENST00000300098.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007853.hg.1 ENST00000300098.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007853.hg.1 ENST00000300098.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007853.hg.1 ENST00000300098.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007853.hg.1 ENST00000300098.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007853.hg.1 ENST00000300098.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007853.hg.1 ENST00000300098.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007853.hg.1 ENST00000300098.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007853.hg.1 ENST00000556850.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007853.hg.1 ENST00000556850.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007853.hg.1 ENST00000556850.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007853.hg.1 ENST00000622922.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007853.hg.1 NM_007264.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007853.hg.1 NM_007264.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007853.hg.1 NM_007264.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007853.hg.1 NM_007264.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007853.hg.1 NM_007264.3 Refseq
Affymetrix Human Clariom D Assay PSR1200157085.hg.1 ENST00000300098.2 Ensembl
Affymetrix Human Clariom D Assay PSR1200157085.hg.1 NM_007264.3 Refseq
Affymetrix Human Clariom D Assay PSR1200157085.hg.1 ENST00000300098.2 Ensembl
Affymetrix Human Clariom D Assay PSR1200157085.hg.1 NM_007264.3 Refseq
Affymetrix Human Clariom D Assay PSR1200157085.hg.1 TC1200007853.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1200007853.hg.1 ENST00000300098.2 Ensembl
Affymetrix Human Clariom D Assay TC1200007853.hg.1 ENST00000300098.2 Ensembl
Affymetrix Human Clariom D Assay TC1200007853.hg.1 ENST00000300098.2 Ensembl
Affymetrix Human Clariom D Assay TC1200007853.hg.1 ENST00000300098.2 Ensembl
Affymetrix Human Clariom D Assay TC1200007853.hg.1 ENST00000300098.2 Ensembl
Affymetrix Human Clariom D Assay TC1200007853.hg.1 ENST00000300098.2 Ensembl
Affymetrix Human Clariom D Assay TC1200007853.hg.1 ENST00000300098.2 Ensembl
Affymetrix Human Clariom D Assay TC1200007853.hg.1 ENST00000300098.2 Ensembl
Affymetrix Human Clariom D Assay TC1200007853.hg.1 ENST00000300098.2 Ensembl
Affymetrix Human Clariom D Assay TC1200007853.hg.1 ENST00000300098.2 Ensembl
Affymetrix Human Clariom D Assay TC1200007853.hg.1 ENST00000556850.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007853.hg.1 ENST00000556850.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007853.hg.1 ENST00000556850.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007853.hg.1 ENST00000622922.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007853.hg.1 NM_007264.3 Refseq
Affymetrix Human Clariom D Assay TC1200007853.hg.1 NM_007264.3 Refseq
Affymetrix Human Clariom D Assay TC1200007853.hg.1 NM_007264.3 Refseq
Affymetrix Human Clariom D Assay TC1200007853.hg.1 NM_007264.3 Refseq
Affymetrix Human Clariom D Assay TC1200007853.hg.1 NM_007264.3 Refseq
Affymetrix Human Clariom D Assay PSR1200157082.hg.1 ENST00000300098.2 Ensembl
Affymetrix Human Clariom D Assay PSR1200157082.hg.1 ENST00000300098.2 Ensembl
Affymetrix Human Clariom D Assay PSR1200157082.hg.1 TC1200007853.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1200157083.hg.1 ENST00000300098.2 Ensembl
Affymetrix Human Clariom D Assay PSR1200157083.hg.1 NM_007264.3 Refseq
Affymetrix Human Clariom D Assay PSR1200157083.hg.1 ENST00000300098.2 Ensembl
Affymetrix Human Clariom D Assay PSR1200157083.hg.1 NM_007264.3 Refseq
Affymetrix Human Clariom D Assay PSR1200157083.hg.1 TC1200007853.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0