This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00198370_m1
See other VAMP5 GE Assays ›
Gene Symbol:
VAMP5
Gene Name:
vesicle associated membrane protein 5
Gene Aliases:
-
Chromosome Location:
Chr.2: 85584408 - 85593388 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00198370_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: VAMP5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_006634.2 NP_006625.1 1 - 2 91 80
GenBank mRNA AF054825.1 - 1 - 2 80
AK292660.1 - 1 - 2 89 80
BC017891.1 - 1 - 2 91 4617039 80

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200008261.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02000513.hg.1

Additional Information:

This assay is in the 5'UTR region of the AF054825.1,AK292660.1,BC017891.1,NM_006634.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene VAMP5

Gene Ontology Categories:

Function(s) Process(es)

exocytosis
vesicle fusion
muscle organ development
skeletal muscle tissue development
cell differentiation
Golgi to plasma membrane protein transport
SNARE binding
SNAP receptor activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR02007899.hg.1 ENST00000306384.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02007899.hg.1 NM_006634.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02007899.hg.1 TC02000513.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0200008261.hg.1 ENST00000306384.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008261.hg.1 ENST00000306384.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008261.hg.1 ENST00000306384.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008261.hg.1 ENST00000306384.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008261.hg.1 ENST00000306384.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008261.hg.1 ENST00000462451.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008261.hg.1 ENST00000462451.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008261.hg.1 ENST00000462451.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008261.hg.1 ENST00000462451.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008261.hg.1 NM_006634.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008261.hg.1 NM_006634.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008261.hg.1 NM_006634.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008261.hg.1 NM_006634.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008261.hg.1 NM_006634.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02007897.hg.1 ENST00000306384.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02007897.hg.1 NM_006634.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02007897.hg.1 TC02000513.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC0200008261.hg.1 ENST00000306384.4 Ensembl
Affymetrix Human Clariom D Assay TC0200008261.hg.1 ENST00000306384.4 Ensembl
Affymetrix Human Clariom D Assay TC0200008261.hg.1 ENST00000306384.4 Ensembl
Affymetrix Human Clariom D Assay TC0200008261.hg.1 ENST00000306384.4 Ensembl
Affymetrix Human Clariom D Assay TC0200008261.hg.1 ENST00000306384.4 Ensembl
Affymetrix Human Clariom D Assay TC0200008261.hg.1 ENST00000462451.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008261.hg.1 ENST00000462451.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008261.hg.1 ENST00000462451.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008261.hg.1 ENST00000462451.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008261.hg.1 NM_006634.2 Refseq
Affymetrix Human Clariom D Assay TC0200008261.hg.1 NM_006634.2 Refseq
Affymetrix Human Clariom D Assay TC0200008261.hg.1 NM_006634.2 Refseq
Affymetrix Human Clariom D Assay TC0200008261.hg.1 NM_006634.2 Refseq
Affymetrix Human Clariom D Assay TC0200008261.hg.1 NM_006634.2 Refseq
Affymetrix Human Clariom D Assay PSR0200157232.hg.1 ENST00000306384.4 Ensembl
Affymetrix Human Clariom D Assay PSR0200157232.hg.1 NM_006634.2 Refseq
Affymetrix Human Clariom D Assay PSR0200157232.hg.1 ENST00000306384.4 Ensembl
Affymetrix Human Clariom D Assay PSR0200157232.hg.1 NM_006634.2 Refseq
Affymetrix Human Clariom D Assay PSR0200157232.hg.1 TC0200008261.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0200157235.hg.1 ENST00000306384.4 Ensembl
Affymetrix Human Clariom D Assay PSR0200157235.hg.1 ENST00000462451.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200157235.hg.1 NM_006634.2 Refseq
Affymetrix Human Clariom D Assay PSR0200157235.hg.1 ENST00000306384.4 Ensembl
Affymetrix Human Clariom D Assay PSR0200157235.hg.1 ENST00000462451.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200157235.hg.1 NM_006634.2 Refseq
Affymetrix Human Clariom D Assay PSR0200157235.hg.1 TC0200008261.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0