This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00196206_m1
See other GZMA GE Assays ›
Gene Symbol:
GZMA
Gene Name:
granzyme A
Gene Aliases:
CTLA3, HFSP
Chromosome Location:
Chr.5: 55102646 - 55110252 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00196206_m1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information
  • Citations (1)

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GZMA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_006144.3 NP_006135.1 1 - 2 102 101
GenBank mRNA BC015739.1 - 1 - 2 73 4849202 101
BP330335.1 - 1 - 2 102 101
CR456968.1 - 1 - 2 65 101
M18737.1 - 1 - 2 65 101

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0500007413.hg.1
This assay targets the same location as HTA transcript cluster(s) TC05000231.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Protease Immune Tolerance Network Mammalian Gene Collection Homo sapiens Inventoried MicroArray Quality Control (MAQC) Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GZMA

Panther Classification:

Molecular Function -

protease serine protease protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

apoptotic process
immune response
cytolysis
negative regulation of endodeoxyribonuclease activity
positive regulation of apoptotic process
negative regulation of DNA binding
negative regulation of oxidoreductase activity
proteolysis involved in cellular protein catabolic process
serine-type endopeptidase activity
protein binding
protein homodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0500007413.hg.1 ENST00000274306.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007413.hg.1 ENST00000274306.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007413.hg.1 ENST00000274306.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007413.hg.1 ENST00000274306.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007413.hg.1 ENST00000274306.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007413.hg.1 ENST00000274306.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007413.hg.1 ENST00000274306.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007413.hg.1 ENST00000274306.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007413.hg.1 NM_006144.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007413.hg.1 NM_006144.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007413.hg.1 NM_006144.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007413.hg.1 NM_006144.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007413.hg.1 NM_006144.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007413.hg.1 NM_006144.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007413.hg.1 NM_006144.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007413.hg.1 NM_006144.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05002810.hg.1 ENST00000274306.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05002810.hg.1 NM_006144.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05002810.hg.1 TC05000231.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR05002808.hg.1 ENST00000274306.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05002808.hg.1 NM_006144.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05002808.hg.1 TC05000231.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR05002809.hg.1 ENST00000274306.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05002809.hg.1 NM_006144.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05002809.hg.1 TC05000231.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0500150056.hg.1 ENST00000274306.6 Ensembl
Affymetrix Human Clariom D Assay PSR0500150056.hg.1 NM_006144.3 Refseq
Affymetrix Human Clariom D Assay PSR0500150056.hg.1 ENST00000274306.6 Ensembl
Affymetrix Human Clariom D Assay PSR0500150056.hg.1 NM_006144.3 Refseq
Affymetrix Human Clariom D Assay PSR0500150056.hg.1 TC0500007413.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0500150050.hg.1 ENST00000274306.6 Ensembl
Affymetrix Human Clariom D Assay PSR0500150050.hg.1 NM_006144.3 Refseq
Affymetrix Human Clariom D Assay PSR0500150050.hg.1 ENST00000274306.6 Ensembl
Affymetrix Human Clariom D Assay PSR0500150050.hg.1 NM_006144.3 Refseq
Affymetrix Human Clariom D Assay PSR0500150050.hg.1 TC0500007413.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0500150052.hg.1 ENST00000274306.6 Ensembl
Affymetrix Human Clariom D Assay PSR0500150052.hg.1 NM_006144.3 Refseq
Affymetrix Human Clariom D Assay PSR0500150052.hg.1 ENST00000274306.6 Ensembl
Affymetrix Human Clariom D Assay PSR0500150052.hg.1 NM_006144.3 Refseq
Affymetrix Human Clariom D Assay PSR0500150052.hg.1 TC0500007413.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0500150055.hg.1 ENST00000274306.6 Ensembl
Affymetrix Human Clariom D Assay PSR0500150055.hg.1 NM_006144.3 Refseq
Affymetrix Human Clariom D Assay PSR0500150055.hg.1 ENST00000274306.6 Ensembl
Affymetrix Human Clariom D Assay PSR0500150055.hg.1 NM_006144.3 Refseq
Affymetrix Human Clariom D Assay PSR0500150055.hg.1 TC0500007413.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0500007413.hg.1 ENST00000274306.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007413.hg.1 ENST00000274306.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007413.hg.1 ENST00000274306.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007413.hg.1 ENST00000274306.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007413.hg.1 ENST00000274306.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007413.hg.1 ENST00000274306.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007413.hg.1 ENST00000274306.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007413.hg.1 ENST00000274306.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007413.hg.1 NM_006144.3 Refseq
Affymetrix Human Clariom D Assay TC0500007413.hg.1 NM_006144.3 Refseq
Affymetrix Human Clariom D Assay TC0500007413.hg.1 NM_006144.3 Refseq
Affymetrix Human Clariom D Assay TC0500007413.hg.1 NM_006144.3 Refseq
Affymetrix Human Clariom D Assay TC0500007413.hg.1 NM_006144.3 Refseq
Affymetrix Human Clariom D Assay TC0500007413.hg.1 NM_006144.3 Refseq
Affymetrix Human Clariom D Assay TC0500007413.hg.1 NM_006144.3 Refseq
Affymetrix Human Clariom D Assay TC0500007413.hg.1 NM_006144.3 Refseq

Back To Top
 

Citations (1)

Comparative analysis of transcriptional profiling of CD3+, CD4+ and CD8+ T cells identifies novel immune response players in T-cell activation.
Authors
Wang M, Windgassen D, Papoutsakis ET
Journal
Bmc Genomics - Page No 225 (Tue Jan 01 00:00:00 UTC 2008)
Volume (Year)
9 (2008)
Product Usage
-

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0