Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043136.hg.1 |
ENST00000370981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043136.hg.1 |
ENST00000395201.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043136.hg.1 |
NM_004675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043136.hg.1 |
TC01002762.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043135.hg.1 |
ENST00000370981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043135.hg.1 |
ENST00000395201.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043135.hg.1 |
NM_004675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043135.hg.1 |
TC01002762.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014528.hg.1 |
ENST00000370981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014528.hg.1 |
ENST00000370981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014528.hg.1 |
ENST00000370981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014528.hg.1 |
ENST00000370981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014528.hg.1 |
ENST00000370981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014528.hg.1 |
ENST00000370981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014528.hg.1 |
ENST00000370981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014528.hg.1 |
ENST00000395201.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014528.hg.1 |
ENST00000395201.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014528.hg.1 |
ENST00000395201.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014528.hg.1 |
ENST00000395201.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014528.hg.1 |
ENST00000395201.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014528.hg.1 |
NM_004675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014528.hg.1 |
NM_004675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014528.hg.1 |
NM_004675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014528.hg.1 |
NM_004675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014528.hg.1 |
NM_004675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043137.hg.1 |
ENST00000370981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043137.hg.1 |
ENST00000395201.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043137.hg.1 |
NM_004675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043137.hg.1 |
TC01002762.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043134.hg.1 |
ENST00000370981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043134.hg.1 |
ENST00000395201.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043134.hg.1 |
NM_004675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043134.hg.1 |
TC01002762.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0100208512.hg.1 |
ENST00000370981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100208512.hg.1 |
ENST00000395201.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100208512.hg.1 |
NM_004675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100208512.hg.1 |
ENST00000370981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100208512.hg.1 |
ENST00000395201.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100208512.hg.1 |
NM_004675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100208512.hg.1 |
TC0100014528.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100208513.hg.1 |
ENST00000370981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100208513.hg.1 |
ENST00000395201.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100208513.hg.1 |
NM_004675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100208513.hg.1 |
ENST00000370981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100208513.hg.1 |
ENST00000395201.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100208513.hg.1 |
NM_004675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100208513.hg.1 |
TC0100014528.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100014528.hg.1 |
ENST00000370981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014528.hg.1 |
ENST00000370981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014528.hg.1 |
ENST00000370981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014528.hg.1 |
ENST00000370981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014528.hg.1 |
ENST00000370981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014528.hg.1 |
ENST00000370981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014528.hg.1 |
ENST00000370981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014528.hg.1 |
ENST00000395201.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014528.hg.1 |
ENST00000395201.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014528.hg.1 |
ENST00000395201.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014528.hg.1 |
ENST00000395201.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014528.hg.1 |
ENST00000395201.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014528.hg.1 |
NM_004675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014528.hg.1 |
NM_004675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014528.hg.1 |
NM_004675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014528.hg.1 |
NM_004675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014528.hg.1 |
NM_004675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100208510.hg.1 |
ENST00000370981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100208510.hg.1 |
ENST00000395201.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100208510.hg.1 |
NM_004675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100208510.hg.1 |
ENST00000370981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100208510.hg.1 |
ENST00000395201.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100208510.hg.1 |
NM_004675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100208510.hg.1 |
TC0100014528.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100208511.hg.1 |
ENST00000370981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100208511.hg.1 |
ENST00000395201.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100208511.hg.1 |
NM_004675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100208511.hg.1 |
ENST00000370981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100208511.hg.1 |
ENST00000395201.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100208511.hg.1 |
NM_004675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100208511.hg.1 |
TC0100014528.hg.1 |
Clariom D TC |