This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00180702_m1
See other AGR2 GE Assays ›
Gene Symbol:
AGR2
Gene Name:
anterior gradient 2, protein disulphide isomerase family member
Gene Aliases:
AG2, GOB-4, HAG-2, HEL-S-116, PDIA17, XAG-2
Chromosome Location:
Chr.7: 16791811 - 16805114 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00180702_m1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AGR2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_006408.3 NP_006399.1 6 - 7 583 107
XM_005249581.4 XP_005249638.1 6 - 7 552 107
GenBank mRNA AF007791.1 - 5 - 6 406 107
AF038451.1 - 6 - 7 454 107
AF087879.1 - 6 - 7 545 107
AF088867.1 - 6 - 7 543 107
AF115926.1 - 6 - 7 453 107
AK223067.1 - 6 - 7 474 107
AK312942.1 - 6 - 7 464 107
AY359009.1 - 6 - 7 543 107
BC015503.1 - 6 - 7 452 3852448 107
BT007048.1 - 5 - 6 396 107
FJ224307.1 - 5 - 6 406 107
KJ767789.1 - 6 - 7 557 107
KJ767790.1 - 3 - 4 175 107

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0700010355.hg.1
This assay targets the same location as HTA transcript cluster(s) TC07001163.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene AGR2

Panther Classification:

Molecular Function -

reductase

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of gene expression
positive regulation of cell-substrate adhesion
response to endoplasmic reticulum stress
positive regulation of epidermal growth factor receptor signaling pathway
digestive tract morphogenesis
positive regulation of developmental growth
lung goblet cell differentiation
negative regulation of cell death
mucus secretion
positive regulation of establishment of protein localization to plasma membrane
positive regulation of IRE1-mediated unfolded protein response
positive regulation of PERK-mediated unfolded protein response
dystroglycan binding
epidermal growth factor receptor binding
protein binding
protein homodimerization activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR07018195.hg.1 ENST00000401412.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07018195.hg.1 ENST00000419304.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07018195.hg.1 ENST00000419572.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07018195.hg.1 NM_006408.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07018195.hg.1 TC07001163.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000401412.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000401412.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000401412.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000401412.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000401412.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000401412.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000401412.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000401412.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000401412.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000412973.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000412973.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000412973.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000412973.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000412973.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000412973.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000412973.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000450569.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000450569.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000450569.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000450569.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000450569.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000450569.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000450569.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000450569.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000468419.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000468419.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000468419.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000468419.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000468419.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000486219.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000486219.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000489523.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000489523.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000489523.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000489523.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000489523.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 ENST00000489523.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000401412.5 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000401412.5 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000401412.5 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000401412.5 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000401412.5 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000401412.5 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000401412.5 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000401412.5 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000401412.5 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000412973.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000412973.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000412973.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000412973.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000412973.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000412973.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000412973.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000450569.5 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000450569.5 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000450569.5 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000450569.5 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000450569.5 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000450569.5 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000450569.5 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000450569.5 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000468419.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000468419.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000468419.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000468419.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000468419.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000486219.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000486219.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000489523.5 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000489523.5 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000489523.5 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000489523.5 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000489523.5 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 ENST00000489523.5 Ensembl
Affymetrix Human Clariom D Assay TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Clariom D Assay TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Clariom D Assay TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Clariom D Assay TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Clariom D Assay TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Clariom D Assay TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Clariom D Assay TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Clariom D Assay TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Clariom D Assay TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Clariom D Assay TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Clariom D Assay TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Clariom D Assay TC0700010355.hg.1 NM_006408.3 Refseq
Affymetrix Human Clariom D Assay PSR0700173608.hg.1 ENST00000401412.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700173608.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700173608.hg.1 ENST00000450569.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700173608.hg.1 NM_006408.3 Refseq
Affymetrix Human Clariom D Assay PSR0700173608.hg.1 ENST00000401412.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700173608.hg.1 ENST00000419304.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700173608.hg.1 ENST00000450569.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700173608.hg.1 NM_006408.3 Refseq
Affymetrix Human Clariom D Assay PSR0700173608.hg.1 TC0700010355.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 195456 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0