This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00175072_m1
See other NR4A3 GE Assays ›
Gene Symbol:
NR4A3
Gene Name:
nuclear receptor subfamily 4 group A member 3
Gene Aliases:
CHN, CSMF, MINOR, NOR1, TEC
Chromosome Location:
Chr.9: 99821855 - 99866893 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00175072_m1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NR4A3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_006981.3 NP_008912.2 2 - 3 730 60
NM_173199.2 NP_775291.1 2 - 3 730 60
GenBank mRNA AK226108.1 - 2 - 3 712 60
D78579.1 - 2 - 3 732 60
S81243.1 - 2 - 3 617 60
X89894.1 - 2 - 3 688 60

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0900008219.hg.1
This assay targets the same location as HTA transcript cluster(s) TC09000508.hg.1

Additional Information:

This assay is in the 5'UTR region of the D78579.1,NM_006981.3,NM_173199.2,X89894.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Nuclear Receptor Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NR4A3

Panther Classification:

Molecular Function -

C4 zinc finger nuclear receptor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
mesoderm formation
transcription initiation from RNA polymerase II promoter
gastrulation
axon guidance
biological_process
pyruvate oxidation
positive regulation of cardiac muscle hypertrophy
positive regulation of glucose transport
hippocampus development
intracellular receptor signaling pathway
adult behavior
organ regeneration
positive regulation of mast cell cytokine production
common myeloid progenitor cell proliferation
positive regulation of mast cell activation by Fc-epsilon receptor signaling pathway
response to hydrogen peroxide
mast cell degranulation
steroid hormone mediated signaling pathway
negative regulation of neuron apoptotic process
cellular response to leptin stimulus
cellular respiration
fat cell differentiation
positive regulation of cell cycle
positive regulation of transcription from RNA polymerase II promoter
positive regulation of fatty acid oxidation
regulation of smooth muscle cell proliferation
positive regulation of smooth muscle cell proliferation
semicircular canal morphogenesis
positive regulation of epithelial cell proliferation
neuromuscular process controlling balance
vestibular reflex
regulation of type B pancreatic cell proliferation
cellular response to corticotropin-releasing hormone stimulus
cellular response to catecholamine stimulus
energy homeostasis
positive regulation of monocyte aggregation
negative regulation of hydrogen peroxide-induced neuron death
positive regulation of leukocyte apoptotic process
positive regulation of feeding behavior
regulation of energy homeostasis
RNA polymerase II core promoter proximal region sequence-specific DNA binding
core promoter sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription coactivator binding
DNA binding
steroid hormone receptor activity
thyroid hormone receptor activity
protein binding
zinc ion binding
protein kinase binding
histone acetyltransferase binding
glucocorticoid receptor binding
protein homodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR09005513.hg.1 ENST00000330847.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09005513.hg.1 ENST00000338488.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09005513.hg.1 ENST00000395096.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09005513.hg.1 ENST00000395097.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09005513.hg.1 NM_006981.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09005513.hg.1 NM_173199.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09005513.hg.1 NM_173200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09005513.hg.1 TC09000508.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000330847.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000330847.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000330847.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000330847.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000330847.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000330847.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000330847.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000330847.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000330847.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000330847.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000338488.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000338488.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000338488.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000338488.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000338488.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000338488.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000338488.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000395097.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000395097.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000395097.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000395097.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000395097.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000395097.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000395097.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000395097.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000395097.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000618101.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000618101.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000618101.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000618101.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000618101.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000618101.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000618101.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000618101.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000618101.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000618101.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000618101.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000618101.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_006981.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_006981.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_006981.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_006981.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_006981.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_006981.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_006981.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_006981.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_006981.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_006981.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_173199.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_173199.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_173199.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_173199.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_173199.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_173199.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_173199.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_173200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_173200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_173200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_173200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_173200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_173200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_173200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_173200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_173200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_173200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_173200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_173200.2 Refseq
Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000330847.1 Ensembl
Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000330847.1 Ensembl
Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000330847.1 Ensembl
Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000330847.1 Ensembl
Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000330847.1 Ensembl
Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000330847.1 Ensembl
Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000330847.1 Ensembl
Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000330847.1 Ensembl
Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000330847.1 Ensembl
Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000330847.1 Ensembl
Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000338488.8 Ensembl
Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000338488.8 Ensembl
Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000338488.8 Ensembl
Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000338488.8 Ensembl
Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000338488.8 Ensembl
Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000338488.8 Ensembl
Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000338488.8 Ensembl
Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000395097.6 Ensembl
Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000395097.6 Ensembl
Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000395097.6 Ensembl
Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000395097.6 Ensembl
Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000395097.6 Ensembl
Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000395097.6 Ensembl
Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000395097.6 Ensembl
Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000395097.6 Ensembl
Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000395097.6 Ensembl
Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000618101.4 Ensembl
Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000618101.4 Ensembl
Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000618101.4 Ensembl
Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000618101.4 Ensembl
Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000618101.4 Ensembl
Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000618101.4 Ensembl
Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000618101.4 Ensembl
Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000618101.4 Ensembl
Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000618101.4 Ensembl
Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000618101.4 Ensembl
Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000618101.4 Ensembl
Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000618101.4 Ensembl
Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_006981.3 Refseq
Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_006981.3 Refseq
Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_006981.3 Refseq
Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_006981.3 Refseq
Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_006981.3 Refseq
Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_006981.3 Refseq
Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_006981.3 Refseq
Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_006981.3 Refseq
Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_006981.3 Refseq
Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_006981.3 Refseq
Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_173199.2 Refseq
Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_173199.2 Refseq
Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_173199.2 Refseq
Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_173199.2 Refseq
Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_173199.2 Refseq
Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_173199.2 Refseq
Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_173199.2 Refseq
Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_173200.2 Refseq
Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_173200.2 Refseq
Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_173200.2 Refseq
Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_173200.2 Refseq
Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_173200.2 Refseq
Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_173200.2 Refseq
Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_173200.2 Refseq
Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_173200.2 Refseq
Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_173200.2 Refseq
Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_173200.2 Refseq
Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_173200.2 Refseq
Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_173200.2 Refseq
Affymetrix Human Clariom D Assay PSR0900155057.hg.1 ENST00000330847.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900155057.hg.1 ENST00000338488.8 Ensembl
Affymetrix Human Clariom D Assay PSR0900155057.hg.1 ENST00000395097.6 Ensembl
Affymetrix Human Clariom D Assay PSR0900155057.hg.1 ENST00000618101.4 Ensembl
Affymetrix Human Clariom D Assay PSR0900155057.hg.1 NM_006981.3 Refseq
Affymetrix Human Clariom D Assay PSR0900155057.hg.1 NM_173199.2 Refseq
Affymetrix Human Clariom D Assay PSR0900155057.hg.1 NM_173200.2 Refseq
Affymetrix Human Clariom D Assay PSR0900155057.hg.1 ENST00000330847.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900155057.hg.1 ENST00000338488.8 Ensembl
Affymetrix Human Clariom D Assay PSR0900155057.hg.1 ENST00000395097.6 Ensembl
Affymetrix Human Clariom D Assay PSR0900155057.hg.1 ENST00000618101.4 Ensembl
Affymetrix Human Clariom D Assay PSR0900155057.hg.1 NM_006981.3 Refseq
Affymetrix Human Clariom D Assay PSR0900155057.hg.1 NM_173199.2 Refseq
Affymetrix Human Clariom D Assay PSR0900155057.hg.1 NM_173200.2 Refseq
Affymetrix Human Clariom D Assay PSR0900155057.hg.1 TC0900008219.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header