This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00174781_m1
See other EDIL3 GE Assays ›
Gene Symbol:
EDIL3
Gene Name:
EGF like repeats and discoidin domains 3
Gene Aliases:
DEL1
Chromosome Location:
Chr.5: 83940595 - 84384867 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00174781_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EDIL3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005711.4 NP_005702.3 2 - 3 691 74
GenBank mRNA AK312858.1 - 2 - 3 208 74
BC030828.1 - 2 - 3 617 4791845 74
U70312.1 - 2 - 3 308 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0500011352.hg.1
This assay targets the same location as HTA transcript cluster(s) TC05001558.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Immune Tolerance Network Mammalian Gene Collection Homo sapiens Human Angiogenesis 1700 Inventoried Human_Angiogenesis Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene EDIL3

Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
multicellular organism development
positive regulation of cell-substrate adhesion
integrin binding
calcium ion binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 ENST00000296591.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 ENST00000296591.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 ENST00000296591.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 ENST00000296591.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 ENST00000296591.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 ENST00000296591.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 ENST00000296591.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 ENST00000296591.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 ENST00000296591.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 ENST00000296591.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 ENST00000296591.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 ENST00000296591.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 ENST00000296591.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 ENST00000296591.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 ENST00000296591.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 ENST00000296591.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 ENST00000296591.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 ENST00000380138.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 ENST00000380138.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 ENST00000380138.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 ENST00000380138.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 ENST00000380138.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 ENST00000380138.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 ENST00000380138.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 ENST00000380138.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 ENST00000380138.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 ENST00000380138.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 ENST00000380138.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 ENST00000380138.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 ENST00000380138.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 ENST00000507663.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 ENST00000507663.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 ENST00000507663.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 ENST00000507663.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 ENST00000510271.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 ENST00000510271.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 ENST00000510271.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 ENST00000510271.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 ENST00000510271.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 NM_001278642.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 NM_001278642.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 NM_001278642.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 NM_001278642.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 NM_001278642.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 NM_001278642.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 NM_001278642.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 NM_001278642.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 NM_001278642.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 NM_001278642.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 NM_001278642.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 NM_001278642.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 NM_001278642.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 NM_001278642.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 NM_001278642.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 NM_001278642.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 NM_005711.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 NM_005711.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 NM_005711.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 NM_005711.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 NM_005711.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 NM_005711.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 NM_005711.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 NM_005711.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 NM_005711.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 NM_005711.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 NM_005711.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 NM_005711.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 NM_005711.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 NM_005711.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 NM_005711.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 NM_005711.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011352.hg.1 NM_005711.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05021272.hg.1 ENST00000296591.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05021272.hg.1 ENST00000380138.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05021272.hg.1 NM_005711.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05021272.hg.1 TC05001558.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0500178611.hg.1 ENST00000296591.9 Ensembl
Affymetrix Human Clariom D Assay PSR0500178611.hg.1 NM_005711.4 Refseq
Affymetrix Human Clariom D Assay PSR0500178611.hg.1 ENST00000296591.9 Ensembl
Affymetrix Human Clariom D Assay PSR0500178611.hg.1 NM_005711.4 Refseq
Affymetrix Human Clariom D Assay PSR0500178611.hg.1 TC0500011352.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0500011352.hg.1 ENST00000296591.9 Ensembl
Affymetrix Human Clariom D Assay TC0500011352.hg.1 ENST00000296591.9 Ensembl
Affymetrix Human Clariom D Assay TC0500011352.hg.1 ENST00000296591.9 Ensembl
Affymetrix Human Clariom D Assay TC0500011352.hg.1 ENST00000296591.9 Ensembl
Affymetrix Human Clariom D Assay TC0500011352.hg.1 ENST00000296591.9 Ensembl
Affymetrix Human Clariom D Assay TC0500011352.hg.1 ENST00000296591.9 Ensembl
Affymetrix Human Clariom D Assay TC0500011352.hg.1 ENST00000296591.9 Ensembl
Affymetrix Human Clariom D Assay TC0500011352.hg.1 ENST00000296591.9 Ensembl
Affymetrix Human Clariom D Assay TC0500011352.hg.1 ENST00000296591.9 Ensembl
Affymetrix Human Clariom D Assay TC0500011352.hg.1 ENST00000296591.9 Ensembl
Affymetrix Human Clariom D Assay TC0500011352.hg.1 ENST00000296591.9 Ensembl
Affymetrix Human Clariom D Assay TC0500011352.hg.1 ENST00000296591.9 Ensembl
Affymetrix Human Clariom D Assay TC0500011352.hg.1 ENST00000296591.9 Ensembl
Affymetrix Human Clariom D Assay TC0500011352.hg.1 ENST00000296591.9 Ensembl
Affymetrix Human Clariom D Assay TC0500011352.hg.1 ENST00000296591.9 Ensembl
Affymetrix Human Clariom D Assay TC0500011352.hg.1 ENST00000296591.9 Ensembl
Affymetrix Human Clariom D Assay TC0500011352.hg.1 ENST00000296591.9 Ensembl
Affymetrix Human Clariom D Assay TC0500011352.hg.1 ENST00000380138.3 Ensembl
Affymetrix Human Clariom D Assay TC0500011352.hg.1 ENST00000380138.3 Ensembl
Affymetrix Human Clariom D Assay TC0500011352.hg.1 ENST00000380138.3 Ensembl
Affymetrix Human Clariom D Assay TC0500011352.hg.1 ENST00000380138.3 Ensembl
Affymetrix Human Clariom D Assay TC0500011352.hg.1 ENST00000380138.3 Ensembl
Affymetrix Human Clariom D Assay TC0500011352.hg.1 ENST00000380138.3 Ensembl
Affymetrix Human Clariom D Assay TC0500011352.hg.1 ENST00000380138.3 Ensembl
Affymetrix Human Clariom D Assay TC0500011352.hg.1 ENST00000380138.3 Ensembl
Affymetrix Human Clariom D Assay TC0500011352.hg.1 ENST00000380138.3 Ensembl
Affymetrix Human Clariom D Assay TC0500011352.hg.1 ENST00000380138.3 Ensembl
Affymetrix Human Clariom D Assay TC0500011352.hg.1 ENST00000380138.3 Ensembl
Affymetrix Human Clariom D Assay TC0500011352.hg.1 ENST00000380138.3 Ensembl
Affymetrix Human Clariom D Assay TC0500011352.hg.1 ENST00000380138.3 Ensembl
Affymetrix Human Clariom D Assay TC0500011352.hg.1 ENST00000507663.1 Ensembl
Affymetrix Human Clariom D Assay TC0500011352.hg.1 ENST00000507663.1 Ensembl
Affymetrix Human Clariom D Assay TC0500011352.hg.1 ENST00000507663.1 Ensembl
Affymetrix Human Clariom D Assay TC0500011352.hg.1 ENST00000507663.1 Ensembl
Affymetrix Human Clariom D Assay TC0500011352.hg.1 ENST00000510271.1 Ensembl
Affymetrix Human Clariom D Assay TC0500011352.hg.1 ENST00000510271.1 Ensembl
Affymetrix Human Clariom D Assay TC0500011352.hg.1 ENST00000510271.1 Ensembl
Affymetrix Human Clariom D Assay TC0500011352.hg.1 ENST00000510271.1 Ensembl
Affymetrix Human Clariom D Assay TC0500011352.hg.1 ENST00000510271.1 Ensembl
Affymetrix Human Clariom D Assay TC0500011352.hg.1 NM_001278642.1 Refseq
Affymetrix Human Clariom D Assay TC0500011352.hg.1 NM_001278642.1 Refseq
Affymetrix Human Clariom D Assay TC0500011352.hg.1 NM_001278642.1 Refseq
Affymetrix Human Clariom D Assay TC0500011352.hg.1 NM_001278642.1 Refseq
Affymetrix Human Clariom D Assay TC0500011352.hg.1 NM_001278642.1 Refseq
Affymetrix Human Clariom D Assay TC0500011352.hg.1 NM_001278642.1 Refseq
Affymetrix Human Clariom D Assay TC0500011352.hg.1 NM_001278642.1 Refseq
Affymetrix Human Clariom D Assay TC0500011352.hg.1 NM_001278642.1 Refseq
Affymetrix Human Clariom D Assay TC0500011352.hg.1 NM_001278642.1 Refseq
Affymetrix Human Clariom D Assay TC0500011352.hg.1 NM_001278642.1 Refseq
Affymetrix Human Clariom D Assay TC0500011352.hg.1 NM_001278642.1 Refseq
Affymetrix Human Clariom D Assay TC0500011352.hg.1 NM_001278642.1 Refseq
Affymetrix Human Clariom D Assay TC0500011352.hg.1 NM_001278642.1 Refseq
Affymetrix Human Clariom D Assay TC0500011352.hg.1 NM_001278642.1 Refseq
Affymetrix Human Clariom D Assay TC0500011352.hg.1 NM_001278642.1 Refseq
Affymetrix Human Clariom D Assay TC0500011352.hg.1 NM_001278642.1 Refseq
Affymetrix Human Clariom D Assay TC0500011352.hg.1 NM_005711.4 Refseq
Affymetrix Human Clariom D Assay TC0500011352.hg.1 NM_005711.4 Refseq
Affymetrix Human Clariom D Assay TC0500011352.hg.1 NM_005711.4 Refseq
Affymetrix Human Clariom D Assay TC0500011352.hg.1 NM_005711.4 Refseq
Affymetrix Human Clariom D Assay TC0500011352.hg.1 NM_005711.4 Refseq
Affymetrix Human Clariom D Assay TC0500011352.hg.1 NM_005711.4 Refseq
Affymetrix Human Clariom D Assay TC0500011352.hg.1 NM_005711.4 Refseq
Affymetrix Human Clariom D Assay TC0500011352.hg.1 NM_005711.4 Refseq
Affymetrix Human Clariom D Assay TC0500011352.hg.1 NM_005711.4 Refseq
Affymetrix Human Clariom D Assay TC0500011352.hg.1 NM_005711.4 Refseq
Affymetrix Human Clariom D Assay TC0500011352.hg.1 NM_005711.4 Refseq
Affymetrix Human Clariom D Assay TC0500011352.hg.1 NM_005711.4 Refseq
Affymetrix Human Clariom D Assay TC0500011352.hg.1 NM_005711.4 Refseq
Affymetrix Human Clariom D Assay TC0500011352.hg.1 NM_005711.4 Refseq
Affymetrix Human Clariom D Assay TC0500011352.hg.1 NM_005711.4 Refseq
Affymetrix Human Clariom D Assay TC0500011352.hg.1 NM_005711.4 Refseq
Affymetrix Human Clariom D Assay TC0500011352.hg.1 NM_005711.4 Refseq
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 139374 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0