This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00171926_m1
See other MRAS GE Assays ›
Gene Symbol:
MRAS
Gene Name:
muscle RAS oncogene homolog
Gene Aliases:
M-RAs, R-RAS3, RRAS3
Chromosome Location:
Chr.3: 138347648 - 138405535 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00171926_m1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MRAS
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001085049.2 NP_001078518.1 4 - 5 566 65
NM_001252090.1 NP_001239019.1 4 - 5 716 65
NM_001252091.1 NP_001239020.1 3 - 4 931 65
NM_001252092.1 NP_001239021.1 3 - 4 436 65
NM_001252093.1 NP_001239022.1 3 - 4 355 65
NM_012219.4 NP_036351.3 4 - 5 1142 65
XM_005247228.1 XP_005247285.1 4 - 5 647 65
XM_017005887.1 XP_016861376.1 4 - 5 702 65
GenBank mRNA AF022080.1 - 3 - 4 446 65
AF043938.1 - 4 - 5 549 65
AF493918.1 - 3 - 4 446 65
AI216578.1 - 3 - 4 327 65
AK124954.1 - 4 - 5 1093 65
AK295640.1 - 3 - 4 436 65
AK316071.1 - 3 - 4 297 65
BC035939.1 - 3 - 4 475 65
BC047690.1 - 4 - 5 559 65
BT020057.1 - 3 - 4 446 65
CR542067.1 - 3 - 4 446 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300008945.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03000744.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Human PDGF Pathway Human Erythropoietin Pathway Human IL-2 Pathway Human NGF Pathway Human EGF Pathway Human FGF Pathway HUMAN 14-3-3 INDUCED INTRACELLULAR SIGNALING HUMAN IL-6 PATHWAY 1700 Inventoried Human_FGF_Pathway Human_EGF_Pathway Human_PDGF_Pathway Human_Erythropoietin_Pathway Human_NGF_Pathway Human_IL-2_Pathway Human_IL-6_Pathway Human_14-3-3_Induced_Intracellular_Signaling Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MRAS

Panther Classification:

Molecular Function -

small GTPase

Gene Ontology Categories:

Function(s) Process(es)

Ras protein signal transduction
multicellular organism development
muscle organ development
actin cytoskeleton organization
GTPase activity
protein binding
GTP binding
GTP-dependent protein binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000289104.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000289104.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000289104.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000289104.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000289104.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000289104.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000289104.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000289104.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000289104.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000289104.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000289104.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000289104.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000423968.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000423968.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000423968.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000423968.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000423968.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000423968.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000423968.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000423968.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000423968.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000423968.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000423968.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000423968.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000461114.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000461114.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000461114.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000461114.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000464896.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000464896.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000464896.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000464896.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000464896.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000464896.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000464896.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000464896.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000464896.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000464896.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000474559.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000474559.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000474559.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000474559.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000474559.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000474559.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000474559.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000474559.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000474559.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000475711.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000475711.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000475711.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000477784.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000477784.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000478647.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000478647.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000494949.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000494949.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000494949.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000494949.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000494949.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000614350.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000614350.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000614350.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000614350.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000614350.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000614350.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000614350.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000614350.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000614350.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000614350.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000614350.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000621127.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000621127.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000621127.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000621127.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000621127.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000621127.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000621127.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000621127.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000621127.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000621127.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 ENST00000621127.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001085049.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001085049.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001085049.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001085049.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001085049.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001085049.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001085049.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001085049.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001085049.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001085049.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001085049.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001085049.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001085049.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001252090.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001252090.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001252090.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001252090.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001252090.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001252090.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001252090.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001252090.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001252090.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001252090.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001252090.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001252090.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001252090.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001252091.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001252091.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001252091.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001252091.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001252091.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001252091.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001252091.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001252091.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001252091.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001252091.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001252091.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001252092.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001252092.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001252092.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001252092.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001252092.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001252092.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001252092.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001252092.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001252092.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001252092.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001252092.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001252093.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001252093.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001252093.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001252093.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001252093.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001252093.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001252093.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001252093.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001252093.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001252093.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_001252093.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_012219.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_012219.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_012219.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_012219.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_012219.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_012219.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_012219.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_012219.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_012219.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_012219.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_012219.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_012219.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008945.hg.1 NM_012219.4 Refseq
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 101584 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0