This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00170143_m1
See other NEO1 GE Assays ›
Gene Symbol:
NEO1
Gene Name:
neogenin 1
Gene Aliases:
IGDCC2, NGN, NTN1R2
Chromosome Location:
Chr.15: 73051715 - 73305206 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00170143_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NEO1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001172623.1 NP_001166094.1 21 - 22 3390 67
NM_002499.3 NP_002490.2 21 - 22 3390 67
XM_005254408.1 XP_005254465.1 22 - 23 3700 67
XM_011521628.2 XP_011519930.1 21 - 22 3455 67
XM_011521630.2 XP_011519932.1 21 - 22 3401 67
XM_011521633.2 XP_011519935.1 21 - 22 3449 67
XM_017022234.1 XP_016877723.1 22 - 23 3444 67
GenBank mRNA BC117161.1 - 21 - 22 3264 40125712 67
BC143270.1 - 21 - 22 3265 9051772 67
U61262.1 - 21 - 22 3334 67
U72391.1 - 21 - 22 3324 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1500007814.hg.1
This assay targets the same location as HTA transcript cluster(s) TC15000664.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Homo sapiens 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NEO1

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
axon guidance
positive regulation of BMP signaling pathway
iron ion homeostasis
receptor activity
co-receptor binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR15005731.hg.1 ENST00000261908.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR15005731.hg.1 ENST00000339362.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR15005731.hg.1 ENST00000379842.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR15005731.hg.1 ENST00000558964.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR15005731.hg.1 ENST00000560262.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR15005731.hg.1 ENST00000560328.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR15005731.hg.1 NM_001172623.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR15005731.hg.1 NM_001172624.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR15005731.hg.1 NM_002499.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR15005731.hg.1 TC15000664.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1500155488.hg.1 ENST00000261908.10 Ensembl
Affymetrix Human Clariom D Assay PSR1500155488.hg.1 ENST00000339362.9 Ensembl
Affymetrix Human Clariom D Assay PSR1500155488.hg.1 ENST00000558964.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500155488.hg.1 ENST00000560262.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500155488.hg.1 ENST00000560328.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500155488.hg.1 NM_001172623.1 Refseq
Affymetrix Human Clariom D Assay PSR1500155488.hg.1 NM_001172624.1 Refseq
Affymetrix Human Clariom D Assay PSR1500155488.hg.1 NM_002499.3 Refseq
Affymetrix Human Clariom D Assay PSR1500155488.hg.1 ENST00000261908.10 Ensembl
Affymetrix Human Clariom D Assay PSR1500155488.hg.1 ENST00000339362.9 Ensembl
Affymetrix Human Clariom D Assay PSR1500155488.hg.1 ENST00000558964.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500155488.hg.1 ENST00000560262.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500155488.hg.1 ENST00000560328.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500155488.hg.1 NM_001172623.1 Refseq
Affymetrix Human Clariom D Assay PSR1500155488.hg.1 NM_001172624.1 Refseq
Affymetrix Human Clariom D Assay PSR1500155488.hg.1 NM_002499.3 Refseq
Affymetrix Human Clariom D Assay PSR1500155488.hg.1 TC1500007814.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 186099 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0