This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00168617_m1
See other PDE3A GE Assays ›
Gene Symbol:
PDE3A
Gene Name:
phosphodiesterase 3A
Gene Aliases:
CGI-PDE, CGI-PDE A, CGI-PDE-A, HTNB
Chromosome Location:
Chr.12: 20369245 - 20684107 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00168617_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PDE3A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000921.4 NP_000912.3 1 - 2 1004 63
XM_017019420.1 XP_016874909.1 1 - 2 985 63
GenBank mRNA AJ005036.1 - 1 - 2 986 63
BC117369.1 - 1 - 2 1004 40125920 63
BC117371.1 - 1 - 2 1004 40125922 63
M91667.2 - 1 - 2 993 63
U36798.1 - 1 - 2 964 63

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1200007037.hg.1
This assay targets the same location as HTA transcript cluster(s) TC12000227.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens HUMAN SPERM MOTILITY 1700 Inventoried Human_Sperm_Motility Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PDE3A

Panther Classification:

Molecular Function -

phosphodiesterase

Gene Ontology Categories:

Function(s) Process(es)

oocyte maturation
cAMP catabolic process
lipid metabolic process
diterpenoid metabolic process
cAMP-mediated signaling
cGMP-mediated signaling
regulation of meiotic nuclear division
response to drug
negative regulation of apoptotic process
negative regulation of vascular permeability
positive regulation of vascular permeability
response to cAMP
positive regulation of oocyte development
cellular response to cGMP
cellular response to transforming growth factor beta stimulus
3',5'-cyclic-AMP phosphodiesterase activity
cGMP-inhibited cyclic-nucleotide phosphodiesterase activity
cAMP binding
metal ion binding
3',5'-cyclic-GMP phosphodiesterase activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR12002753.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12002753.hg.1 NM_000921.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12002753.hg.1 TC12000227.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1200010092.hg.1 ENST00000535755.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010092.hg.1 ENST00000535755.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000542675.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000542675.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000542675.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000544307.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000544307.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000544307.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000544307.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000544307.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000544307.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000544307.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000544307.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000544307.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000544307.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000544307.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000544307.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000544307.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000544307.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000544307.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000544307.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000544307.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_000921.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_000921.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_000921.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_000921.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_000921.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_000921.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_000921.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_000921.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_000921.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_000921.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_000921.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_000921.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_000921.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_000921.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_000921.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_000921.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_000921.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_000921.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_000921.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_000921.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_000921.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_000921.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_000921.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_001244683.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_001244683.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_001244683.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_001244683.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_001244683.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_001244683.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_001244683.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_001244683.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_001244683.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_001244683.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_001244683.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_001244683.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_001244683.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_001244683.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_001244683.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_001244683.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_001244683.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_001244683.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_001244683.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_001244683.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12002754.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12002754.hg.1 NM_000921.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12002754.hg.1 TC12000227.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1200149218.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Clariom D Assay PSR1200149218.hg.1 NM_000921.4 Refseq
Affymetrix Human Clariom D Assay PSR1200149218.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Clariom D Assay PSR1200149218.hg.1 NM_000921.4 Refseq
Affymetrix Human Clariom D Assay PSR1200149218.hg.1 TC1200007037.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1200149219.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Clariom D Assay PSR1200149219.hg.1 ENST00000542675.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200149219.hg.1 NM_000921.4 Refseq
Affymetrix Human Clariom D Assay PSR1200149219.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Clariom D Assay PSR1200149219.hg.1 ENST00000542675.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200149219.hg.1 NM_000921.4 Refseq
Affymetrix Human Clariom D Assay PSR1200149219.hg.1 TC1200007037.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1200010092.hg.1 ENST00000535755.1 Ensembl
Affymetrix Human Clariom D Assay TC1200010092.hg.1 ENST00000535755.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200149220.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Clariom D Assay PSR1200149220.hg.1 ENST00000542675.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200149220.hg.1 NM_000921.4 Refseq
Affymetrix Human Clariom D Assay PSR1200149220.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Clariom D Assay PSR1200149220.hg.1 ENST00000542675.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200149220.hg.1 NM_000921.4 Refseq
Affymetrix Human Clariom D Assay PSR1200149220.hg.1 TC1200007037.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000542675.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000542675.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000542675.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000544307.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000544307.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000544307.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000544307.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000544307.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000544307.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000544307.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000544307.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000544307.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000544307.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000544307.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000544307.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000544307.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000544307.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000544307.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000544307.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000544307.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_000921.4 Refseq
Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_000921.4 Refseq
Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_000921.4 Refseq
Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_000921.4 Refseq
Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_000921.4 Refseq
Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_000921.4 Refseq
Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_000921.4 Refseq
Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_000921.4 Refseq
Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_000921.4 Refseq
Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_000921.4 Refseq
Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_000921.4 Refseq
Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_000921.4 Refseq
Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_000921.4 Refseq
Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_000921.4 Refseq
Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_000921.4 Refseq
Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_000921.4 Refseq
Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_000921.4 Refseq
Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_000921.4 Refseq
Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_000921.4 Refseq
Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_000921.4 Refseq
Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_000921.4 Refseq
Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_000921.4 Refseq
Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_000921.4 Refseq
Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_001244683.1 Refseq
Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_001244683.1 Refseq
Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_001244683.1 Refseq
Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_001244683.1 Refseq
Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_001244683.1 Refseq
Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_001244683.1 Refseq
Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_001244683.1 Refseq
Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_001244683.1 Refseq
Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_001244683.1 Refseq
Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_001244683.1 Refseq
Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_001244683.1 Refseq
Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_001244683.1 Refseq
Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_001244683.1 Refseq
Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_001244683.1 Refseq
Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_001244683.1 Refseq
Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_001244683.1 Refseq
Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_001244683.1 Refseq
Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_001244683.1 Refseq
Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_001244683.1 Refseq
Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_001244683.1 Refseq
Affymetrix Human Clariom D Assay PSR1200149217.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Clariom D Assay PSR1200149217.hg.1 NM_000921.4 Refseq
Affymetrix Human Clariom D Assay PSR1200149217.hg.1 ENST00000359062.3 Ensembl
Affymetrix Human Clariom D Assay PSR1200149217.hg.1 NM_000921.4 Refseq
Affymetrix Human Clariom D Assay PSR1200149217.hg.1 TC1200007037.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 123743 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0