This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00162632_m1
See other GCFC2 GE Assays ›
Gene Symbol:
GCFC2
Gene Name:
GC-rich sequence DNA-binding factor 2
Gene Aliases:
C2orf3, DNABF, GCF, TCF9
Chromosome Location:
Chr.2: 75662655 - 75710986 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00162632_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GCFC2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001201334.1 NP_001188263.1 11 - 12 1584 72
NM_003203.4 NP_003194.3 11 - 12 1816 72
XM_005264520.3 XP_005264577.1 11 - 12 1841 72
XM_011533074.2 XP_011531376.1 10 - 11 1592 72
GenBank mRNA AB026911.1 - 11 - 12 1754 72
AK097564.1 - 11 - 12 1584 72
BC064559.1 - 11 - 12 1713 5548897 72
EF158468.1 - 10 - 11 1630 72
EF158469.1 - 7 - 8 1030 72

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200013137.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02002008.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GCFC2

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
spliceosomal complex assembly
transcription, DNA-templated
regulation of transcription, DNA-templated
negative regulation of transcription, DNA-templated
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000321027.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000321027.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000321027.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000321027.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000321027.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000321027.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000321027.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000321027.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000321027.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000321027.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000321027.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000321027.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000321027.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000321027.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000321027.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000321027.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000321027.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000321027.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000321027.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000321027.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000321027.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000321027.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000321027.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000321027.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000321027.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000321027.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000321027.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000409857.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000409857.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000409857.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000409857.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000409857.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000409857.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000409857.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000409857.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000409857.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000409857.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000409857.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000409857.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000409857.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000409857.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000409857.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000409857.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000409857.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000409857.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000409857.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000409857.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000409857.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000409857.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000409857.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000427862.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000427862.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000427862.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000427862.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000427862.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000427862.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000427862.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000442309.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000442309.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000442309.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000442309.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000442309.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000442309.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000470197.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000470197.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000470197.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000470197.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000470197.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000470197.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000470197.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000470197.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000470197.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000470197.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000470197.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000470197.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000470197.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000470197.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000470197.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000470197.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000470197.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000470197.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000470197.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000470197.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000470285.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000470285.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000470285.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000470503.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000470503.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000470503.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000470503.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000470503.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000470503.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000470503.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000472230.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000472230.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000472230.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000472230.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000472230.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000472230.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000472230.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000472230.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000472230.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000472230.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000472230.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000472230.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000472230.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000472230.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000472230.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000472230.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000472230.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000472230.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000472230.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000472230.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000472230.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000486016.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000486016.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000486016.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000486016.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000486016.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000492826.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000492826.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000492826.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000492826.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000541687.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000541687.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000541687.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000541687.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000541687.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000541687.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000541687.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000541687.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000541687.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000541687.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000541687.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000541687.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000541687.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000541687.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000541687.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000541687.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000541687.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000541687.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000541687.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000541687.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000541687.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000541687.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000541687.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000541687.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 ENST00000541687.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_001201334.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_001201334.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_001201334.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_001201334.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_001201334.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_001201334.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_001201334.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_001201334.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_001201334.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_001201334.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_001201334.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_001201334.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_001201334.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_001201334.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_001201334.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_001201334.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_001201334.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_001201334.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_001201334.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_001201334.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_001201334.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_001201334.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_001201334.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_001201334.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_001201335.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_001201335.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_001201335.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_001201335.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_001201335.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_001201335.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_001201335.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_001201335.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_001201335.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_003203.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_003203.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_003203.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_003203.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_003203.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_003203.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_003203.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_003203.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_003203.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_003203.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_003203.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_003203.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_003203.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_003203.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_003203.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_003203.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_003203.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_003203.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_003203.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_003203.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_003203.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_003203.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_003203.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_003203.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_003203.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_003203.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013137.hg.1 NM_003203.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0