Assay Details
Gene Symbol: Ceacam20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004873123.3 | XP_004873180.1 | 6 - 7 | 1513 | 67 | |
XM_013068644.2 | XP_012924098.1 | 5 - 6 | 1231 | 67 | ||
XM_021250929.1 | XP_021106588.1 | 6 - 7 | 1513 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
