This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05264224_g1
See other RDM1 GE Assays ›
Gene Symbol:
Rdm1
Gene Name:
RAD52 motif containing 1
Gene Aliases:
GW7_15084
Chromosome Location:
Chr.Un|NW_004624795: 2859519 - 2869757 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05264224_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rdm1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004859436.2 XP_004859493.1 1 - 2 187 100
XM_021239998.1 XP_021095657.1 1 - 2 187 100
XM_021239999.1 XP_021095658.1 1 - 2 187 100
XM_021240000.1 XP_021095659.1 1 - 2 188 100
XM_021240001.1 XP_021095660.1 1 - 2 188 100

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0